Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: REWVTQATA LWTANKIVSD YGKDPSITSI LDALDIFLLP VTNPDGYVFS QTKNRMWRKT RSKVSGSLCV GVDPNRNWDA GFGGPGASSN PCSDSYHGPS ANSEVEVKSI VDFIKSHGKV KAFITLHSYS QLLMFPYGYK CTKLDDFDEL SEVAQKAAQS LRSLHGTKYK VGPICSVIYQ ASGGSIDWSY DYGIKYSFAF ELRDTGRYGF LLPARQILPT AEETWLGLKA IMEHVRDHPY (184-419 aa encoded by BC014571)
Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase A2; carboxypeptidase A2 (pancreatic); CBPA2; EC 3.4.17; EC 3.4.17.15
Gene Aliases: CPA2
UniProt ID: (Human) P48052, (Mouse) Q504N0
Entrez Gene ID: (Human) 1358, (Rat) 296959, (Mouse) 232680
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support