Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: RVSEKESQA IQSALDQNKM HYEILIHDLQ EEIEKQFDVK EDIPGRHSYA KYNNWEKIVA WTEKMMDKYP EMVSRIKIGS TVEDNPLYVL KIGEKNERRK AIFMDCGIHA REWVSPAFCQ WFVYQATKTY GRNKIMTKLL DRMNFYILPV FNVDGYIWSW TKNRMWRKNR SKNQNSKCIG TDLNRNFNAS WNSIPNTNDP CADNYRGSAP ESEKETKAVT NFIRSHLNEI KVYITFHSYS QMLLFPYGYT SKLPPNHEDL AKVAKIGTDV LSTRYETRYI YGPIESTIYP ISGSSLDWAY DLGIKHTFAF ELRDKGKFGF LLPESRIKPT CRETMLAVKF IAKYILKHTS (69-417 aa encoded by BC012613)
Three different forms of human pancreatic procarboxypeptidase A have been isolated. This gene encodes a form which is obtained as a binary complex of a procarboxypeptidase A with proproteinase E and functions as a secretory granule metalloexopeptidase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase A3; carboxypeptidase A3 (mast cell); Mast cell carboxypeptidase A; MC-CPA; R-CPA; RMC-CP; tissue carboxypeptidase A
Gene Aliases: CPA3; MC-CPA; mMC-CPA
UniProt ID: (Human) P15088, (Rat) P21961, (Mouse) P15089
Entrez Gene ID: (Human) 1359, (Rat) 54242, (Mouse) 12873
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support