Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: ATGHSYEKYN KWETIEAWTQ QVATENPALI SRSVIGTTFE GRAIYLLKVG KAGQNKPAIF MDCGFHAREW ISPAFCQWFV REAVRTYGRE IQVTELLDKL DFYVLPVLNI DGYIYTWTKS RFWRKTRSTH TGSSCIGTDP NRNFDAGWCE IGASRNPCDE TYCGPAAESE KETKALADFI RNKLSSIKAY LTIHSYSQMM IYPYSYAYKL GENNAELNAL AKATVKELAS LHGTKYTYGP GATTIYPAAG GSDDWAYDQG IRYSFTFELR DTGRYGFLLP ESQIRATCEE TFLAIKYVAS YVLEHLY; Positive Samples: SW620, Mouse pancreas, Mouse liver, Mouse spleen, Rat pancreas
Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase B; carboxypeptidase B1 (tissue); Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; procarboxypeptidase B; protaminase; tissue carboxypeptidase B
Gene Aliases: 0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; CPB; CPB1; PASP; PCPB
UniProt ID: (Human) P15086
Entrez Gene ID: (Human) 1360, (Mouse) 76703, (Rat) 24271
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support