Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Specificity: This antibody binds specificially to the cardiac Troponin I Peptide KISASRKLQLKT. This epitope is 100% conserved among a great variety of mammalian species, including most monkeys, Squirrel, Hamster, Mouse, Rat, Rabbit, Whales, Mole-Rat, Antelope and several other mammals.
Cardiac Troponin I (Cardiac-specific troponin I, TnI, Troponin I) is an integral inhibitory protein in cardiac muscle that exists as part of a complex with troponin C (TnC) and troponin T (TnT). The TnT subunit of troponin binds to tropomyosin-t form, a troponin-tropomyosin complex, anchored in place by the binding of TnI to actin, within muscle thin filaments. Structural change resulting from the binding of calcium to specific sites on the regulatory TnC subunit, releases the inhibitory region of TnI from actin, anabling the attachment of the molecular motor protein myosin, allowing for muscle contraction and hence movement. The measurements of blood TnI and TnT levels is an important diagnostics indicator of heart muscle damage, and can be used to differentiate between angina and myocardial infarction in patients with chest pains. Cardiac Troponin I has also been shown to inhibit angiogenesis in vivo and in vitro. Mutations in the Cardiac Troponin I gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cardiac troponin I; cardiomyopathy, dilated 2A (autosomal recessive); MGC116817; Tpn1; troponin I type 3 (cardiac); Troponin I, cardiac muscle
Gene Aliases: CMD1FF; CMD2A; CMH7; cTnI; RCM1; TNNC1; TNNI3
UniProt ID: (Human) P19429
Entrez Gene ID: (Human) 7137
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support