Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
AA Sequence of recombinant protein: SRREGSLEDP QTDSSVSLLP HLEAKIRQTH SLAHLLTKYA EQLLQEYVQL QGDPFGLPSF SPPRLPVAGL SAPAPSHAGL PVHERLRLDA AALAALPPLL DAVCRRQAEL NPRAPRLLRR LEDAARQARA LGAAVEALLA ALGAANRGPR AEPPAATASA ASATGVFPAK VLGLRVCGLY REWLSRTEGD LGQLLPGGSA.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Cardiotrophin-1. Anti-Human Cardiotrophin-1-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect Human Cardiotrophin-1 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Cardiotrophin-1 (500-P101) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Cardiotrophin-1.
Western Blot: To detect hCardiotrophin-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hCardiotrophin-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P101BT-1MG will be provided as 2 x 500 µg
Cardiotrophin-1 (CT-1) is a member of the IL-6 family of cytokines, which signal through gp130 receptor complexes. gp130 complexes with several different receptor subunits to transmit signals from Cardiotrophin-1, IL-6, LIF, OSM, CNTF and IL-11. Cardiotrophin-1 binds to and activates the leukemia inhibitory factor (LIF) receptor/gp130 receptor complex and has been shown to induce hypertrophy in cardiac myocytes in vitro. Cardiotrophin-1, a secreted protein expressed at high levels in myocardium during cardiogenesis, has been shown to promote proliferation and survival of embryonic cardiomyocytes, suggesting a role for Cardiotrophin-1 in the activation of gp130 during cardiac development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiophin 1; Cardiotrophin; Cardiotrophin-1; Cardiotrophin1; CT 1; CT-1; CT-1CT1; CTF 1
Gene Aliases: CT-1; CT1; CTF1
UniProt ID: (Human) Q16619
Entrez Gene ID: (Human) 1489
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support