Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: KTVLLLADQ MISRIEYIHS KNFIHRDVKP DNFLMGLGKK GNLVYIIDFG LAKKYRDART HQHIPYRENK NLTGTARYAS INTHLGIEQS RRDDLESLGY VLMYFNLGSL PWQGLKAATK RQKYERISEK KMSTPIEVLC KGYPSEFATY LNFCRSLRFD DKPDYSYLRQ LFRNLFHRQG FSYDYVFDWN MLKFGASRAA DDAERERRDR EERLRHSRNP ATRGLPSTAS GRLRGTQEVA PPTPLTPTSH TANTSPRPVS GMERERKVSM RL (103-373 aa encoded by BC003558)
CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway. This pathway is important in both development and cancer. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for CK1 delta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: casein kinase 1; casein kinase 1, delta; Casein kinase I isoform delta; CK1d; CK1delta; CKI-delta; CKId; HGNC:2452; Tau-protein kinase CSNK1D
Gene Aliases: 1200006A05Rik; AA409348; ASPS; CKIdelta; CSNK1D; D930010H05Rik; FASPS2; HCKID
UniProt ID: (Human) P48730, (Rat) Q06486, (Mouse) Q9DC28
Entrez Gene ID: (Human) 1453, (Rat) 64462, (Mouse) 104318
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support