Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AIQIVTPPV DKEAESYQGE EELVSQTDVK TFLEALPQES WQNKHAGSNG NRATNGAPSL VSRGMQGASA NTLNSETSTK RAAVYRMNRN HRGLCVIVNN HSFTSLKDRQ GTHKDAEILS HVFQWLGFTV HIHNNVTKVE MEMVLQKQKC NPAHADGDCF VFCILTHGRF GAVYSSDEAL IPIREIMSHF TALQCPRLAE KPKLFFIQAC QGEEIQPSVS IEADALNPEQ APTSLQDSIP AEADFLLGLA TVPGYVSFRH VEEGSWYIQS LCNHLKKLVP RHEDILSILT AVNDDVSRRV DKQGTKKQMP QPAFTLRKKL VFPVPLDALS L (193-522 aa encoded by BC042844)
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein cleaves and activates caspases 3 and 7, and the protein itself is processed by caspase 8. Mutations in this gene are associated with apoptosis defects seen in type II autoimmune lymphoproliferative syndrome. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptotic protease Mch-4; CASP-10; caspase 10 apoptosis-related cysteine peptidase; caspase 10, apoptosis-related cysteine peptidase; caspase 10, apoptosis-related cysteine protease; Caspase-10; FADD-like ICE2; FAS-associated death domain protein interleukin-1B-converting enzyme 2; FLICE2; ICE-like apoptotic protease 4; interleukin-1B-converting enzyme 2
Gene Aliases: ALPS2; CASP10; FLICE2; MCH4
UniProt ID: (Human) Q92851
Entrez Gene ID: (Human) 843
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support