Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can recognize the 32 kDa pro-caspase 2 as well as 18 kDa cleaved-caspase 2.
Immunogen sequence: GPLCLQVKP CTPEFYQTHF QLAYRLQSRP RGLALVLSNV HFTGEKELEF RSGGDVDHST LVTLFKLLGY DVHVLCDQTA QEMQEKLQNF AQLPAHRVTD SCIVALLSHG VEGAIYGVDG KLLQLQEVFQ LFDNANCPSL QNKPKMFFIQ ACRGDET (170-325 aa encoded by BC002427 )
Caspase 2 gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CASP-2; caspase 2 apoptosis-related cysteine peptidase; Caspase-2; ICH-1L; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; Protease ICH-1; protein phosphatase 1, regulatory subunit 57
Gene Aliases: CASP-2; CASP2; ICH1; NEDD-2; NEDD2; PPP1R57
UniProt ID: (Human) P42575
Entrez Gene ID: (Human) 835
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support