Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAADRGRRI LGVCGMHPHH QETLKKNRVV LAKQLLLSEL LEHLLEKDII TLEMRELIQA KVGSFSQNVE LLNLLPKRGP QAFDAFCEAL RETKQGHLED MLLTTLSGLQ HVLPPLSCDY DLSLPFPVCE SCPLYKKLRL STDTVEHSLD NKDGPLCLQV KPCTPEFYQT HFQLAYRLQS RPRGLALVLS NVHFTGEKEL E (18-217 aa encoded by BC002427)
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CASP-2; caspase 2 apoptosis-related cysteine peptidase; Caspase-2; ICH-1 protease; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; Protease ICH-1; protein phosphatase 1, regulatory subunit 57
Gene Aliases: CASP-2; CASP2; ICH-1; ICH1; NEDD-2; NEDD2; PPP1R57
UniProt ID: (Human) P42575, (Mouse) P29594, (Rat) P55215
Entrez Gene ID: (Human) 835, (Mouse) 12366, (Rat) 64314
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support