Product References
NaAsO2 regulates TLR4/MyD88/NF-κB signaling pathway through DNMT1/SOCS1 to cause apoptosis and inflammation in hepatic BRL-3A cells.
Biological trace element research
Li S,Zhang J,Ma M,Zhang M,Li L,Chen W,Li S
Mon Jan 01 00:00:00 EST 2024
Chlorin e6-induced photodynamic effect facilitates immunogenic cell death of lung cancer as a result of oxidative endoplasmic reticulum stress and DNA damage.
International immunopharmacology
Yu TT,Hu J,Li QR,Peng XC,Xu HZ,Han N,Li LG,Yang XX,Xu X,Yang ZY,Chen H,Chen X,Wang MF,Li TF
Wed Feb 01 00:00:00 EST 2023
Chlorin e6-induced photodynamic effect facilitates immunogenic cell death of lung cancer as a result of oxidative endoplasmic reticulum stress and DNA damage.
International immunopharmacology
Yu TT,Hu J,Li QR,Peng XC,Xu HZ,Han N,Li LG,Yang XX,Xu X,Yang ZY,Chen H,Chen X,Wang MF,Li TF
Wed Feb 01 00:00:00 EST 2023
Chlorin e6-induced photodynamic effect facilitates immunogenic cell death of lung cancer as a result of oxidative endoplasmic reticulum stress and DNA damage.
International immunopharmacology
Yu TT,Hu J,Li QR,Peng XC,Xu HZ,Han N,Li LG,Yang XX,Xu X,Yang ZY,Chen H,Chen X,Wang MF,Li TF
Wed Feb 01 00:00:00 EST 2023
Chlorin e6-induced photodynamic effect facilitates immunogenic cell death of lung cancer as a result of oxidative endoplasmic reticulum stress and DNA damage.
International immunopharmacology
Yu TT,Hu J,Li QR,Peng XC,Xu HZ,Han N,Li LG,Yang XX,Xu X,Yang ZY,Chen H,Chen X,Wang MF,Li TF
Wed Feb 01 00:00:00 EST 2023
JFD, a Novel Natural Inhibitor of Keap1 Alkylation, Suppresses Intracellular Mycobacterium Tuberculosis Growth through Keap1/Nrf2/SOD2-Mediated ROS Accumulation.
Oxidative medicine and cellular longevity
Wan H,Cai Y,Xiao L,Ling Y,Ge L,Mo S,Xie Q,Peng S,Zhou B,Zeng X,Chen X
Tue Jan 23 00:00:00 EST 2024
Insulin ameliorates dim blue light at night-induced apoptosis in hippocampal neurons via the IR/IRS1/AKT/GSK3β/β-catenin signaling pathway.
Ecotoxicology and environmental safety
Liu Q,Wang Z,Cao J,Dong Y,Chen Y
Sun Jan 15 00:00:00 EST 2023
Insulin ameliorates dim blue light at night-induced apoptosis in hippocampal neurons via the IR/IRS1/AKT/GSK3β/β-catenin signaling pathway.
Ecotoxicology and environmental safety
Liu Q,Wang Z,Cao J,Dong Y,Chen Y
Sun Jan 15 00:00:00 EST 2023
Homoplantaginin attenuates high glucose-induced vascular endothelial cell apoptosis through promoting autophagy via the AMPK/TFEB pathway.
Phytotherapy research : PTR
Fan L,Zhang X,Huang Y,Zhang B,Li W,Shi Q,Lin Y,Wu F
Sat Jul 01 00:00:00 EDT 2023
Lipoxin A4 methyl ester attenuated ketamine-induced neurotoxicity in SH-SY5Y cells via regulating leptin pathway.
Toxicology in vitro : an international journal published in association with BIBRA
Zhang R,Wang X,Xie Z,Cao T,Jiang S,Huang L
Thu Jun 01 00:00:00 EDT 2023
KIAA1199 promotes oxaliplatin resistance and epithelial mesenchymal transition of colorectal cancer via protein O-GlcNAcylation.
Translational oncology
Hua Q,Lu Y,Wang D,Da J,Peng W,Sun G,Gu K,Wang H,Zhu Y
Wed Feb 01 00:00:00 EST 2023
Baicalin and baicalein attenuate hyperuricemic nephropathy via inhibiting PI3K/AKT/NF-κB signalling pathway.
Nephrology (Carlton, Vic.)
Liu Z,Xiang H,Deng Q,Fu W,Li Y,Yu Z,Qiu Y,Mei Z,Xu L
Thu Jun 01 00:00:00 EDT 2023
LAMP-2A ablation in hippocampal CA1 astrocytes confers cerebroprotection and ameliorates neuronal injury after global brain ischemia.
Brain pathology (Zurich, Switzerland)
Fu HY,Cui Y,Li Q,Wang D,Li H,Yang L,Wang DJ,Zhou JW
Wed Mar 01 00:00:00 EST 2023
Taxifolin attenuates neuroinflammation and microglial pyroptosis via the PI3K/Akt signaling pathway after spinal cord injury.
International immunopharmacology
Hu Z,Xuan L,Wu T,Jiang N,Liu X,Chang J,Wang T,Han N,Tian X
Sun Jan 01 00:00:00 EST 2023
Novel antioxidant peptides from MFGM protein Hydrolysates: Separation, identification and effects on Dexamethasone-induced mitochondrial dysfunction.
Food chemistry
Li H,Guan K,Liu M,Liu D,Wang W,Zhu A
Wed Mar 01 00:00:00 EST 2023
Novel antioxidant peptides from MFGM protein Hydrolysates: Separation, identification and effects on Dexamethasone-induced mitochondrial dysfunction.
Food chemistry
Li H,Guan K,Liu M,Liu D,Wang W,Zhu A
Wed Mar 01 00:00:00 EST 2023
Lycopene ameliorates islet function and down-regulates the TLR4/MyD88/NF-κB pathway in diabetic mice and Min6 cells.
Food & function
Liu Y,Tian Y,Dai X,Liu T,Zhang Y,Wang S,Shi H,Yin J,Xu T,Zhu R,Zhang Y,Zhao D,Gao S,Wang XD,Wang L,Zhang D
Tue Jun 06 00:00:00 EDT 2023
Nose-to-brain delivery of self-assembled curcumin-lactoferrin nanoparticles: Characterization, neuroprotective effect and in vivo pharmacokinetic study.
Frontiers in bioengineering and biotechnology
Li L,Tan L,Zhang Q,Cheng Y,Liu Y,Li R,Hou S
Sat Apr 15 00:00:00 EDT 2023
Mass spectrometry-based metabolomics reveal Dendrobium huoshanense polysaccharide effects and potential mechanism of N-methyl-N'-nitro-N-nitrosoguanidine -induced damage in GES-1 cells.
Journal of ethnopharmacology
Xie H,Hu M,Yu J,Yang X,Li J,Yu N,Han L,Peng D
Wed Jun 28 00:00:00 EDT 2023
PM2.5 exposure exacerbates mice thoracic aortic aneurysm and dissection by inducing smooth muscle cell apoptosis via the MAPK pathway.
Chemosphere
Ming Y,Zhou X,Liu G,Abudupataer M,Zhu S,Xiang B,Yin X,Lai H,Sun Y,Wang C,Li J,Zhu K
Wed Feb 01 00:00:00 EST 2023
Peptide deregulated in hypertrophic scar-1 alleviates hypertrophic scar fibrosis by targeting focal adhesion kinase and pyruvate kinase M2 and remodeling the metabolic landscape.
International journal of biological macromolecules
Li J,Yin Y,Zhang E,Gui M,Chen L,Li J
Sun Apr 30 00:00:00 EDT 2023
Echinacea Polyphenols Inhibit NLRP3-Dependent Pyroptosis, Apoptosis, and Necroptosis via Suppressing NO Production during Lipopolysaccharide-Induced Acute Lung Injury.
Journal of agricultural and food chemistry
Guo J,Luo Y,Zuo J,Teng J,Shen B,Liu X
Wed May 17 00:00:00 EDT 2023
METTL3 depletion contributes to tumour progression and drug resistance via N6 methyladenosine-dependent mechanism in HR+HER2-breast cancer.
Breast cancer research : BCR
Ouyang D,Hong T,Fu M,Li Y,Zeng L,Chen Q,He H,Wen Y,Cheng Y,Zhou M,Zou Q,Yi W
Fri Feb 10 00:00:00 EST 2023
ZY-444 inhibits the growth and metastasis of prostate cancer by targeting TNFAIP3 through TNF signaling pathway.
American journal of cancer research
Han MT,Pei H,Sun QQ,Wang CL,Li P,Xie YY,Cao LJ,Zhang XX,Sun ZL
Sun May 14 00:00:00 EDT 2023
ZY-444 inhibits the growth and metastasis of prostate cancer by targeting TNFAIP3 through TNF signaling pathway.
American journal of cancer research
Han MT,Pei H,Sun QQ,Wang CL,Li P,Xie YY,Cao LJ,Zhang XX,Sun ZL
Sun May 14 00:00:00 EDT 2023
ZY-444 inhibits the growth and metastasis of prostate cancer by targeting TNFAIP3 through TNF signaling pathway.
American journal of cancer research
Han MT,Pei H,Sun QQ,Wang CL,Li P,Xie YY,Cao LJ,Zhang XX,Sun ZL
Sun May 14 00:00:00 EDT 2023
ZY-444 inhibits the growth and metastasis of prostate cancer by targeting TNFAIP3 through TNF signaling pathway.
American journal of cancer research
Han MT,Pei H,Sun QQ,Wang CL,Li P,Xie YY,Cao LJ,Zhang XX,Sun ZL
Sun May 14 00:00:00 EDT 2023
Inherent Capability of Self-Assembling Nanostructures in Specific Proteasome Activation for Cancer Cell Pyroptosis.
Small (Weinheim an der Bergstrasse, Germany)
Luo QW,Yao L,Li L,Yang Z,Zhao MM,Zheng YZ,Zhuo FF,Liu TT,Zhang XW,Liu D,Tu PF,Zeng KW
Wed Mar 01 00:00:00 EST 2023
Treadmill exercise exerts a synergistic effect with bone marrow mesenchymal stem cell-derived exosomes on neuronal apoptosis and synaptic-axonal remodeling.
Neural regeneration research
Jiang XH,Li HF,Chen ML,Zhang YX,Chen HB,Chen RH,Xiao YC,Liu N
Thu Jun 01 00:00:00 EDT 2023
Ginsenoside Rg3 enhances the radiosensitivity of lung cancer A549 and H1299 cells via the PI3K/AKT signaling pathway.
In vitro cellular & developmental biology. Animal
Li J,Yang B
Sun Jan 01 00:00:00 EST 2023
Roles of TRPV4 in Regulating Circulating Angiogenic Cells to Promote Coronary Microvascular Regeneration.
Journal of cardiovascular translational research
Yang W,Wang H,Guo Q,Xu X,Guo T,Sun L
Sat Apr 01 00:00:00 EDT 2023
Naringin against doxorubicin-induced hepatotoxicity in mice through reducing oxidative stress, inflammation, and apoptosis via the up-regulation of SIRT1.
Environmental toxicology
Xi Y,Chi Z,Tao X,Zhai X,Zhao Z,Ren J,Yang S,Dong D
Mon May 01 00:00:00 EDT 2023
Toosendanin-induced apoptosis of CMT-U27 is mediated through the mitochondrial apoptotic pathway.
Veterinary and comparative oncology
Yang Y,Mei C,Xian H,Zhang X,Li J,Liang ZX,Zhi Y,Ma Y,Wang HJ
Thu Jun 01 00:00:00 EDT 2023
Dexpramipexole ameliorates cognitive deficits in sepsis-associated encephalopathy through suppressing mitochondria-mediated pyroptosis and apoptosis.
Neuroreport
Zhang Y,Fu Q,Ruan J,Shi C,Lu W,Wu J,Zhou Z
Wed Mar 01 00:00:00 EST 2023
Glutamine and amino acid metabolism as a prognostic signature and therapeutic target in endometrial cancer.
Cancer medicine
Zhai L,Yang X,Cheng Y,Wang J
Tue Aug 01 00:00:00 EDT 2023
The pan-cancer analysis of the oncogenic role of FAM72A as a BRCA prognostic biomarker and immunotherapeutic target.
Environmental toxicology
Xu Y,Hirachan S,Shen Y,Huang Q,Bhandari A,Xia E
Mon May 01 00:00:00 EDT 2023
USP39 attenuates the antitumor activity of cisplatin on colon cancer cells dependent on p53.
Cell biology and toxicology
Yuan J,Li X,Zhang Y,Zhang G,Cheng W,Wang W,Lei Y,Song G
Sun Oct 01 00:00:00 EDT 2023
DON entry into the nucleus induces DNA damage, apoptosis and cycle arrest in GES-1 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Hou S,Cheng Y,Wang Z,Xia L,Wang J,Wang H,Sun J,Ma J,Yan Y
Sun Jan 01 00:00:00 EST 2023
Chlorogenic Acid Alleviates Hepatic Ischemia-Reperfusion Injury by Inhibiting Oxidative Stress, Inflammation, and Mitochondria-Mediated Apoptosis In Vivo and In Vitro.
Inflammation
Li K,Feng Z,Wang L,Ma X,Wang L,Liu K,Geng X,Peng C
Thu Jun 01 00:00:00 EDT 2023
XBP1s gene of endoplasmic reticulum stress enhances proliferation and osteogenesis of human periodontal ligament cells.
Tissue & cell
Cui Z,Qin R,Feng J,Liu Y,Zhou X,Qin X,Li Y,Zhang Z,He X
Tue Aug 01 00:00:00 EDT 2023
Liquiritigenin promotes osteogenic differentiation and prevents bone loss via inducing auto-lysosomal degradation and inhibiting apoptosis.
Genes & diseases
Qiu Y,Zhao Y,Long Z,Song A,Huang P,Wang K,Xu L,Molloy DP,He G
Sun Jan 01 00:00:00 EST 2023
Phospholipase A2 induces acute kidney injury by complement mediated mitochondrial apoptosis via TNF-α/NF-κB signaling pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Tang X,Wei T,Guan M,Li P,Pu Y,Cheng L,Zhou Z,Fu P,Zhang L
Wed Feb 01 00:00:00 EST 2023
Hydrogen sulfide regulates SERCA2a SUMOylation by S-Sulfhydration of SENP1 to ameliorate cardiac systole-diastole function in diabetic cardiomyopathy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Peng S,Wang M,Zhang S,Liu N,Li Q,Kang J,Chen L,Li M,Pang K,Huang J,Lu F,Zhao D,Zhang W
Sat Apr 01 00:00:00 EDT 2023
Role of Orai3-Mediated Store-Operated Calcium Entry in Radiation-Induced Brain Microvascular Endothelial Cell Injury.
International journal of molecular sciences
Wu Q,Fang Y,Huang X,Zheng F,Ma S,Zhang X,Han T,Gao H,Shen B
Thu Apr 06 00:00:00 EDT 2023
Cytotoxic steroidal glycosides from the rhizomes of Paris polyphylla var. yunnanensis.
Phytochemistry
Liu Y,Liu MY,Bi LL,Tian YY,Qiu PC,Qian XY,Wang MC,Tang HF,Lu YY,Zhang BL
Wed Mar 01 00:00:00 EST 2023
Alleviative Effect of Threonine on Cadmium-Induced Liver Injury in Mice.
Biological trace element research
Fang Z,Li Y,Wang J,Wang X,Huang L,Sun L,Deng Q
Fri Sep 01 00:00:00 EDT 2023
PHLDA1 is a P53 target gene involved in P53-mediated cell apoptosis.
Molecular and cellular biochemistry
Song X,Zhou L,Yang W,Li X,Ma J,Qi K,Liang R,Li M,Xie L,Su T,Huang D,Liang B
Mon May 08 00:00:00 EDT 2023
Capsella bursa-pastoris (L.) Medic. extract alleviate cataract development by regulating the mitochondrial apoptotic pathway of the lens epithelial cells.
Journal of ethnopharmacology
Xie LK,Xu XJ,Wu X,Wang MJ,Gao CF,Wang DM,Ren SM,Pan YN,Liu XQ
Thu Feb 10 00:00:00 EST 2022
MBD1/HDAC3-miR-5701-FGFR2 axis promotes the development of gastric cancer.
Aging
Zhao C,Miao J,Sun R,Liang R,Chen W,Gao Y,Wang X,Han S,Zhao W,Lei T,Huang C
Fri Jul 22 00:00:00 EDT 2022
Oxidative stress and Cx43-mediated apoptosis are involved in PFOS-induced nephrotoxicity.
Toxicology
Tang L,Yu J,Zhuge S,Chen H,Zhang L,Jiang G
Mon Aug 01 00:00:00 EDT 2022
LS-HB-Mediated Photodynamic Therapy Inhibits Proliferation and Induces Cell Apoptosis in Melanoma.
Molecular pharmaceutics
Wang L,Wang L,Zhang Y,Zhao Z,Liu C,Li M,Liu J,Wang S,Yang D,Luo F,Yan J
Mon Jul 04 00:00:00 EDT 2022
CRISPR screening of E3 ubiquitin ligases reveals Ring Finger Protein 185 as a novel tumor suppressor in glioblastoma repressed by promoter hypermethylation and miR-587.
Journal of translational medicine
Lin K,Shen SH,Lu F,Zheng P,Wu S,Liao J,Jiang X,Zeng G,Wei D
Sat Feb 19 00:00:00 EST 2022
Thioredoxin-Interacting Protein (TXNIP) Knockdown Protects against Sepsis-Induced Brain Injury and Cognitive Decline in Mice by Suppressing Oxidative Stress and Neuroinflammation.
Oxidative medicine and cellular longevity
Zhang Y,Xing CJ,Liu X,Li YH,Jia J,Feng JG,Yang CJ,Chen Y,Zhou J
Thu May 19 00:00:00 EDT 2022
Moderate Aerobic Exercise Regulates Follicular Dysfunction by Initiating Brain-Derived Neurotrophic Factor (BDNF)-Mediated Anti-Apoptotic Signaling Pathways in Polycystic Ovary Syndrome.
Journal of clinical medicine
Zhang Y,Chen D,Wang D,Wang L,Weng Y,Wang H,Wu X,Wang Y
Fri Sep 23 00:00:00 EDT 2022
Multistage targeting and dual inhibiting strategies based on bioengineered tumor matrix microenvironment-mediated protein nanocages for enhancing cancer biotherapy.
Bioengineering & translational medicine
Hu F,Deng C,Zhou Y,Liu Y,Zhang T,Zhang P,Zhao Z,Miao H,Zheng W,Zhang W,Wang M,Ma X
Sun May 01 00:00:00 EDT 2022
HRK inhibits colorectal cancer cells proliferation by suppressing the PI3K/AKT/mTOR pathway.
Frontiers in oncology
Wang H,Chen Y,Yuan Q,Chen L,Dai P,Li X
Tue Jan 03 00:00:00 EST 2023
SS31 Ameliorates Oxidative Stress via the Restoration of Autophagic Flux to Protect Aged Mice From Hind Limb Ischemia.
Frontiers in cardiovascular medicine
Yang Q,Li C,Chen Q
Tue May 03 00:00:00 EDT 2022
Naoluo Xintong Decoction in the Treatment of Ischemic Stroke: A Network Analysis of the Mechanism of Action.
Frontiers in pharmacology
Wang N,Chu F,Fei C,Pan L,Wang Y,Chen W,Peng D,Duan X,He L
Sat Jul 16 00:00:00 EDT 2022
Chinese Medicine, Succinum, Ameliorates Cognitive Impairment of Carotid Artery Ligation Rats and Inhibits Apoptosis of HT22 Hippocampal Cells via Regulation of the GSK3β/β-Catenin Pathway.
Frontiers in pharmacology
Wei C,Zhu Z,Zheng JN,Lu Y,Cao C,Qu S,Liu M,Meng XE,Lou Q,Wang Q,Duan JA,Shang EX,Han Z,Zhu Y
Sat Jul 16 00:00:00 EDT 2022
Tetrandrine alleviates inflammation and neuron apoptosis in experimental traumatic brain injury by regulating the IRE1α/JNK/CHOP signal pathway.
Brain and behavior
Liu H,He S,Li C,Wang J,Zou Q,Liao Y,Chen R
Thu Dec 01 00:00:00 EST 2022
Didymin Ameliorates Liver Fibrosis by Alleviating Endoplasmic Reticulum Stress and Glycerophospholipid Metabolism: Based on Transcriptomics and Metabolomics.
Drug design, development and therapy
Li Y,Li C,Xiong Y,Fang B,Lin X,Huang Q
Wed Jun 15 00:00:00 EDT 2022
Identification and verification of the role of crucial genes through which methionine restriction inhibits the progression of colon cancer cells.
Oncology letters
Zhou L,Chen Z,Liu C
Mon Aug 01 00:00:00 EDT 2022
Sevoflurane preconditioning prevents acute renal injury caused by ischemia-reperfusion in mice via activation of the Nrf2 signaling pathway.
Experimental and therapeutic medicine
Wang WX,Zhao ZR,Bai Y,Li YX,Gao XN,Zhang S,Sun YB
Fri Apr 01 00:00:00 EDT 2022
Sevoflurane preconditioning prevents acute renal injury caused by ischemia-reperfusion in mice via activation of the Nrf2 signaling pathway.
Experimental and therapeutic medicine
Wang WX,Zhao ZR,Bai Y,Li YX,Gao XN,Zhang S,Sun YB
Fri Apr 01 00:00:00 EDT 2022
Protection of pancreatic β-cell by phosphocreatine through mitochondrial improvement via the regulation of dual AKT/IRS-1/GSK-3β and STAT3/Cyp-D signaling pathways.
Cell biology and toxicology
Wang H,Ai J,Shopit A,Niu M,Ahmed N,Tesfaldet T,Tang Z,Li X,Jamalat Y,Chu P,Peng J,Ma X,Qaed E,Han G,Zhang W,Wang J,Tang Z
Wed Jun 01 00:00:00 EDT 2022
Activation of CXCL13/CXCR5 axis aggravates experimental autoimmune cystitis and interstitial cystitis/bladder pain syndrome.
Biochemical pharmacology
Zhao J,Chen S,Yang C,Zhou M,Yang T,Sun B,Zhu J,Zhang H,Lu Q,Li L,Yang Z,Song B,Shen W,Yi S,Dai S
Wed Jun 01 00:00:00 EDT 2022
Exploration of the Effect and Potential Mechanism of Echinacoside Against Endometrial Cancer Based on Network Pharmacology and in vitro Experimental Verification.
Drug design, development and therapy
Shu W,Wang Z,Zhao R,Shi R,Zhang J,Zhang W,Wang H
Fri Jun 24 00:00:00 EDT 2022
Depletion of B7-H4 from C3H10 T1/2 Mesenchymal Stem Cells Attenuates their Immunomodulatory Therapy in Experimental Autoimmune Encephalomyelitis Mice.
Neurotoxicity research
Li H,Sun S,Yin Z,Cao S,Ji X,Duan X,Gao H,Wang M,Fang Q,Dong W,Zhang X,Gu Y,Xue Q
Wed Jun 01 00:00:00 EDT 2022
Acrylonitrile exposure triggers ovarian inflammation and decreases oocyte quality probably via mitochondrial dysfunction induced apoptosis in mice.
Chemico-biological interactions
Luo YS,He QK,Sun MX,Qiao FX,Liu YC,Xu CL,Xu ZR,Zhao SC,Wang HL,Qi ZQ,Liu Y
Wed Jun 01 00:00:00 EDT 2022
Glucocorticoids induce osteonecrosis of the femoral head in rats via PI3K/AKT/FOXO1 signaling pathway.
PeerJ
Sun F,Zhou JL,Wei SX,Jiang ZW,Peng H
Fri Jan 13 00:00:00 EST 2023
Antitumor Effects of Astaxanthin on Esophageal Squamous Cell Carcinoma by up-Regulation of PPARγ.
Nutrition and cancer
Cui L,Li Z,Xu F,Tian Y,Chen T,Li J,Guo Y,Lyu Q
Tue Apr 26 00:00:00 EDT 2022
Protective effect of baicalin against arsenic trioxide-induced acute hepatic injury in mice through JAK2/STAT3 signaling pathway.
International journal of immunopathology and pharmacology
He Q,Sun X,Zhang M,Chu L,Zhao Y,Wu Y,Zhang J,Han X,Guan S,Ding C
Wed Mar 30 00:00:00 EDT 2022
Resveratrol ameliorates iron overload induced liver fibrosis in mice by regulating iron homeostasis.
PeerJ
Wang H,Jiang C,Yang Y,Li J,Wang Y,Wang C,Gao Y
Mon Jan 16 00:00:00 EST 2023
Hernandezine Regulates Proliferation and Autophagy-Induced Apoptosis in Melanoma Cells.
Journal of natural products
Wang X,Li X,Xia Y,Wang D,Li X,Liu L,Zheng Q,Li D,Jiang Q
Fri May 27 00:00:00 EDT 2022
c-Fos is a mechanosensor that regulates inflammatory responses and lung barrier dysfunction during ventilator-induced acute lung injury.
BMC pulmonary medicine
Zhou L,Xue C,Chen Z,Jiang W,He S,Zhang X
Thu Jan 06 00:00:00 EST 2022
Chemical Composition, Antitumor Properties, and Mechanism of the Essential Oil from Plagiomnium acutum T. Kop.
International journal of molecular sciences
Li M,Wang L,Li S,Hua C,Gao H,Ning D,Li C,Zhang C,Jiang F
Sat Nov 26 00:00:00 EST 2022
Dominant negative TGF-β receptor type II in T lymphocytes promotes anti-tumor immunity by modulating T cell subsets and enhancing CTL responses.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li H,Guan Y,Han C,Zhang Y,Chen Y,Jiang L,Zhang P,Chen X,Wei W,Ma Y
Fri Apr 01 00:00:00 EDT 2022
Dandelion Seed Extract Affects Tumor Progression and Enhances the Sensitivity of Cisplatin in Esophageal Squamous Cell Carcinoma.
Frontiers in pharmacology
Li Y,Deng Y,Zhang X,Fu H,Han X,Guo W,Zhao W,Zhao X,Yu C,Li H,Lei K,Wang T
Sat Jul 16 00:00:00 EDT 2022
OLFM4 depletion sensitizes gallbladder cancer cells to cisplatin through the ARL6IP1/caspase-3 axis.
Translational oncology
Lin Z,Yang S,Zhou Y,Hou Z,Li L,Meng M,Ge C,Zeng B,Lai J,Gao H,Zhao Y,Xie Y,He S,Tang W,Li R,Tan J,Wang W
Tue Feb 01 00:00:00 EST 2022
IL-18BP Improves Early Graft Function and Survival in Lewis-Brown Norway Rat Orthotopic Liver Transplantation Model.
Biomolecules
Meng Q,Wu W,Zhang W,Yuan J,Yang L,Zhang X,Tao K
Thu Dec 01 00:00:00 EST 2022
MiR-9-5p Inhibits the MMP+-Induced Neuron Apoptosis through Regulating SCRIB/β-Catenin Signaling in Parkinson's Disease.
Oxidative medicine and cellular longevity
Xiao Z,Yan Z,Sun X,Zhu Z,Wang B,Gao M,Lu F,Liu J,Zong Z,Zhang H,Guo Y
Fri May 06 00:00:00 EDT 2022
BNC1 deficiency-triggered ferroptosis through the NF2-YAP pathway induces primary ovarian insufficiency.
Nature communications
Wang F,Liu Y,Ni F,Jin J,Wu Y,Huang Y,Ye X,Shen X,Ying Y,Chen J,Chen R,Zhang Y,Sun X,Wang S,Xu X,Chen C,Guo J,Zhang D
Wed Oct 05 00:00:00 EDT 2022
BNC1 deficiency-triggered ferroptosis through the NF2-YAP pathway induces primary ovarian insufficiency.
Nature communications
Wang F,Liu Y,Ni F,Jin J,Wu Y,Huang Y,Ye X,Shen X,Ying Y,Chen J,Chen R,Zhang Y,Sun X,Wang S,Xu X,Chen C,Guo J,Zhang D
Wed Oct 05 00:00:00 EDT 2022
Inhibition of PDK1 enhances radiosensitivity and reverses epithelial-mesenchymal transition in nasopharyngeal carcinoma.
Head & neck
Zhang W,Li L,Guo E,Zhou H,Ming J,Sun L,Hu G,Zhang L
Fri Jul 01 00:00:00 EDT 2022
The role of ER stress and ATP/AMPK in oxidative stress meditated hepatotoxicity induced by citrinin.
Ecotoxicology and environmental safety
Wu J,Yang C,Yang M,Liang Z,Wu Y,Kong X,Fan H,Wang S,Ning C,Xiao W,Jin Y,Yi J,Yuan Z
Wed Jun 01 00:00:00 EDT 2022
Proanthocyanidins inhibit the apoptosis and aging of nucleus pulposus cells through the PI3K/Akt pathway delaying intervertebral disc degeneration.
Connective tissue research
Chen HW,Liu MQ,Zhang GZ,Zhang CY,Wang ZH,Lin AX,Kang JH,Liu WZ,Guo XD,Wang YD,Kang XW
Tue Nov 01 00:00:00 EDT 2022
Nephroprotective effect of magnesium isoglycyrrhizinate against arsenic trioxide-induced acute kidney damage in mice.
Experimental and therapeutic medicine
Wei Z,Sun X,He Q,Zhao Y,Wu Y,Han X,Wu Z,Chu X,Guan S
Fri Apr 01 00:00:00 EDT 2022
Gender Differential Expression of AR/miR-21 Signaling Axis and Its Protective Effect on Renal Ischemia-Reperfusion Injury.
Frontiers in cell and developmental biology
Huang G,Yao Q,Ye Z,Huang Y,Zhang C,Jiang Y,Xi X
Wed Nov 01 00:00:00 EDT 2023
Yiqi Huoxue Tongluo recipe regulates NR4A1 to improve renal mitochondrial function in unilateral ureteral obstruction (UUO) rats.
Pharmaceutical biology
He Z,Zhang M,Xu H,Zhou W,Xu C,Wang Z,He M,Wang X
Thu Dec 01 00:00:00 EST 2022
Yiqi Huoxue Tongluo recipe regulates NR4A1 to improve renal mitochondrial function in unilateral ureteral obstruction (UUO) rats.
Pharmaceutical biology
He Z,Zhang M,Xu H,Zhou W,Xu C,Wang Z,He M,Wang X
Thu Dec 01 00:00:00 EST 2022
Yiqi Huoxue Tongluo recipe regulates NR4A1 to improve renal mitochondrial function in unilateral ureteral obstruction (UUO) rats.
Pharmaceutical biology
He Z,Zhang M,Xu H,Zhou W,Xu C,Wang Z,He M,Wang X
Thu Dec 01 00:00:00 EST 2022
Yiqi Huoxue Tongluo recipe regulates NR4A1 to improve renal mitochondrial function in unilateral ureteral obstruction (UUO) rats.
Pharmaceutical biology
He Z,Zhang M,Xu H,Zhou W,Xu C,Wang Z,He M,Wang X
Thu Dec 01 00:00:00 EST 2022
The LPA-CDK5-tau pathway mediates neuronal injury in an in vitro model of ischemia-reperfusion insult.
BMC neurology
Wang Y,Zhang J,Huang L,Mo Y,Wang C,Li Y,Zhang Y,Zhang Z
Mon May 02 00:00:00 EDT 2022
Derepression of the USP22-FASN axis by p53 loss under oxidative stress drives lipogenesis and tumorigenesis.
Cell death discovery
Han Z,Liu M,Xie Y,Zeng K,Zhan Z,Chen Y,Wang L,Chen X,Luo Y,Zeng Y,Zhan H,Lin Y,Zhang K,Zhu X,Liu S,Luo X,Zhou A
Fri Nov 04 00:00:00 EDT 2022
Role of Nrf2 Nucleus Translocation in Beauvericin-Induced Cell Damage in Rat Hepatocytes.
Toxins
Shi J,Wang Y,Xu W,Cai G,Zou H,Yuan Y,Gu J,Liu Z,Bian J
Wed May 25 00:00:00 EDT 2022
Sevelamer arsenite nanoparticle as a Pi-responsive drug carrier and embolic agent for chemoembolization.
Drug delivery
Bi QC,Tang JJ,Zhao J,Lv YF,Deng ZQ,Chen H,Xu YH,Xie CS,Liang QR,Luo RG,Tang Q
Thu Dec 01 00:00:00 EST 2022
Cold Exposure Induces Intestinal Barrier Damage and Endoplasmic Reticulum Stress in the Colon via the SIRT1/Nrf2 Signaling Pathway.
Frontiers in physiology
Guo J,Hu H,Chen Z,Xu J,Nie J,Lu J,Ma L,Ji H,Yuan J,Xu B
Sat Jul 16 00:00:00 EDT 2022
Baicalein Ameliorates Myocardial Ischemia Through Reduction of Oxidative Stress, Inflammation and Apoptosis via TLR4/MyD88/MAPKS/NF-κB Pathway and Regulation of Ca2+ Homeostasis by L-type Ca2+ Channels.
Frontiers in pharmacology
Li J,Yang Y,Wang H,Ma D,Wang H,Chu L,Zhang Y,Gao Y
Tue Apr 05 00:00:00 EDT 2022
A bavachinin analog, D36, induces cell death by targeting both autophagy and apoptosis pathway in acute myeloid leukemia cells.
Cancer chemotherapy and pharmacology
Zhang W,Liu J,Li Y,Guo F
Thu Sep 01 00:00:00 EDT 2022
Enhanced photodynamic therapy/photothermo therapy for nasopharyngeal carcinoma via a tumour microenvironment-responsive self-oxygenated drug delivery system.
Asian journal of pharmaceutical sciences
Wu N,Tu Y,Fan G,Ding J,Luo J,Wang W,Zhang C,Yuan C,Zhang H,Chen P,Tan S,Xiao H
Tue Mar 01 00:00:00 EST 2022
Alnustone inhibits the growth of hepatocellular carcinoma via ROS- mediated PI3K/Akt/mTOR/p70S6K axis.
Phytotherapy research : PTR
Wang L,Cheng L,Ma L,Ahmad Farooqi A,Qiao G,Zhang Y,Ye H,Liu M,Huang J,Yang X,Lin X,Cao S
Sat Jan 01 00:00:00 EST 2022
High expression of 14-3-3¿ indicates poor prognosis and progression of lung adenocarcinoma.
Oncology letters
Feng J,Leng J,Zhao C,Guo J,Chen Y,Li H
Fri Jul 01 00:00:00 EDT 2022
Erectile dysfunction in hypospadiac male adult rats induced by maternal exposure to di-n-butyl phthalate.
Toxicology
Zhou X,Wang S,Zhou R,Zhang T,Wang Y,Zhang Q,Cong R,Ji C,Luan J,Yao L,Zhou X,Song N
Wed Jun 15 00:00:00 EDT 2022
Erectile dysfunction in hypospadiac male adult rats induced by maternal exposure to di-n-butyl phthalate.
Toxicology
Zhou X,Wang S,Zhou R,Zhang T,Wang Y,Zhang Q,Cong R,Ji C,Luan J,Yao L,Zhou X,Song N
Wed Jun 15 00:00:00 EDT 2022
Diosgenin Ameliorated Type II Diabetes-Associated Nonalcoholic Fatty Liver Disease through Inhibiting De Novo Lipogenesis and Improving Fatty Acid Oxidation and Mitochondrial Function in Rats.
Nutrients
Zhong Y,Li Z,Jin R,Yao Y,He S,Lei M,Wang X,Shi C,Gao L,Peng X
Thu Nov 24 00:00:00 EST 2022
Calycosin Improves Intestinal Mucosal Barrier Function after Gastrectomy in Rats through Alleviating Bacterial Translocation, Inflammation, and Oxidative Stress.
Evidence-based complementary and alternative medicine : eCAM
Peng H,Jin L,Zhang Q,Shen Y,Wang Z,Zhou F,Yu Q
Sat Jul 16 00:00:00 EDT 2022
Human breast milk-derived exosomes through inhibiting AT II cell apoptosis to prevent bronchopulmonary dysplasia in rat lung.
Journal of cellular and molecular medicine
Zhou Y,Liu Y,Xu G,Liu L,Li H,Li Y,Yin J,Wang X,Yu Z
Mon Aug 01 00:00:00 EDT 2022
Human breast milk-derived exosomes through inhibiting AT II cell apoptosis to prevent bronchopulmonary dysplasia in rat lung.
Journal of cellular and molecular medicine
Zhou Y,Liu Y,Xu G,Liu L,Li H,Li Y,Yin J,Wang X,Yu Z
Mon Aug 01 00:00:00 EDT 2022
Human breast milk-derived exosomes through inhibiting AT II cell apoptosis to prevent bronchopulmonary dysplasia in rat lung.
Journal of cellular and molecular medicine
Zhou Y,Liu Y,Xu G,Liu L,Li H,Li Y,Yin J,Wang X,Yu Z
Mon Aug 01 00:00:00 EDT 2022
Human breast milk-derived exosomes through inhibiting AT II cell apoptosis to prevent bronchopulmonary dysplasia in rat lung.
Journal of cellular and molecular medicine
Zhou Y,Liu Y,Xu G,Liu L,Li H,Li Y,Yin J,Wang X,Yu Z
Mon Aug 01 00:00:00 EDT 2022
miR-141-3p regulates saturated fatty acid-induced cardiomyocyte apoptosis through Notch1/PTEN/AKT pathway via targeting PSEN1.
Environmental toxicology
Xin X,Duan L,Yang H,Yu H,Bao Y,Jia D,Wu N,Qiao Y
Fri Apr 01 00:00:00 EDT 2022
Prognostic value and biological function of LRRN4 in colorectal cancer.
Cancer cell international
Xu C,Chen Y,Long F,Ye J,Li X,Huang Q,Yao D,Wang X,Zhao J,Meng W,Mo X,Lu R,Fan C,Zhang T
Tue Apr 19 00:00:00 EDT 2022
Blocking PARP activity with the inhibitor veliparib enhances radiotherapy sensitivity in endometrial carcinoma.
Journal of clinical laboratory analysis
Wang J,Xing W,Lin Y,Uskenbayeva N,Yan H,Xu Y,Fang L
Sun May 01 00:00:00 EDT 2022
Exploring the pharmacological mechanism of calculus bovis in cerebral ischaemic stroke using a network pharmacology approach.
Journal of ethnopharmacology
Du X,Li C,Zhang S,Sun C,Zhang X,Chen C,Wang X,Cheng F,Wang Q
Thu Feb 10 00:00:00 EST 2022
Periplaneta americana Extract Pretreatment Alleviates Oxidative Stress and Inflammation and Increases the Abundance of Gut Akkermansia muciniphila in Diquat-Induced Mice.
Antioxidants (Basel, Switzerland)
Lu S,Xu S,Chen L,Deng Y,Feng J
Tue Sep 13 00:00:00 EDT 2022
Inhibition of human peptide deformylase by actinonin sensitizes glioblastoma cells to temozolomide chemotherapy.
Experimental cell research
Lan B,Zhao H,He Y,Zhao Z,Wang N,Gao Y
Tue Nov 15 00:00:00 EST 2022
Hsa_circ_0005050 interacts with ILF3 and affects cell apoptosis and proliferation by disrupting the balance between p53 and p65.
Chemico-biological interactions
Tan J,Sun M,Yin J,Zhou Q,Zhao R,Chen Q,Sun H,Jiang C,Li S,He Y
Thu Dec 01 00:00:00 EST 2022
Mesenchymal stem cells-derived exosomes prevent sepsis-induced myocardial injury by a CircRTN4/miR-497-5p/MG53 pathway.
Biochemical and biophysical research communications
Li J,Jiang R,Hou Y,Lin A
Sat Aug 27 00:00:00 EDT 2022
Hepatocyte-Conditional Knockout of Phosphatidylethanolamine Binding Protein 4 Aggravated LPS/D-GalN-Induced Acute Liver Injury via the TLR4/NF-κB Pathway.
Frontiers in immunology
Qu XQ,Chen QF,Shi QQ,Luo QQ,Zheng SY,Li YH,Bai LY,Gan S,Zhou XY
Tue Jul 26 00:00:00 EDT 2022
Protective Effect of Resveratrol on Immortalized Duck Intestinal Epithelial Cells Exposed to H2O2.
Molecules (Basel, Switzerland)
Zhou N,Tian Y,Wu H,Cao Y,Li R,Zou K,Xu W,Lu L
Tue May 31 00:00:00 EDT 2022
Novel NPR2 Gene Mutations Affect Chondrocytes Function via ER Stress in Short Stature.
Cells
Li Q,Fan X,Lu W,Sun C,Pei Z,Zhang M,Ni J,Wu J,Yu FX,Luo F
Fri Apr 08 00:00:00 EDT 2022
Mechanisms of cinnamic aldehyde against myocardial ischemia/hypoxia injury in vivo and in vitro: Involvement of regulating PI3K/AKT signaling pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zheng B,Qi J,Yang Y,Li L,Liu Y,Han X,Qu W,Chu L
Tue Mar 01 00:00:00 EST 2022
Curcumin Inhibits Hyperandrogen-Induced IRE1α-XBP1 Pathway Activation by Activating the PI3K/AKT Signaling in Ovarian Granulosa Cells of PCOS Model Rats.
Oxidative medicine and cellular longevity
Zhang Y,Wang L,Weng Y,Wang D,Wang R,Wang H,Wang L,Shen S,Wang H,Li Y,Wang Y
Wed Sep 07 00:00:00 EDT 2022
Curcumin Inhibits Hyperandrogen-Induced IRE1α-XBP1 Pathway Activation by Activating the PI3K/AKT Signaling in Ovarian Granulosa Cells of PCOS Model Rats.
Oxidative medicine and cellular longevity
Zhang Y,Wang L,Weng Y,Wang D,Wang R,Wang H,Wang L,Shen S,Wang H,Li Y,Wang Y
Wed Sep 07 00:00:00 EDT 2022
Dihydroartemisinin alleviates deoxynivalenol induced liver apoptosis and inflammation in piglets.
Ecotoxicology and environmental safety
Li J,Bai Y,Ma K,Ren Z,Li J,Zhang J,Shan A
Mon Aug 01 00:00:00 EDT 2022
Mechanism of a new photosensitizer (TBZPy) in the treatment of high-risk human papillomavirus-related diseases.
Photodiagnosis and photodynamic therapy
Li Z,Xiao Z,Feng Y,Wang Q,Teng M
Tue Mar 01 00:00:00 EST 2022
RNA binding motif protein 3 (RBM3) promotes protein kinase B (AKT) activation to enhance glucose metabolism and reduce apoptosis in skeletal muscle of mice under acute cold exposure.
Cell stress & chaperones
Liu Y,Shi H,Hu Y,Yao R,Liu P,Yang Y,Li S
Tue Nov 01 00:00:00 EDT 2022
Design, synthesis, and in vitro protective effect evaluation of α-carboline derivatives against H2O2-induced cardiomyocyte injury.
European journal of medicinal chemistry
Zhang RH,Yang D,Liao XM,Zhang H,Chen GQ,Zhang WL,Wang YC,Wang C,Liao SG,Zhou M,Wang YL,Li YJ
Fri Aug 05 00:00:00 EDT 2022
Protective Effect and Potential Mechanism of the Traditional Chinese Medicine Shaoyao-Gancao Decoction on Ethanol-Induced Gastric Ulcers in Rats.
Evidence-based complementary and alternative medicine : eCAM
Jin Y,Zhang M,Wang Y,Lu Y,Liu T,Yang G,Song S,Liu W
Sat Jul 16 00:00:00 EDT 2022
N-acetylserotonin protects PC12 cells from hydrogen peroxide induced damage through ROS mediated PI3K / AKT pathway.
Cell cycle (Georgetown, Tex.)
Kang J,Wang Y,Guo X,He X,Liu W,Chen H,Wang Z,Lin A,Kang X
Tue Nov 01 00:00:00 EDT 2022
Chlorine disinfection elevates the toxicity of polystyrene microplastics to human cells by inducing mitochondria-dependent apoptosis.
Journal of hazardous materials
Qin J,Xia PF,Yuan XZ,Wang SG
Sat Mar 05 00:00:00 EST 2022
Systemically Silencing Long Non-coding RNAs Maclpil With Short Interfering RNA Nanoparticles Alleviates Experimental Ischemic Stroke by Promoting Macrophage Apoptosis and Anti-inflammatory Activation.
Frontiers in cardiovascular medicine
Wang Y,Liu C,Chen Y,Chen T,Han T,Xue L,Xu B
Tue May 24 00:00:00 EDT 2022
Renoprotective effects of levosimendan on acute kidney injury following cardiac arrest via anti-inflammation, anti-apoptosis, and ERK activation.
FEBS open bio
Tian L,Wang S,Zhao L,Lu X,Zhu C,Gong H,Yang W
Sun Aug 01 00:00:00 EDT 2021
Renoprotective effects of levosimendan on acute kidney injury following cardiac arrest via anti-inflammation, anti-apoptosis, and ERK activation.
FEBS open bio
Tian L,Wang S,Zhao L,Lu X,Zhu C,Gong H,Yang W
Sun Aug 01 00:00:00 EDT 2021
The Potential Protective Effect and Possible Mechanism of Peptides from Oyster (Crassostrea hongkongensis) Hydrolysate on Triptolide-Induced Testis Injury in Male Mice.
Marine drugs
Zhang X,Peng Z,Zheng H,Zhang C,Lin H,Qin X
Sat Oct 09 00:00:00 EDT 2021
Hydroxysafflor yellow A acutely attenuates blood-brain barrier permeability, oxidative stress, inflammation and apoptosis in traumatic brain injury in rats1.
Acta cirurgica brasileira
Xu J,Zhan T,Zheng W,Huang YK,Chen K,Zhang XH,Ren P,Huang X
Fri Jan 29 00:00:00 EST 2021
CircCDK8 regulates osteogenic differentiation and apoptosis of PDLSCs by inducing ER stress/autophagy during hypoxia.
Annals of the New York Academy of Sciences
Zheng J,Zhu X,He Y,Hou S,Liu T,Zhi K,Hou T,Gao L
Mon Feb 01 00:00:00 EST 2021
Wogonin alleviates liver injury in sepsis through Nrf2-mediated NF-κB signalling suppression.
Journal of cellular and molecular medicine
Dai JM,Guo WN,Tan YZ,Niu KW,Zhang JJ,Liu CL,Yang XM,Tao KS,Chen ZN,Dai JY
Tue Jun 01 00:00:00 EDT 2021
Rutaecarpine Inhibits Doxorubicin-Induced Oxidative Stress and Apoptosis by Activating AKT Signaling Pathway.
Frontiers in cardiovascular medicine
Liao ZQ,Jiang YN,Su ZL,Bi HL,Li JT,Li CL,Yang XL,Zhang Y,Xie X
Tue Jan 25 00:00:00 EST 2022
Oxidative stress-mediated apoptosis is involved in bisphenol S-induced reproductive toxicity in male C57BL/6 mice.
Journal of applied toxicology : JAT
Dai W,He QZ,Zhu BQ,Zeng HC
Mon Nov 01 00:00:00 EDT 2021
[8]-Gingerol exerts anti-myocardial ischemic effects in rats via modulation of the MAPK signaling pathway and L-type Ca2+ channels.
Pharmacology research & perspectives
Xue Y,Zhang M,Zheng B,Zhang Y,Chu X,Liu Y,Li Z,Han X,Chu L
Fri Oct 01 00:00:00 EDT 2021
Oxymatrine pretreatment protects H9c2 cardiomyocytes from hypoxia/reoxygenation injury by modulating the PI3K/Akt pathway.
Experimental and therapeutic medicine
Zhang Z,Qin X,Wang Z,Li Y,Chen F,Chen R,Li C,Zhang W,Zhang M
Tue Jun 01 00:00:00 EDT 2021
Aronia melanocarpa polysaccharide ameliorates inflammation and aging in mice by modulating the AMPK/SIRT1/NF-κB signaling pathway and gut microbiota.
Scientific reports
Zhao Y,Liu X,Zheng Y,Liu W,Ding C
Mon Oct 18 00:00:00 EDT 2021
Carnosol alleviates nonalcoholic fatty liver disease by inhibiting mitochondrial dysfunction and apoptosis through targeting of PRDX3.
Toxicology and applied pharmacology
Geng Y,Wang Y,Sun R,Kang X,Zhao H,Zhu M,Sun Y,Hu Y,Wang Z,Tian X,Zhao Y,Yao J
Wed Dec 01 00:00:00 EST 2021
Matrix metalloproteinase 3 regulates angiotensin II‑induced myocardial fibrosis cell viability, migration and apoptosis.
Molecular medicine reports
Shu J,Gu Y,Jin L,Wang H
Mon Feb 01 00:00:00 EST 2021
Prognostic signature composed of transcription factors accurately predicts the prognosis of gastric cancer patients.
Cancer cell international
Zhou L,Chen Z,Wu Y,Lu H,Xin L
Wed Jul 07 00:00:00 EDT 2021
Inhibition of JAK2/STAT3 signaling pathway by panaxadiol limits the progression of pancreatic cancer.
Aging
Fan X,Fu H,Xie N,Guo H,Fu T,Shan Y
Fri Oct 08 00:00:00 EDT 2021
Inhibition of JAK2/STAT3 signaling pathway by panaxadiol limits the progression of pancreatic cancer.
Aging
Fan X,Fu H,Xie N,Guo H,Fu T,Shan Y
Fri Oct 08 00:00:00 EDT 2021
Inhibition of JAK2/STAT3 signaling pathway by panaxadiol limits the progression of pancreatic cancer.
Aging
Fan X,Fu H,Xie N,Guo H,Fu T,Shan Y
Fri Oct 08 00:00:00 EDT 2021
Elevated Serum FGG Levels Prognosticate and Promote the Disease Progression in Prostate Cancer.
Frontiers in genetics
Peng HH,Wang JN,Xiao LF,Yan M,Chen SP,Wang L,Yang K
Tue May 18 00:00:00 EDT 2021
Koumiss promotes Mycobacterium bovis infection by disturbing intestinal flora and inhibiting endoplasmic reticulum stress.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wang H,Hussain T,Yao J,Li J,Sabir N,Liao Y,Liang Z,Wang Y,Liu Y,Zhao D,Zhou X
Wed Sep 01 00:00:00 EDT 2021
A systematic approach to decode the mechanism of Cornus in the treatment of hepatocellular carcinoma (HCC).
European journal of pharmacology
Shen HY,Li XQ,Fan WQ,Wang YW,Huang F,Wu JQ,Zhang W,Feng XS,Chao X
Fri Oct 15 00:00:00 EDT 2021
Salubrinal Exposes Anticancer Properties in Inflammatory Breast Cancer Cells by Manipulating the Endoplasmic Reticulum Stress Pathway.
Frontiers in oncology
Alsterda A,Asha K,Powrozek O,Repak M,Goswami S,Dunn AM,Memmel HC,Sharma-Walia N
Tue Jun 08 00:00:00 EDT 2021
Role of GRK2 in Trophoblast Necroptosis and Spiral Artery Remodeling: Implications for Preeclampsia Pathogenesis.
Frontiers in cell and developmental biology
Lv Z,Xiong LL,Qin X,Zhang H,Luo X,Peng W,Kilby MD,Saffery R,Baker PN,Qi HB
Sat Dec 18 00:00:00 EST 2021
MUSTN1 is an indispensable factor in the proliferation, differentiation and apoptosis of skeletal muscle satellite cells in chicken.
Experimental cell research
Hu Z,Xu H,Lu Y,He Q,Yan C,Zhao X,Tian Y,Yang C,Zhang Z,Qiu M,Wang Y
Fri Oct 15 00:00:00 EDT 2021
Design, synthesis and biological evaluation of novel 1,3,4,9-tetrahydropyrano[3,4-b]indoles as potential treatment of triple negative breast cancer by suppressing PI3K/AKT/mTOR pathway.
Bioorganic & medicinal chemistry
Qin J,Sun X,Ma Y,Cheng Y,Ma Q,Jing W,Qu S,Liu L
Fri Dec 31 00:00:00 EST 2021
Magnesium Isoglycyrrhizinate Alleviates Arsenic Trioxide-Induced Cardiotoxicity: Contribution of Nrf2 and TLR4/NF-κB Signaling Pathway.
Drug design, development and therapy
Zheng B,Yang Y,Li J,Li J,Zuo S,Chu X,Xu S,Ma D,Chu L
Wed Oct 06 00:00:00 EDT 2021
LncRNA COL1A2-AS1 promotes skin fibroblast apoptosis by repressing p-Smad3 and promoting β-catenin expression.
Experimental dermatology
Li J,Gao Y,Li Q,Chen L,Chen Y,Li J
Sun Aug 01 00:00:00 EDT 2021
Protocatechuic aldehyde protects cardiomycoytes against ischemic injury via regulation of nuclear pyruvate kinase M2.
Acta pharmaceutica Sinica. B
Wu X,Liu L,Zheng Q,Hao H,Ye H,Li P,Yang H
Mon Nov 01 00:00:00 EDT 2021
Protocatechuic aldehyde protects cardiomycoytes against ischemic injury via regulation of nuclear pyruvate kinase M2.
Acta pharmaceutica Sinica. B
Wu X,Liu L,Zheng Q,Hao H,Ye H,Li P,Yang H
Mon Nov 01 00:00:00 EDT 2021
Limonin induces apoptosis of HL-60 cells by inhibiting NQO1 activity.
Food science & nutrition
Chen Y,Liang J,Liang X,Chen J,Wang Y,Cao J,Sun C,Ye J,Chen Q
Thu Apr 01 00:00:00 EDT 2021
The miR-623/CXCL12 axis inhibits LPS-induced nucleus pulposus cell apoptosis and senescence.
Mechanisms of ageing and development
Zhong H,Zhou Z,Guo L,Liu F,Zheng B,Bi S,Tian C
Mon Mar 01 00:00:00 EST 2021
Exploration of the hepatoprotective effect and mechanism of magnesium isoglycyrrhizinate in mice with arsenic trioxide‑induced acute liver injury.
Molecular medicine reports
Liu M,Zheng B,Liu P,Zhang J,Chu X,Dong C,Shi J,Liang Y,Chu L,Liu Y,Han X
Tue Jun 01 00:00:00 EDT 2021
Upregulated LINC00319 aggravates neuronal injury induced by oxygen-glucose deprivation via modulating miR-200a-3p.
Experimental and therapeutic medicine
Yang H,Wang H,Zhang X,Yang Y,Li H
Sun Aug 01 00:00:00 EDT 2021
Structural analysis of Panax ginseng glycoproteins and its anti-oligoasthenozoospermia effect in vivo.
International journal of biological macromolecules
Shan M,Zhang X,Fang X,Tian J,Song L,Chen Y,Qiu Z,Zhu D,Luo H,Wang Z
Wed Dec 15 00:00:00 EST 2021
An Endogenous Retroviral LTR-Derived Long Noncoding RNA lnc-LTR5B Interacts With BiP to Modulate ALV-J Replication in Chicken Cells.
Frontiers in microbiology
Chen S,Zhao R,Wu T,Wang D,Wang B,Pan S,Hu X,Pan Z,Cui H
Fri Dec 17 00:00:00 EST 2021
ECHS1, an interacting protein of LASP1, induces sphingolipid-metabolism imbalance to promote colorectal cancer progression by regulating ceramide glycosylation.
Cell death & disease
Li R,Hao Y,Wang Q,Meng Y,Wu K,Liu C,Xu L,Liu Z,Zhao L
Wed Oct 06 00:00:00 EDT 2021
Nephrotoxicity and possible mechanisms of decabrominated diphenyl ethers (BDE-209) exposure to kidney in broilers.
Ecotoxicology and environmental safety
Sun S,Jin Y,Yang J,Zhao Z,Rao Q
Fri Jan 15 00:00:00 EST 2021
BAIAP2L2 promotes the proliferation, migration and invasion of osteosarcoma associated with the Wnt/β-catenin pathway.
Journal of bone oncology
Guo H,Peng J,Hu J,Chang S,Liu H,Luo H,Chen X,Tang H,Chen Y
Wed Dec 01 00:00:00 EST 2021
Hesperetin modulates the Sirt1/Nrf2 signaling pathway in counteracting myocardial ischemia through suppression of oxidative stress, inflammation, and apoptosis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu P,Li J,Liu M,Zhang M,Xue Y,Zhang Y,Han X,Jing X,Chu L
Thu Jul 01 00:00:00 EDT 2021
Harpagide alleviate neuronal apoptosis and blood-brain barrier leakage by inhibiting TLR4/MyD88/NF-κB signaling pathway in Angiotensin II-induced microglial activation in vitro.
Chemico-biological interactions
Lu Y,Hao R,Hu Y,Wei Y,Xie Y,Shen Y,Rui Q,Yu G
Fri Oct 01 00:00:00 EDT 2021
Isosteroidal alkaloids from Fritillaria hupehensis Hsiao et K.C.Hsia: Synthesis and biological evaluation of alkaloid derivatives as potential cytotoxic agents.
Steroids
Liu XH,Zou J,Li YJ,Liu M,He CL,Liu YR,Wang JZ,Chen DL
Wed Dec 01 00:00:00 EST 2021
Tea Polyphenols Prevent Sepsis-Induced Lung Injury via Promoting Translocation of DJ-1 to Mitochondria.
Frontiers in cell and developmental biology
Jia CM,Zhang FW,Wang SJ,Wang W,Li Y
Sat Apr 23 00:00:00 EDT 2022
M2 Macrophage-Derived Exosomal miR-590-3p Attenuates DSS-Induced Mucosal Damage and Promotes Epithelial Repair via the LATS1/YAP/ β-Catenin Signalling Axis.
Journal of Crohn's & colitis
Deng F,Yan J,Lu J,Luo M,Xia P,Liu S,Wang X,Zhi F,Liu D
Tue Apr 06 00:00:00 EDT 2021
Inhibition of autophagy with Chloroquine enhanced apoptosis induced by 5-aminolevulinic acid-photodynamic therapy in secondary hyperparathyroidism primary cells and organoids.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zeng L,Zou Q,Huang P,Xiong L,Cheng Y,Chen Q,Li Y,He H,Yi W,Wei W
Fri Oct 01 00:00:00 EDT 2021
Therapeutic Efficacy of Carbon Ion Irradiation Enhanced by 11-MUA-Capped Gold Nanoparticles: An in vitro and in vivo Study.
International journal of nanomedicine
Zhang P,Yu B,Jin X,Zhao T,Ye F,Liu X,Li P,Zheng X,Chen W,Li Q
Fri Jul 23 00:00:00 EDT 2021
Establishment and Evaluation of a Novel High-Efficiency Model of Graded Traumatic Brain Injury in Mice.
World neurosurgery
Chen B,Shi QX,Nie C,Zhao ZP,Luo L,Zhao QJ,Si SY,Xu BX,Wang T,Gao LY,Gu JW
Fri Oct 01 00:00:00 EDT 2021
Sodium alginate/collagen/stromal cell-derived factor-1 neural scaffold loaded with BMSCs promotes neurological function recovery after traumatic brain injury.
Acta biomaterialia
Ma S,Zhou J,Huang T,Zhang Z,Xing Q,Zhou X,Zhang K,Yao M,Cheng T,Wang X,Wen X,Guan F
Wed Sep 01 00:00:00 EDT 2021
Ketoprofen and Loxoprofen Platinum(IV) Complexes Displaying Antimetastatic Activities by Inducing DNA Damage, Inflammation Suppression, and Enhanced Immune Response.
Journal of medicinal chemistry
Li Z,Wang Q,Li L,Chen Y,Cui J,Liu M,Zhang N,Liu Z,Han J,Wang Z
Thu Dec 23 00:00:00 EST 2021
Cold-Induced RNA-Binding Protein Promotes Glucose Metabolism and Reduces Apoptosis by Increasing AKT Phosphorylation in Mouse Skeletal Muscle Under Acute Cold Exposure.
Frontiers in molecular biosciences
Liu Y,Liu P,Hu Y,Cao Y,Lu J,Yang Y,Lv H,Lian S,Xu B,Li S
Tue Aug 17 00:00:00 EDT 2021
PIM1 inhibitor synergizes the anti-tumor effect of osimertinib via STAT3 dephosphorylation in EGFR-mutant non-small cell lung cancer.
Annals of translational medicine
Sun Z,Zeng L,Zhang M,Zhang Y,Yang N
Sun Mar 01 00:00:00 EST 2020
CCDC57 Cooperates with Microtubules and Microcephaly Protein CEP63 and Regulates Centriole Duplication and Mitotic Progression.
Cell reports
Gurkaslar HK,Culfa E,Arslanhan MD,Lince-Faria M,Firat-Karalar EN
Tue May 12 00:00:00 EDT 2020
Niclosamide: drug repurposing for human chondrosarcoma treatment via the caspase-dependent mitochondrial apoptotic pathway.
American journal of translational research
Yu QS,Xin HR,Qiu RL,Deng ZL,Deng F,Yan ZJ
Mon Sep 28 00:00:00 EDT 2020
A Splice Variant of NCOR2, BQ323636.1, Confers Chemoresistance in Breast Cancer by Altering the Activity of NRF2.
Cancers
Leung MH,Tsoi H,Gong C,Man EP,Zona S,Yao S,Lam EW,Khoo US
Wed Feb 26 00:00:00 EST 2020
Electroacupuncture Inhibits Neuronal Autophagy and Apoptosis via the PI3K/AKT Pathway Following Ischemic Stroke.
Frontiers in cellular neuroscience
Wang MM,Zhang M,Feng YS,Xing Y,Tan ZX,Li WB,Dong F,Zhang F
Mon Sep 28 00:00:00 EDT 2020
2-Methylquinazoline derivative 23BB as a highly selective histone deacetylase 6 inhibitor alleviated cisplatin-induced acute kidney injury.
Bioscience reports
Hao Y,Guo F,Huang Z,Feng Y,Xia Z,Liu J,Li L,Huang R,Lin L,Ma L,Fu P
Fri Jan 31 00:00:00 EST 2020
2-Methylquinazoline derivative 23BB as a highly selective histone deacetylase 6 inhibitor alleviated cisplatin-induced acute kidney injury.
Bioscience reports
Hao Y,Guo F,Huang Z,Feng Y,Xia Z,Liu J,Li L,Huang R,Lin L,Ma L,Fu P
Fri Jan 31 00:00:00 EST 2020
Fucoidan Induces Apoptosis of HT-29 Cells via the Activation of DR4 and Mitochondrial Pathway.
Marine drugs
Bai X,Wang Y,Hu B,Cao Q,Xing M,Song S,Ji A
Mon Apr 20 00:00:00 EDT 2020
Loss of Collapsin Response Mediator Protein 4 Attenuates 6-Hydroxydopamine-Induced Impairments in a Mouse Model of Parkinson's Disease.
Neurochemical research
Li W,Goshima Y,Ohshima T
Thu Oct 01 00:00:00 EDT 2020
Overexpression of Mucin 1 Suppresses the Therapeutical Efficacy of Disulfiram against Canine Mammary Tumor.
Animals : an open access journal from MDPI
Zhao Y,Lin Z,Lin Z,Zhou C,Liu G,Lin J,Zhang D,Lin D
Sun Dec 27 00:00:00 EST 2020
JTE-013 supplementation improves erectile dysfunction in rats with streptozotocin-induced type ¿ diabetes through the inhibition of the rho-kinase pathway, fibrosis, and apoptosis.
Andrology
Liu K,Cui K,Feng H,Li R,Lin H,Chen Y,Zhang Y,Chen Z,Yuan H,Li M,Wang T,Lan R,Liu J,Rao K,Wen B
Sun Mar 01 00:00:00 EST 2020
Intracellular expression of arginine deiminase activates the mitochondrial apoptosis pathway by inhibiting cytosolic ferritin and inducing chromatin autophagy.
BMC cancer
Feng Q,Bian X,Liu X,Wang Y,Zhou H,Ma X,Quan C,Yao Y,Zheng Z
Thu Jul 16 00:00:00 EDT 2020
Activation of PGC-1α and Mitochondrial Biogenesis Protects Against Prenatal Hypoxic-ischemic Brain Injury.
Neuroscience
Jia L,Wang J,Cao H,Zhang X,Rong W,Xu Z
Wed Apr 15 00:00:00 EDT 2020
Fengbaisan suppresses endoplasmic reticulum stress by up-regulating SIRT1 expression to protect rats with chronic obstructive pulmonary diseases.
Pharmaceutical biology
Wang Y,Su NX,Pan SG,Ge XP,Dai XP
Tue Dec 01 00:00:00 EST 2020
Inhibition of Serine Metabolism Promotes Resistance to Cisplatin in Gastric Cancer.
OncoTargets and therapy
Zhao X,Fu J,Tang W,Yu L,Xu W
Fri Apr 15 00:00:00 EDT 2022
Autophagy-inhibiting polymer as an effective nonviral cancer gene therapy vector with inherent apoptosis-sensitizing ability.
Biomaterials
Wang J,Zhou X,Wang H,Xiao Q,Ding K,Dong X,Xu S,Shen B,Sun J,Zhou Z,Tang J,Liu X,Shen Y
Thu Oct 01 00:00:00 EDT 2020
MiR-15b is a key regulator of proliferation and apoptosis of chondrocytes from patients with condylar hyperplasia by targeting IGF1, IGF1R and BCL2.
Osteoarthritis and cartilage
Cao P,Feng Y,Deng M,Li J,Cai H,Meng Q,Fang W,Li Y,Ke J,Long X
Fri Feb 01 00:00:00 EST 2019
MiR-15b is a key regulator of proliferation and apoptosis of chondrocytes from patients with condylar hyperplasia by targeting IGF1, IGF1R and BCL2.
Osteoarthritis and cartilage
Cao P,Feng Y,Deng M,Li J,Cai H,Meng Q,Fang W,Li Y,Ke J,Long X
Fri Feb 01 00:00:00 EST 2019
Silencing TTK expression inhibits the proliferation and progression of prostate cancer.
Experimental cell research
Chen S,Wang J,Wang L,Peng H,Xiao L,Li C,Lin D,Yang K
Sun Dec 01 00:00:00 EST 2019
[JNK/c-Jun signaling pathway mediates arginine vasopressin neuron regeneration by promoting cytoskeleton reconstruction in rats with electrical lesions of the pituitary stalk].
Nan fang yi ke da xue xue bao = Journal of Southern Medical University
Li K,Feng Z,Ou Y,Zhou M,Peng J,Gong H,Wu G,Liu Y,Qi S
Mon Sep 30 00:00:00 EDT 2019
Extracellular RNAs-TLR3 signaling contributes to cognitive decline in a mouse model of postoperative cognitive dysfunction.
Brain, behavior, and immunity
Chen C,Gao R,Li M,Wang Q,Chen H,Zhang S,Mao X,Behensky A,Zhang Z,Gan L,Li T,Liao R,Li Q,Yu H,Yang J,Zhu T,Liu J
Thu Aug 01 00:00:00 EDT 2019
Melatonin protects circulatory death heart from ischemia/reperfusion injury via the JAK2/STAT3 signalling pathway.
Life sciences
Lan H,Su Y,Liu Y,Deng C,Wang J,Chen T,Jules KED,Masau JF,Li H,Wei X
Mon Jul 01 00:00:00 EDT 2019
Knockdown of LncRNA MALAT1 contributes to cell apoptosis via regulating NF-κB/CD80 axis in neonatal respiratory distress syndrome.
The international journal of biochemistry & cell biology
Juan C,Wang Q,Mao Y,Cao Q,Li S,Qiao C,Zhang D,Zhou G
Thu Nov 01 00:00:00 EDT 2018
Knockdown of LncRNA MALAT1 contributes to cell apoptosis via regulating NF-κB/CD80 axis in neonatal respiratory distress syndrome.
The international journal of biochemistry & cell biology
Juan C,Wang Q,Mao Y,Cao Q,Li S,Qiao C,Zhang D,Zhou G
Thu Nov 01 00:00:00 EDT 2018
The Antiapoptosis Effect of Geum japonicum Thunb. var. chinense Extracts on Cerebral Ischemia Reperfusion Injury via PI3K/Akt Pathway.
Evidence-based complementary and alternative medicine : eCAM
Ou B,Tao W,Yang S,Feng J,Wang J,Yang T,Wu H,Huang Y,Tan L,Huang W,Feng Z,Mei Z
Wed Sep 30 00:00:00 EDT 2020
The Antiapoptosis Effect of Geum japonicum Thunb. var. chinense Extracts on Cerebral Ischemia Reperfusion Injury via PI3K/Akt Pathway.
Evidence-based complementary and alternative medicine : eCAM
Ou B,Tao W,Yang S,Feng J,Wang J,Yang T,Wu H,Huang Y,Tan L,Huang W,Feng Z,Mei Z
Wed Sep 30 00:00:00 EDT 2020
The Antiapoptosis Effect of Geum japonicum Thunb. var. chinense Extracts on Cerebral Ischemia Reperfusion Injury via PI3K/Akt Pathway.
Evidence-based complementary and alternative medicine : eCAM
Ou B,Tao W,Yang S,Feng J,Wang J,Yang T,Wu H,Huang Y,Tan L,Huang W,Feng Z,Mei Z
Wed Sep 30 00:00:00 EDT 2020
The Antiapoptosis Effect of Geum japonicum Thunb. var. chinense Extracts on Cerebral Ischemia Reperfusion Injury via PI3K/Akt Pathway.
Evidence-based complementary and alternative medicine : eCAM
Ou B,Tao W,Yang S,Feng J,Wang J,Yang T,Wu H,Huang Y,Tan L,Huang W,Feng Z,Mei Z
Wed Sep 30 00:00:00 EDT 2020
Geniposide attenuates Aβ25-35-induced neurotoxicity via the TLR4/NF-κB pathway in HT22 cells.
RSC advances
Huang XF,Li JJ,Tao YG,Wang XQ,Zhang RL,Zhang JL,Su ZQ,Huang QH,Deng YH
Tue May 22 00:00:00 EDT 2018
MicroRNA‑143 increases cell apoptosis in myelodysplastic syndrome through the Fas/FasL pathway both in vitro and in vivo.
International journal of oncology
Cui J,Wei C,Deng L,Kuang X,Zhang Z,Pierides C,Chi J,Wang L
Thu Nov 01 00:00:00 EDT 2018
UNBS5162 inhibits proliferation of human melanoma cells by inducing apoptosis via the PI3K/Akt pathway.
Molecular medicine reports
Shi X,Yang L,Xie J,Zhao Y,Cong J,Li Z,Li H,Cheng X,Fan J
Sat Sep 01 00:00:00 EDT 2018
Dectin-1 is essential for IL-1β production through JNK activation and apoptosis in Aspergillus fumigatus keratitis.
International immunopharmacology
Yuan K,Zhao G,Che C,Li C,Lin J,Zhu G,He K
Wed Nov 01 00:00:00 EDT 2017
Dectin-1 is essential for IL-1β production through JNK activation and apoptosis in Aspergillus fumigatus keratitis.
International immunopharmacology
Yuan K,Zhao G,Che C,Li C,Lin J,Zhu G,He K
Wed Nov 01 00:00:00 EDT 2017
Induction of autophagy by an oleanolic acid derivative, SZC017, promotes ROS-dependent apoptosis through Akt and JAK2/STAT3 signaling pathway in human lung cancer cells.
Cell biology international
Song Y,Kong L,Sun B,Gao L,Chu P,Ahsan A,Qaed E,Lin Y,Peng J,Ma X,Zhang J,Wang S,Tang Z
Fri Dec 01 00:00:00 EST 2017
Induction of autophagy by an oleanolic acid derivative, SZC017, promotes ROS-dependent apoptosis through Akt and JAK2/STAT3 signaling pathway in human lung cancer cells.
Cell biology international
Song Y,Kong L,Sun B,Gao L,Chu P,Ahsan A,Qaed E,Lin Y,Peng J,Ma X,Zhang J,Wang S,Tang Z
Fri Dec 01 00:00:00 EST 2017
Gaseous signalling molecule SO2 via Hippo‑MST pathway to improve myocardial fibrosis of diabetic rats.
Molecular medicine reports
Liu M,Liu S,Tan W,Tang F,Long J,Li Z,Liang B,Chu C,Yang J
Fri Dec 01 00:00:00 EST 2017
Gaseous signalling molecule SO2 via Hippo‑MST pathway to improve myocardial fibrosis of diabetic rats.
Molecular medicine reports
Liu M,Liu S,Tan W,Tang F,Long J,Li Z,Liang B,Chu C,Yang J
Fri Dec 01 00:00:00 EST 2017
Deoxyelephantopin Induces Reactive Oxygen Species-Mediated Apoptosis and Autophagy in Human Osteosarcoma Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Zou J,Zhang Y,Sun J,Wang X,Tu H,Geng S,Liu R,Chen Y,Bi Z
Thu Nov 16 00:00:00 EST 2017
Deoxyelephantopin Induces Reactive Oxygen Species-Mediated Apoptosis and Autophagy in Human Osteosarcoma Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Zou J,Zhang Y,Sun J,Wang X,Tu H,Geng S,Liu R,Chen Y,Bi Z
Thu Nov 16 00:00:00 EST 2017
Selective CDK7 inhibition with BS-181 suppresses cell proliferation and induces cell cycle arrest and apoptosis in gastric cancer.
Drug design, development and therapy
Wang BY,Liu QY,Cao J,Chen JW,Liu ZS
Mon Dec 26 00:00:00 EST 2016