Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MAEGNHRKKP LKVLESLGKD FLTGVLDNLV EQNVLNWKEE EKKKYYDAKT EDKVRVMADS MQEKQRMAGQ MLLQTFFNID QISPNKKAHP NMEAGPPESG ESTDALKLCP HEEFLRLCKE RAEEIYPIKE RNNRTRLALI ICNTEFDHLP PRNGADFDIT GMKELLEGLD YSVDVEENLT ARDMESALRA FATRPEHKSS DSTFLVLMSH GILEGICGTV HDEKKPDVLL YDTIFQIFNN RNCLSLKDKP KVIIVQACRG ANRGELWVRD SPASLEVASS QSSENLEEDA VYKTHVEKDF IAFCSSTPHN VSWRDSTMGS IFITQLITCF QKYSWCCHLE EVFRKVQQSF ETPRAKAQMP TIERLSMTRY FYLFPGN
This gene encodes a protein that is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a prodomain and a large and small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. This caspase is able to cleave and activate its own precursor protein, as well as caspase 1 precursor. When overexpressed, this gene induces cell apoptosis. Alternative splicing results in transcript variants encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apoptotic cysteine protease Mih1/TX; CASP-4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; Caspase-4; ICE and Ced-3 homolog 2; ICE(rel)-II; ICH-2; Mih1; protease ICH-2; Protease TX
Gene Aliases: CASP4; ICE(rel)II; ICEREL-II; ICH-2; ICH2; Mih1; Mih1/TX; TX
UniProt ID: (Human) P49662
Entrez Gene ID: (Human) 837
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support