Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEGNHRKK PLKVLESLGK DFLTGVLDNL VEQNVLNWKE EEKKKYYDAK TEDKVRVMAD SMQEKQRMAG QMLLQTFFNI DQISPNKKAH PNMEAGPPES GESTDALKLC PHEEFLRLCK ERAEEIYPIK ERNNRTRLAL IICNTEFDHL PPRNGADFDI TGMKELLEGL DYSVDVEENL TARDMESALR AFATRPEHKS SDSTFLVLMS HGILEGICGT VHDEKKPDVL LYDTIFQIFN NRNCLSLKDK PKVIIVQACR GANRGELWVR DSPASLEVAS SQSSENLEED AVYKTHVEKD FIAFCSSTPH NVSWRDSTMG SIFITQLITC FQKYSWCCHL EEVFRKVQQS FETPRAKAQM PTIERLSMTR YFYLFPGN (1-377 aa encoded by BC017839)
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apoptotic cysteine protease Mih1/TX; CASP-4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; Caspase-4; ICE and Ced-3 homolog 2; ICE(rel)-II; ICH-2; Mih1; protease ICH-2; Protease TX
Gene Aliases: CASP4; ICE(rel)II; ICEREL-II; ICH-2; ICH2; Mih1; Mih1/TX; TX
UniProt ID: (Human) P49662
Entrez Gene ID: (Human) 837
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support