Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: Mouse spinal cord, Rat brain
Immunogen sequence: HLDHLDSASA DFPYNPGQGQ AIRNGVNRNS AIIGGVIAVV IFTILCTLVF LIRYMFRHKG TYHTNEAKGA ESAESADAAI MNNDPNFTET IDESKKEWLI
This gene encodes a member of the neurexin family which functions in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, thrombospondin N-terminal-like domains and a putative PDZ binding site. This protein is localized at the juxtaparanodes of myelinated axons, and mediates interactions between neurons and glia during nervous system development and is also involved in localization of potassium channels within differentiating axons. This gene encompasses almost 1.5% of chromosome 7 and is one of the largest genes in the human genome. It is directly bound and regulated by forkhead box protein P2 (FOXP2), a transcription factor related to speech and language development. This gene has been implicated in multiple neurodevelopmental disorders, including Gilles de la Tourette syndrome, schizophrenia, epilepsy, autism, ADHD and mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell recognition molecule Caspr2; contactin associated protein-like 2; Contactin-associated protein-like 2; DKFZp781D1846
Gene Aliases: 5430425M22Rik; Caspr2; Cntnap2; Kiaa0868; mKIAA0868
UniProt ID: (Mouse) Q9CPW0
Entrez Gene ID: (Rat) 500105, (Mouse) 66797
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support