Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: EAVYCKFHYK TDQGIKNLSV EDAARLSQED PDYGIRDLFN AIATGKYPSW TFYIQVMTFN QAETFPFNPF DLTKVWPHKD YPLIPVGKLV LNRNPVNYFA EVEQIAFDPS NMPPGIEASP DKMLQGRLFA YPDTHRHRLG PNYLHIPVNC PYRARVANYQ RDGPMCMQDN QGGAPNYYPN SFGAPEQQPS ALEHSIQYSG EVRRFNTAND DNVTQVRAFY VNVLNEEQRK RLCENIAGHL KDAQIFIQKK AVKNFTEVHP DYGSHIQALL DKYNAEKPKN AIHTFVQSGS HLAAREKANL
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Catalase; catalase 1; CTA1; HGNC:1516; MGC138422; MGC138424
Gene Aliases: 2210418N07; Cas-1; Cas1; CAT; Cat01; Catl; Cs-1; CS1
UniProt ID: (Human) P04040, (Pig) O62839, (Rat) P04762, (Mouse) P24270
Entrez Gene ID: (Human) 847, (Pig) 397568, (Rat) 24248, (Mouse) 12359
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support