Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: MCF7, A-549, PC-3, SP2/0, mouse testis, mouse spleen. The target is usually found in the following locations: Secreted.
Immunogen sequence: NKRFALLGDF FRKSKEKIGK EFKRIVQRIK DFLRNLVPR
Cathelicidin is an epithelial-derived antimicrobial protein found in specific granules of polymorphonuclear leukocytes of bone marrow, testis and neutrophils. Cathelicidins are one family of antimicrobial peptides characterized by conserved pro-peptide sequences that have been identified in epithelial tissues and some myeloid cells of humans and animals. LL-37/hCAP-18 is the only Cathelicidin found in humans and is expressed in inflammatory and epithelial cells. The presence of these molecules is essential for defense against invasive bacterial infection in skin. Besides their direct antimicrobial function, Cathelicidins have multiple roles in mediating innate and adaptive immunity, such as endotoxin neutralizing, angiogenesis, wound healing and promoting neutrophil chemotaxis and mast cell recruitment. Cathelicidin antimicrobial peptides qualify as prototypes of innovative drugs that may be used to treat infection and/or modulate the immune response.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18 kDa cationic antimicrobial protein; CAP-18; Cathelicidin antimicrobial peptide; cathelin-related antimicrobial peptide; hCAP-18; hCAP18
Gene Aliases: CAMP; CAP-18; CAP18; CRAMP; FALL-39; FALL39; HSD26; LL37
UniProt ID: (Human) P49913
Entrez Gene ID: (Human) 820, (Rat) 316010
Molecular Function: antimicrobial response protein defense/immunity protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support