Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
This antibody is specific to CTSD.
Immunogen sequence: VPAVTEGPI PEVLKNYMDA QYYGEIGIGT PPQCFTVVFD TGSSNLWVPS IHCKLLDIAC WIHHKYNSDK SSTYVKNGTS FDIHYGSGSL SGYLSQDTVS VPCQSASSAS ALGGVKVERQ VFGEATKQPG ITFIAAKFDG ILGMAYPRIS VNNVLPVFDN LMQQKLVDQN IFSFYLSRDP DAQPGGELML GGTDSKYYKG SLSYLNVTRK AYWQVHLDQV EVASGLTLCK EGCEAIVDTG TSLMVGPVDE VRELQKAIGA VPLIQGEYMI PCEKVSTLPA ITLKLGGKGY KLSPEDYTLK VSQAGKTLCL SGFMGMDIPP PSGPLWILGD VFIGRYYTVF DRDNNRVGFA EAARL (59-412 aa encoded by B C016320)
Cathepsin D has been reported to be involved in malignant tumor progression and with prognosis of various human cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cathepsin D; CathepsinD; Ceroid lipofuscinosis neuronal 10; ceroid-lipofuscinosis, neuronal 10; epididymis secretory sperm binding protein Li 130P; lysosomal aspartyl peptidase; lysosomal aspartyl protease; MGC2311; OTTHUMP00000196039; OTTHUMP00000198692
Gene Aliases: CLN10; CPSD; CTSD; HEL-S-130P
UniProt ID: (Human) P07339
Entrez Gene ID: (Human) 1509
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support