Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Immunogen sequence: GQSFSIQYGT GSLSGIIGAD QVSVEGLTVV GQQFGESVTE PGQTFVDAEF DGILGLGYPS LAVGGVTPVF DNMMAQNLVD LPMFSVYMSS NPEGGAGSEL IFGGYDHSHF SGS
Cathepsin E may have a role in immune function. It is most likely involved in the processing of antigenic peptides during MHC class II-mediated antigen presentation. It may also play a role in activation-induced lymphocyte depletion in the thymus, and in neuronal degeneration and glial cell activation in the brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cathepsin E; EC 3.4.23.34
Gene Aliases: A430072O03Rik; C920004C08Rik; CatE; CE; CEA; CEB; Ctse; Ctsea
UniProt ID: (Mouse) P70269, (Rat) P16228
Entrez Gene ID: (Mouse) 13034, (Rat) 25424
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support