Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Antibody detects endogenous levels of total Cathepsin G.
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cathepsin G; CG; CTSG protein; MGC23078; Vimentin-specific protease; VSP
Gene Aliases: CATG; CG; CTSG
UniProt ID: (Human) P08311, (Rat) P17977, (Mouse) P28293
Entrez Gene ID: (Human) 1511, (Rat) 290257, (Mouse) 13035
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support