Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Important for the overall degradation of proteins in lysosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aleurain; Cat H; Cathepsin B3; Cathepsin BA; N-benzoylarginine-beta-naphthylamide hydrolase; Pro-cathepsin H
Gene Aliases: ACC-4; ACC-5; ACC4; ACC5; AL022844; CPSB; CTSH
UniProt ID: (Human) P09668, (Rat) P00786, (Mouse) P49935
Entrez Gene ID: (Human) 1512, (Rat) 25425, (Mouse) 13036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support