Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 8
In direct ELISAs and Western blots, no cross-reactivity with recombinant human Cathepsin B, C, L2, O, S, or X/Z/P is observed. In Western blots, both the pro and active forms of recombinant human and mouse Cathspsin L are recognized.
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. At least two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cat L; Cathepsin L1; furless; Major excreted protein; MEP; nackt; p39 cysteine proteinase; Procathepsin L
Gene Aliases: 1190035F06Rik; CATL; CTSL; CTSL1; fs; MEP; nkt
UniProt ID: (Human) P07711, (Mouse) P06797
Entrez Gene ID: (Human) 1514, (Mouse) 13039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support