Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Samples: HeLa, HCT116, HepG2, Mouse lung, Mouse brain, Mouse spleen, Rat lung
Immunogen sequence: MSAAIAALAA SYGSGSGSES DSDSESSRCP LPAADSLMHL TKSPSSKPSL AVAVDSAPEV AVKEDLETGV HLDPAVKEVQ YNPTYETMFA PEFGPENPFR
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is found to be essential for the catalytic step II in pre-mRNA splicing process. It is found in the spliceosome, and contains seven WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp17 protein, which functions in two different cellular processes: pre-mRNA splicing and cell cycle progression. It suggests that this protein may play a role in cell cycle progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell division cycle 40 homolog; EH-binding protein 3; Ehb3; hPRP17; pre-mRNA splicing factor 17; Pre-mRNA-processing factor 17; PRP17 homolog
Gene Aliases: 1200003H23Rik; CDC40; EHB3; PRP17; PRPF17
UniProt ID: (Human) O60508, (Mouse) Q9DC48
Entrez Gene ID: (Human) 51362, (Mouse) 71713, (Rat) 361859
Molecular Function:
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support