Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAKERRRAVL ELLQRPGNAR CADCGAPDPD WASYTLGVFI CLSCSGIHRN IPQVSKVKSV RLDAWEEAQV EFMASHGNDA ARARFESKVP SFYYRPTPSD CQLLREQWIR AKYERQEFIY PEKQEPYSAG YREGFLWKRG RDNGQFLSRK FVLTEREGAL KYFNRNDAKE PKAVMKIEHL NATFQPAKIG HPHGLQVTYL KDNSTRNIFI YHEDGKEIVD WFNALRAARF HYLQVAFPGA SDADLVPKLS RNYLKEGYME KTGPKQTEGF RKRWFTMDDR RLMYFKDPLD AFARGEVFIG SKESGYTVLH GFPPSTQGHH WPHGITIVTP DRKFLFACET ESDQREWVAA FQKAVDRPML PQEYAVEAHF KHKP
Centaurin alpha-1 (ADAP1, ArfGAP With Dual PH Domains 1) is a Protein Coding gene. Among its related pathways are B Cell Receptor Signaling Pathway (sino) and Arf6 signaling events. Gene Ontology (GO) annotations related to this gene include GTPase activator activity and inositol 1,3,4,5 tetrakisphosphate binding. An important paralog of this gene is ADAP2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arf-GAP with dual PH domain-containing protein 1; centaurin, alpha 1; centaurin-alpha; Centaurin-alpha-1; Cnt-a1; Putative MAPK-activating protein PM25
Gene Aliases: 4930431P11Rik; ADAP1; CENTA1; GC1L; GCS1L; p42IP4
UniProt ID: (Human) O75689
Entrez Gene ID: (Human) 11033, (Mouse) 231821, (Rat) 171097
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support