Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
Positive Samples: Mouse serum, Rat testis, Rat serum
Immunogen sequence: IRGKHVRHYY IAAEEIIWNY APSGIDIFTK ENLTAPGSDS AVFFEQGTTR IGGSYKKLVY REYTDASFTN RKERGPEEEH LGILGPVIWA EVGDTIRVTF HNKGAYPLSI EPIGVRFNKN NEGTYYSPNY NPQSRSVPPS ASHVAPTETF TYEWTVPKEV GPTNADPVCL AKMYYSAVDP TKDIFTGLIG PMKICKKGSL HANGRQK
Ceruloplasmin encodes a protein that is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bilitranslocase; Ceruloplasmin; Cuproxidase ceruloplasmin; ferroxidase; Ferroxidase ceruloplasmin
Gene Aliases: CERP; CP; CP-2; D3Ertd555e
UniProt ID: (Human) P00450, (Mouse) Q61147
Entrez Gene ID: (Human) 1356, (Mouse) 12870, (Rat) 24268
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support