Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: PALAPAKPGG PGRPRDPNTC FFEGQQRPHG ARWAPNYDPL CSLCTCQRRT VICDPVVCPP PSCPHPVQAP DQCCPVCPEK QDVRDLPGLP RSRDPGEGCY FDGDRSWRAA GTRWHPVVPP FGLIKCAVCT CKGGTGEVHC EKVQCPRLAC AQPVRVNPTD CCKQCPVGSG AHPQLGDPMQ ADGPRGCRFA GQWFPESQSW HPSVPPFGEM SCITCRCGAG VPHCERDDCS LPLSCGSGKE SRCCSRCTAH RRPAPETRTD PELEKEAEGS
This gene encodes a secreted protein that dorsalizes early vertebrate embryonic tissues by binding to ventralizing TGF-beta-like bone morphogenetic proteins and sequestering them in latent complexes. The encoded protein may also have roles in organogenesis and during adulthood. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequence has not been determined.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chordin
Gene Aliases: Chd; Chrd
UniProt ID: (Mouse) Q9Z0E2
Entrez Gene ID: (Rat) 117275, (Mouse) 12667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support