Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Recommended positive controls: NT2D1, IMR32, U87-MG.
Predicted reactivity: Human (99%), Pig (84%), Rhesus Monkey (96%), Bovine (80%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Studies in rodents suggest that the full-length protein, secretogranin II, is involved in the packaging or sorting of peptide hormones and neuropeptides into secretory vesicles. The full-length protein is cleaved to produce the active peptide secretoneurin, which exerts chemotaxic effects on specific cell types, and EM66, whose function is unknown.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chromogranin C (Secretogranin II); Chromogranin-C; secretogranin 2; Secretogranin II; secretogranin II (chromogranin C); Secretogranin-2; secretoneurin; SgII
Gene Aliases: Chcg; CHGC; EM66; Scg-2; SCG2; SgII; SN
UniProt ID: (Human) P13521, (Mouse) Q03517, (Rat) P10362
Entrez Gene ID: (Human) 7857, (Mouse) 20254, (Rat) 24765
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support