Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHC17; Clathrin light chain A; Clathrin light chain B; clathrin, light chain (Lcb); clathrin, light chain A; clathrin, light chain B; clathrin, light polypeptide (Lca); clathrin, light polypeptide (Lcb); CLH17; CLTC; CTLA; CTLB; KIAA0034; Lca; Lcb
Gene Aliases: CLTA; CLTB; LCA; LCB
UniProt ID: (Human) P09496, (Human) P09497
Entrez Gene ID: (Human) 1211, (Human) 1212
Molecular Function:
vesicle coat protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support