Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Immunogen sequence: EVQKMRMAVI GGAIFLLAGL AILVATAWYG NRIVQEFYDP MTPVNARYEF GQALFTGWAA ASLCLLGGAL LCCSCPRKTT SYPTPRPYPK PAPSSGKDYV
Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claudin-1; Senescence-associated epithelial membrane protein; senescence-associated epithelial membrane protein 1
Gene Aliases: AI596271; CLD1; CLDN1; ILVASC; SEMP1; UNQ481/PRO944
UniProt ID: (Human) O95832, (Rat) P56745, (Mouse) O88551
Entrez Gene ID: (Human) 9076, (Rat) 65129, (Mouse) 12737
Molecular Function: tight junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support