Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TLIGGVMMC IACRGLAPEE TNYKAVSYHA SGHSVAYKPG GFKASTGFGS NTKNKKIYDG GARTEDEVQS YPSKHDYV (185-261 aa encoded by BC146668)
Claudin-18 is a tight junction protein expressed as lung-and stomach-specific isoforms which are generated by alternative splicing of the claudin-18 gene. The lung-specific form is a downstream target gene regulated by the T/EBP/NKX2.1 transcription factor. A splice variant lacking the C-terminal cytoplasmic domain also exists in mouse, but has not been confirmed in human. Human claudin-18 demonstrates 88% amino acid sequence identity to the mouse protein. Immunohistochemical studies have demonstrated complete membrane localization of claudin-18 in lung and stomach epithelial cells, while electron microscopy has shown that it is concentrated in the cell-cell borders of these cells. These features suggest a potentially important role for claudin-18 in the structure and function of tight junctions in the lung and stomach. Claudin-18 expression has also been reported in the inner ear.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claudin-18; surfactant associated 5; surfactant associated protein J; surfactant, pulmonary associated protein J
Gene Aliases: CLDN18; SFTA5; SFTPJ; UNQ778/PRO1572
UniProt ID: (Human) P56856, (Mouse) P56857
Entrez Gene ID: (Human) 51208, (Mouse) 56492
Molecular Function:
tight junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support