Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: Clavesin 1.
Required for normal morphology of late endosomes and/or lysosomes in neurons. Binds phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cellular retinaldehyde-binding protein-like; clathrin vesicle-associated Sec14 protein 1; Clavesin-1; clavesin-1 {ECO:0000303|PubMed:19651769}; clvs1 {ECO:0000303|PubMed:19651769}; retinaldehyde binding protein 1-like 1; Retinaldehyde-binding protein 1-like 1; Retinaldehyde-binding protein 1-like protein 1
Gene Aliases: 4933402J24Rik; Clavesin-1; Clvl1; CLVS1; CRALBPL; RGD1564200; RLBP1L1
UniProt ID: (Human) Q8IUQ0, (Rat) A6JFQ6, (Mouse) Q9D4C9
Entrez Gene ID: (Human) 157807, (Rat) 366311, (Mouse) 74438
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support