Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NVISCQGLA PSQGRPGLSL VKENYAELLE DAFLKNVTAQ ICIDKKCPDY TCPITFSSPA DITILLDGSA SVGSHNFDTT KRFAKRLAER FLTAGRTDPA HDVRVAVVQY SGTGQQRPER ASLQFLQNYT ALASAVDAMD FINDATDVND ALGYVTRFYR EASSGAAKKR LLLFSDGNSQ GATPAAIEKA VQEAQRAGIE IFVVVVGRQV NEPHIRVLVT GKTAEYDVAY GESHLFRVPS YQALLRGVFH QTVSRKVALG (770-1028 aa encoded by BC052575)
The extensive family of COL gene products (collagens) is composed of several chain types, including fibril-forming interstitial collagens (typesI, II, III and V) and basement membrane collagens (type IV), each type containing multiple isoforms. Collagens are fibrous, extracellular matrix proteins with high tensile strength and are the major components of connective tissue, such as tendons and cartilage. All collagens contain a triple helix domain and frequently show lateral self-association in order to form complex connective tissues. Several collagens also play a role in cell adhesion, important for maintaining normal tissue architecture and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha 1 (VI) chain (61 AA); Collagen alpha-1(VI) chain; collagen VI, alpha-1 polypeptide; collagen, type VI, alpha 1
Gene Aliases: BTHLM1; COL6A1; OPLL; UCHMD1
UniProt ID: (Human) P12109
Entrez Gene ID: (Human) 1291
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support