Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: Rat lung, Rat ovary
Immunogen sequence: MPEGFIKAGQ RPSLSGTPLV SANQGVTGMP VSAFTVILSK AYPAIGTPIP FDKILYNRQQ HYDPRTGIFT CQIPGIYYFS YHVHVKGTHV WVGLYKNGTP VMYTYDEYTK GYLDQASGSA IIDLTENDQV WLQLPNAESN GLYSSEYVHS SFSGFLVAPM
Type X collagen is a product of hypertrophic chondrocytes and has been localized to presumptive mineralization zones of hyaline cartilage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collagen alpha-1(X) chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia
Gene Aliases: COL10A1
UniProt ID: (Human) Q03692
Entrez Gene ID: (Human) 1300, (Rat) 25681
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support