Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
COL2A1 is the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1 type II collagen; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; CGA; chondrocalcin; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen, type II, alpha 1; disproportionate micromelia; MGC131516; Procollagen II alpha 1; procollagen, type II, alpha 1
Gene Aliases: ANFH; AOM; CG2A1A; COL11A3; Col2; Col2a; Col2a-1; COL2A1; COLLII; Del1; Dmm; Lpk; M100856; Rgsc856; SEDC; STL1
UniProt ID: (Human) P02458, (Rat) P05539, (Mouse) Q8CEF7
Entrez Gene ID: (Human) 1280, (Rat) 25412, (Mouse) 12824
Molecular Function:
extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support