Search Thermo Fisher Scientific
Complex III, or ubiquinol cytochrome c oxidoreductase, is a complex of 11 subunits and catalyses electron transfer from reduced ubiquinone or coenzyme Q 10 to cytochrome c. The complex contains 11 different subunits, one of which, cytochrome b, is encoded on mitochondrial DNA, while the remainder subunits are nuclear-encoded. Genetic complex III deficiencies occur but are rare.
Protein Aliases: Complex III subunit 1; Complex III subunit 10; Complex III subunit 2; Complex III subunit 3; Complex III subunit 4; Complex III subunit 5; Complex III subunit 6; Complex III subunit 7; Complex III subunit 8; Complex III subunit 9; Complex III subunit III; Complex III subunit IV; Complex III subunit VII; Complex III subunit VIII; Complex III subunit X; Complex III subunit XI; Core protein I; Core protein II; Cytochrome b; Cytochrome b-c1 complex subunit 1, mitochondrial; Cytochrome b-c1 complex subunit 10; Cytochrome b-c1 complex subunit 2, mitochondrial; Cytochrome b-c1 complex subunit 3; Cytochrome b-c1 complex subunit 4; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 6, mitochondrial; Cytochrome b-c1 complex subunit 7; Cytochrome b-c1 complex subunit 8; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; cytochrome bc-1 complex core protein II; Cytochrome c-1; Cytochrome c1 non-heme 11 kDa protein; Cytochrome c1 non-heme 7 kDa protein; Cytochrome c1, heme protein, mitochondrial; cytochrome C1, nonheme 7kDa protein; low molecular mass ubiquinone-binding protein (9.5kD); Mitochondrial hinge protein; mitochondrial ubiquinone-binding protein; QP-C; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RISP; ubiquinol cytochrome c reductase, 6.4kDa subunit; ubiquinol-cytochrome c reductase complex (7.2 kD); Ubiquinol-cytochrome c reductase complex 11 kDa protein; Ubiquinol-cytochrome c reductase complex 14 kDa protein; Ubiquinol-cytochrome c reductase complex 6.4 kDa protein; Ubiquinol-cytochrome c reductase complex 7.2 kDa protein; Ubiquinol-cytochrome c reductase complex 9.5 kDa protein; Ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; Ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, 6.4kDa subunit; ubiquinol-cytochrome c reductase, complex III subunit VI; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa; ubiquinol-cytochrome c reductase, complex III subunit VIII; ubiquinol-cytochrome c reductase, complex III subunit X, 7.2kDa; Ubiquinol-cytochrome-c reductase complex core protein 1; Ubiquinol-cytochrome-c reductase complex core protein 2; Ubiquinol-cytochrome-c reductase complex cytochrome b subunit; Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit
Gene Aliases: 0710008D09Rik; COB; CYC1; CYTB; D3S3191; HSPC051; HSPC119; HSPC151; MC3DN3; MC3DN4; MC3DN5; MC3DN6; MT-CYB; MTCYB; QCR1; QCR10; QCR2; QCR6; QCR7; QCR8; QCR9; QP-C; QPC; RIP1; RIS1; RISP; UCCR7.2; UCRC; UQBC; UQBP; UQCR; UQCR1; UQCR10; UQCR11; UQCR2; UQCR4; UQCR5; UQCR6; UQCR7; UQCR8; UQCRB; UQCRC1; UQCRC2; UQCRFS1; UQCRH; UQCRQ; UQPC
UniProt ID: (Human) P08574, (Human) Q9UDW1, (Human) O14957, (Human) P14927, (Human) P31930, (Human) P22695, (Human) P47985, (Human) P07919, (Human) O14949
Entrez Gene ID: (Human) 1537, (Human) 29796, (Human) 10975, (Human) 7381, (Human) 7384, (Human) 7385, (Human) 7386, (Human) 7388, (Human) 27089