Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDWGTLQTI LGGVNKHSTS IGKIWLTVLF IFRIMILVVA AKEVWGDEQA DFVCNTLQPG CKNVCYDHYF PISHIRLWAL QLIFVSTPAL LVAMHVAYRR HEKKRKFIKG EIKSEFKDIE EIKTQKVRIE GSLWWTYTSS IFFRVIFEAA FMYVFYVMYD GFSMQRLVKC NAWPCPNTVD CFVSRPTEKT VFTVFMIAVS GICILLNVTE LCYLLIRYCS GKSKKPV (1-226 aa encoded by BC017048)
Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variables (EKV) is an autosomal dominant genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: connexin 26; Connexin-26; Cx26; Gap junction beta-2 protein; gap junction channel protein connexin 26; gap junction membrane channel protein beta 2; gap junction protein, beta 2, 26kDa; mutant gap junction protein beta 2
Gene Aliases: AI325222; Cnx26; CX26; CXN-26; DFNA3; DFNA3A; DFNB1; DFNB1A; Gjb-2; GJB2; HID; KID; NSRD1; PPK
UniProt ID: (Human) P29033, (Mouse) Q00977, (Rat) P21994
Entrez Gene ID: (Human) 2706, (Mouse) 14619, (Rat) 394266
Molecular Function:
gap junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support