Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: A549 or Heart
Cellular location: Cell membrane, Cell junction.
Connexin-36 (CX36), is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. Mutations in connexin-encoding genes can lead to functional and developmental abnormalities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: connexin 36; Connexin-36; Cx36; Gap junction alpha-9 protein; Gap junction delta-2 protein; gap junction protein, delta 2, 36kDa; GJA10
Gene Aliases: CX36; GJA9; GJD2
UniProt ID: (Human) Q9UKL4
Entrez Gene ID: (Human) 57369
Molecular Function:
gap junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support