Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide shares 95% identity with rat sequence.
Specificity of this antibody: Connexin-40.
Connexin 40 (Cx40), belongs to the connexin family of gap junction proteins, which are expressed in pancreas, kidney, skeletal muscle, liver, placenta and heart. Connexin 40 is highly expressed in vascular endothelial cells, and the mammalian heart (specifically in the atrial myocardium, and the His-Purkinje conduction system). At least three other connexins are expressed in the heart, including Cx37 (endocardium), Cx43 (atrial and ventricular myocardium, and distal parts of the conduction system) and Cx45 (throughout the heart). Cx40 and Cx43 frequently co-localize in cells of the cardiovascular system and participate in the formation of heterotypic gap junction channels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Connexin-40; Cx40; Gap junction alpha-5 protein; gap junction membrane channel protein alpha 5
Gene Aliases: 5730555N10Rik; Cnx40; Cx40; Cxn-40; Gja-5; Gja5
UniProt ID: (Mouse) Q01231
Entrez Gene ID: (Mouse) 14613
Molecular Function:
gap junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support