Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Contactin-1; Glycoprotein gp135; Neural cell surface protein F3
Gene Aliases: AW495098; CNTN; CNTN1; F3; F3cam; GP135; MYPCN
UniProt ID: (Human) Q12860, (Rat) Q63198, (Mouse) P12960
Entrez Gene ID: (Human) 1272, (Rat) 117258, (Mouse) 12805
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support