Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 8
Immunogen sequence: MWKASAGHAV SIAQDDAGAD DWETDPDFVN DVSEKEQRWG AKTVQGSGHQ EHINIHKLRE NVFQEHQTLK EKELETGPKA SHGYGGKFGV EQDRMDKSAV
Cortactin (Cttn) is a ubiquitous actin-binding protein that was originally identified as a substrate for Src. It contributes to the organization of the actin cytoskeleton and cell shape. Cortactin also plays a role in the formation of lamellipodia and cell migration. It is accumulated in peripheral, actin-enriched structures of cells suggesting that cortactin facilitates actin network formation. Cortactin has four major domains of interest the N-terminal acidic (NTA) and tandem repeats domains and the C-terminal proline-rich and SH3 Domains. NTA associates with the Arp2/3 and WASP complex at F-actin branches. Cortactin is involved in promoting cell motility and invasion, including a critical role in invadopodia, actin rich-subcellular protrusions associated with degradation of the ECM by cancer cells. Cortactin is phosphorylated by src family kinases at Y421, Y466, and Y482 and S405 and S418 that are phosphorylated by Erk family kinases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1110020L01Rik; Amplaxin; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); FLJ34459; mammary tumor and squamous cell carcinoma associated (p80/85 src substrate); Oncogene EMS1; Oncogene SRC8; Src substrate cortactin; src8
Gene Aliases: 1110020L01Rik; CTTN; Cttnb; EMS1
UniProt ID: (Human) Q14247, (Mouse) Q60598, (Rat) Q66HL2
Entrez Gene ID: (Human) 2017, (Mouse) 13043, (Rat) 60465
Molecular Function:
actin or actin-binding cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support