Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 46 kD coxsackievirus and adenovirus receptor (CAR) protein; CAR; Coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog; Coxsackievirus B-adenovirus receptor; CVB3-binding protein; HCVADR; rCAR
Gene Aliases: 2610206D03Rik; AU016810; AW553441; CAR; Car1; CAR4/6; CXADR; HCAR; MCAR; MCVADR
UniProt ID: (Human) P78310, (Mouse) P97792, (Rat) Q9R066
Entrez Gene ID: (Human) 1525, (Mouse) 13052, (Rat) 89843
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support