Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive test controls include: A-549, MCF7, NIH/3T3, K-562, C6, Mouse liver, Mouse brain, Rat testis.
Immunogen sequence: KNPEEAELED TLNQVMVVFK YIEDKDVFQK FYAKMLAKRL VHQNSASDDA EASMISKLKQ ACGFEYTSKL QRMFQDIGVS KDLNEQFKKH LTNSEPLDLD FSIQVLSSGS WPFQQSCTFA LPSELERSYQ RFTAFYASRH SGRKLTWLYQ LSKGELVTN
CUL1 gene ontology annotations related to this gene include animal organ morphogenesis; apoptotic process; cell proliferation; protein monoubiquitination; SCF-dependent proteasomal ubiquitin-dependent protein catabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CUL-1; Cullin-1
Gene Aliases: CUL1
UniProt ID: (Human) Q13616, (Mouse) Q9WTX6
Entrez Gene ID: (Human) 8454, (Mouse) 26965, (Rat) 362356
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support