Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MELLCCEGT RHAPRAGPDP RLLGDQRVLQ SLLRLEERYV PRASYFQCVQ REIKPHMRKM LAYWMLEVCE EQRCEEEVFP LAMNYLDRYL SCVPTRKAQL QLLGAVCMLL ASKLRETTPL TIEKLCIYTD HAVSPRQLRD WEVLVLGKLK WDLAAVIAHD FLAFILHRLS LPRDRQALVK KHAQTFLALC ATDYTFAMYP PSMIATGSIG AAVQGLGACS MSGDELTELL AGITGTEVDC LRACQEQIEA ALRESLREAS QTSSSPAPKA PRGSSSQGPS QTSTPTDVTA IHL (1-292 aa encoded by BC011616)
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with and functions as a regulatory subunit of CDK4 or CDK6, whose activity is required for cell cycle G1/S transition. This protein has been shown to interact with and be involved in the phosphorylation of tumor suppressor protein Rb. The CDK4 activity associated with this cyclin was reported to be necessary for cell cycle progression through G2 phase into mitosis after UV radiation. Several transcript variants encoding different isoforms have been found for this gene. Diseases associated with CCND3 include Retinoblastoma and Plasma Cell Leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCND 3; CGD3; CyclinD3; D3 type cyclin; D3-type cyclin; G1/S-specific cyclin-D3; OTTHUMP00000223415; RP5-973N23.3
Gene Aliases: CCND3
UniProt ID: (Human) P30281
Entrez Gene ID: (Human) 896
Molecular Function:
kinase activator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support