Product References
WITHDRAWN: Expression of Tmem41b and MMP13 associated with poor outcome in osteosarcomas.
Neoplasma
Li GH,Liu X,Feng LJ,Zhang L
Sun Aug 20 00:00:00 EDT 2023
Effects of Ginsenoside Rg1 on the Biological Behavior of Human Amnion-Derived Mesenchymal Stem/Stromal Cells (hAD-MSCs).
Stem cells international
Ling L,Shu H,Huang Y,Hou J,Hua Y
Tue Feb 28 00:00:00 EST 2023
RSL1D1 knockdown induces ferroptosis and mediates ferrous iron accumulation in senescent cells by inhibiting FTH1 mRNA stability.
Carcinogenesis
Jin Y,Zhao L,Wang S,Zhang X,Quan J,Lin Z,Piao J
Fri May 26 00:00:00 EDT 2023
Luteoloside Induces G0/G1 Phase Arrest of Neuroblastoma Cells by Targeting p38 MAPK.
Molecules (Basel, Switzerland)
He Y,Luo M,Lei S,Zeng Z,Chen T,Wu Y,Wang D,Wang L,Wang L
Sun Feb 12 00:00:00 EST 2023
Oroxylin A activates ferritinophagy to induce hepatic stellate cell senescence against hepatic fibrosis by regulating cGAS-STING pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Sun Y,Weng J,Chen X,Ma S,Zhang Y,Zhang F,Zhang Z,Wang F,Shao J,Zheng S
Thu Jun 01 00:00:00 EDT 2023
Shikonin suppresses small cell lung cancer growth via inducing ATF3-mediated ferroptosis to promote ROS accumulation.
Chemico-biological interactions
Qian X,Zhu L,Xu M,Liu H,Yu X,Shao Q,Qin J
Fri Sep 01 00:00:00 EDT 2023
Atractylenolide III suppresses senescence-associated secretome via inhibiting cGAS/NF-κB pathway in hepatic stellate cells.
Clinical and experimental pharmacology & physiology
Wu H,Wu L,Xiao L,Gu Y,Liu H,Zhang L,Zhang M,Qi L
Sat Apr 01 00:00:00 EDT 2023
8-Methoxyflindersine-Induced Apoptosis and Cell Cycle Disorder Involving MAPK Signaling Activation in Human Colorectal Cancer Cells.
International journal of molecular sciences
Zhang D,Fu Y,Liu Y,Wu Y,Chen J,Zhang L,Wang R,Chen Z,Liu T
Fri Apr 28 00:00:00 EDT 2023
Identifying CDC7 as a synergistic target of chemotherapy in resistant small-cell lung cancer via CRISPR/Cas9 screening.
Cell death discovery
Deng L,Yang L,Zhu S,Li M,Wang Y,Cao X,Wang Q,Guo L
Thu Feb 02 00:00:00 EST 2023
N6-methyladenosine reader YTHDF2 promotes multiple myeloma cell proliferation through EGR1/p21cip1/waf1/CDK2-Cyclin E1 axis-mediated cell cycle transition.
Oncogene
Liu R,Miao J,Jia Y,Kong G,Hong F,Li F,Zhai M,Zhang R,Liu J,Xu X,Wang T,Liu H,Hu J,Yang Y,He A
Mon May 01 00:00:00 EDT 2023
N6-methyladenosine reader YTHDF2 promotes multiple myeloma cell proliferation through EGR1/p21cip1/waf1/CDK2-Cyclin E1 axis-mediated cell cycle transition.
Oncogene
Liu R,Miao J,Jia Y,Kong G,Hong F,Li F,Zhai M,Zhang R,Liu J,Xu X,Wang T,Liu H,Hu J,Yang Y,He A
Mon May 01 00:00:00 EDT 2023
N6-methyladenosine reader YTHDF2 promotes multiple myeloma cell proliferation through EGR1/p21cip1/waf1/CDK2-Cyclin E1 axis-mediated cell cycle transition.
Oncogene
Liu R,Miao J,Jia Y,Kong G,Hong F,Li F,Zhai M,Zhang R,Liu J,Xu X,Wang T,Liu H,Hu J,Yang Y,He A
Mon May 01 00:00:00 EDT 2023
N6-methyladenosine reader YTHDF2 promotes multiple myeloma cell proliferation through EGR1/p21cip1/waf1/CDK2-Cyclin E1 axis-mediated cell cycle transition.
Oncogene
Liu R,Miao J,Jia Y,Kong G,Hong F,Li F,Zhai M,Zhang R,Liu J,Xu X,Wang T,Liu H,Hu J,Yang Y,He A
Mon May 01 00:00:00 EDT 2023
Inhibition of ATF4-mediated elevation of both autophagy and AKT/mTOR was involved in antitumorigenic activity of curcumin.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Yang J,Bao S,Luo S,Jiang L,Li Q,Kong Y,Cao J
Wed Mar 01 00:00:00 EST 2023
Venlafaxine antagonizes the noradrenaline-promoted colon cancer progression by inhibiting the norepinephrine transporter.
Cell death discovery
Zhang H,Han J,Zhang J,Miao J,Li F,Tang K,Zhou K,Duan B,Li W,Cheng J,Sun Y,Hou N,Huang C
Mon May 08 00:00:00 EDT 2023
PAQR5 inhibits the growth and metastasis of clear cell renal cell carcinoma by suppressing the JAK/STAT3 signaling pathway.
Cellular oncology (Dordrecht)
Wang L,Yue Y,Zhang L,Jing M,Ma M,Liu C,Li Y,Xu S,Wang K,Wang X,Fan J,Zhang M
Sun Oct 01 00:00:00 EDT 2023
Emodin promotes hepatic stellate cell senescence and alleviates liver fibrosis via a nuclear receptor (Nur77)-mediated epigenetic regulation of glutaminase 1.
British journal of pharmacology
Chen L,Liang B,Xia S,Wang F,Li Z,Shao J,Zhang Z,Chen A,Zheng S,Zhang F
Sun Oct 01 00:00:00 EDT 2023
The location of estrogen receptor variant ER-α36 is associated with the invasion of glioblastoma.
Steroids
Li H,Ge N,Guan X,Han C,Li Y,Shen L,Chen M,Zhang B,Qu C,Zou W
Thu Jun 01 00:00:00 EDT 2023
221S-1a inhibits endothelial proliferation in pathological angiogenesis through ERK/c-Myc signaling.
European journal of pharmacology
Yao X,Xue Y,Ma Q,Bai Y,Jia P,Zhang Y,Lai B,He S,Ma Q,Zhang J,Tian H,Yin Q,Zheng X,Zheng X
Sat Aug 05 00:00:00 EDT 2023
Proline-rich 11 (PRR11) promotes the progression of cutaneous squamous cell carcinoma by activating the EGFR signaling pathway.
Molecular carcinogenesis
Wang S,Zhang X,Lei H,Song L,Huang Y,Kang T,Zhang M,Wang N,Yang P,Feng S,Wang J,Bai R,Wang N,Wang W,Zheng Y
Mon May 01 00:00:00 EDT 2023
IGFBP3 induced by the TGF-β/EGFRvIII transactivation contributes to the malignant phenotype of glioblastoma.
iScience
Zhang X,Wang G,Gong Y,Zhao L,Song P,Zhang H,Zhang Y,Ju H,Wang X,Wang B,Ren H,Zhu X,Dong Y
Fri May 19 00:00:00 EDT 2023
ASRGL1 downregulation suppresses hepatocellular carcinoma tumorigenesis in a CDK1-dependent manner.
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver
Wang X,Wang Y,Yang L,Yuan J,Shen W,Zhang W,Wang J,Tao K
Sat Jul 01 00:00:00 EDT 2023
ASRGL1 downregulation suppresses hepatocellular carcinoma tumorigenesis in a CDK1-dependent manner.
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver
Wang X,Wang Y,Yang L,Yuan J,Shen W,Zhang W,Wang J,Tao K
Sat Jul 01 00:00:00 EDT 2023
Lenvatinib inhibits intrahepatic cholangiocarcinoma via Gadd45a-mediated cell cycle arrest.
Discover. Oncology
Yan X,Wang D,Ning Z,Meng ZQ
Thu Feb 23 00:00:00 EST 2023
Lenvatinib inhibits intrahepatic cholangiocarcinoma via Gadd45a-mediated cell cycle arrest.
Discover. Oncology
Yan X,Wang D,Ning Z,Meng ZQ
Thu Feb 23 00:00:00 EST 2023
Sanggenon C inhibits cell proliferation and induces apoptosis by regulating the MIB1/DAPK1 axis in glioblastoma.
MedComm
Chang H,Hou J,Shao Y,Xu M,Weng X,Du Y,Shi J,Zhang L,Cui H
Tue Aug 01 00:00:00 EDT 2023
Antiovarian cancer mechanism of esculetin: inducing G0/G1 arrest and apoptosis via JAK2/STAT3 signalling pathway.
The Journal of pharmacy and pharmacology
Yin W,Fu X,Chang W,Han L,Meng J,Cao A,Ren X,Fan Z,Zhou S
Tue Jan 31 00:00:00 EST 2023
Metformin synergistically enhances the antitumour activity of Lenvatinib in hepatocellular carcinoma by altering AKT-FOXO3 signalling pathway.
Liver international : official journal of the International Association for the Study of the Liver
Cheng Y,Zhan P,Lu J,Lu Y,Luo C,Cen X,Wang F,Xie C,Yin Z
Sat Jul 01 00:00:00 EDT 2023
Differential effects of WRAP53 transcript variants on non-small cell lung cancer cell behaviors.
PloS one
Zhu Y,Sun W,Jiang X,Bai R,Luo Y,Gao Y,Li S,Huang Z,Gong Y,Xie C
Fri Feb 03 00:00:00 EST 2023
Onvansertib inhibits the proliferation and improves the cisplatin-resistance of lung adenocarcinoma via β-catenin/c-Myc signaling pathway.
American journal of cancer research
Wang R,Hou Y,Geng G,Zhu X,Wang Z,Cai W,Ye J,Zhao S,Mi Y,Jiang J
Sat Mar 11 00:00:00 EST 2023
Oroxylin A regulates cGAS DNA hypermethylation induced by methionine metabolism to promote HSC senescence.
Pharmacological research
Zhao D,Gao Y,Su Y,Zhou Y,Yang T,Li Y,Wang Y,Sun Y,Chen L,Zhang F,Zhang Z,Wang F,Shao J,Zheng S
Sun Jan 01 00:00:00 EST 2023
MicroRNA-1205 Suppresses Hepatocellular Carcinoma Cell Proliferation via a CSNK2B/CDK4 Axis.
Technology in cancer research & treatment
Li X,Xie S,Xia Q,Yan J,Chen S,Shen J
Wed Jan 11 00:00:00 EST 2023
Artesunate and cisplatin synergistically inhibit HNSCC cell growth and promote apoptosis with artesunate‑induced decreases in Rb and phosphorylated Rb levels.
Oncology reports
Okamoto H,Yoshikawa K,Shimada A,Sano R,Inukai D,Yamanaka S,Suzuki S,Ueda R,Ueda H,Fujimoto Y,Ogawa T
Tue Aug 01 00:00:00 EDT 2023
Hypoxia Inhibits Cell Cycle Progression and Cell Proliferation in Brain Microvascular Endothelial Cells via the miR-212-3p/MCM2 Axis.
International journal of molecular sciences
Shi Q,Li S,Lyu Q,Zhang S,Bai Y,Ma J
Wed Feb 01 00:00:00 EST 2023
Triptolide inhibits intrahepatic cholangiocarcinoma growth by suppressing glycolysis via the AKT/mTOR pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Li L,Wang C,Qiu Z,Deng D,Chen X,Wang Q,Meng Y,Zhang B,Zheng G,Hu J
Sun Jan 01 00:00:00 EST 2023
Metformin inhibits pulmonary artery smooth muscle cell proliferation by upregulating p21 via NONRATT015587.2.
International journal of molecular medicine
Sun Z,Liu Y,Hu R,Wang T,Li Y,Liu N
Fri Apr 01 00:00:00 EDT 2022
FBXW2 inhibits prostate cancer proliferation and metastasis via promoting EGFR ubiquitylation and degradation.
Cellular and molecular life sciences : CMLS
Zhou T,Chen T,Lai B,Zhang W,Luo X,Xia D,Fu W,Xu J
Mon May 02 00:00:00 EDT 2022
N-glycosylation of GDF15 abolishes its inhibitory effect on EGFR in AR inhibitor-resistant prostate cancer cells.
Cell death & disease
Wang R,Wen P,Yang G,Feng Y,Mi Y,Wang X,Zhu S,Chen YQ
Tue Jul 19 00:00:00 EDT 2022
Exploring the molecular mechanisms underlie the endoplasmic reticulum stress-mediated methylmercury-induced neuronal developmental damage.
Ecotoxicology and environmental safety
Pan J,Li X,Liu H,Wang C,Xu S,Xu B,Deng Y,Yang T,Liu W
Sat Oct 15 00:00:00 EDT 2022
Tetracaine hydrochloride induces cell cycle arrest in melanoma by downregulating hnRNPA1.
Toxicology and applied pharmacology
Huang X,Chen Y,Yi J,Yi P,Jia J,Liao Y,Feng J,Jiang X
Sat Jan 01 00:00:00 EST 2022
KIFC3 Promotes Proliferation, Migration, and Invasion in Colorectal Cancer via PI3K/AKT/mTOR Signaling Pathway.
Frontiers in genetics
Liao H,Zhang L,Lu S,Li W,Dong W
Sat Jul 16 00:00:00 EDT 2022
Integrative and Comprehensive Pan-Cancer Analysis of Lymphocyte-Specific Protein Tyrosine Kinase in Human Tumors.
International journal of molecular sciences
Han M,Li Y,Guo Y,Zhu W,Jiang J
Sun Nov 13 00:00:00 EST 2022
Nav1.6 promotes the progression of human follicular thyroid carcinoma cells via JAK-STAT signaling pathway.
Pathology, research and practice
Li H,Liu J,Fan N,Wang H,Thomas AM,Yan Q,Li S,Qin H
Mon Aug 01 00:00:00 EDT 2022
Rosuvastatin exerts cardioprotective effect in lipopolysaccharide-mediated injury of cardiomyocytes in an MG53-dependent manner.
BMC cardiovascular disorders
Zhuang J,Cheng G,Huang J,Guo H,Lai Y,Wang J,Shan Z,Zheng S
Wed Feb 23 00:00:00 EST 2022
CDK5RAP3 acts as a putative tumor inhibitor in papillary thyroid carcinoma via modulation of Akt/GSK-3β/Wnt/β-catenin signaling.
Toxicology and applied pharmacology
Feng X,Jiang J,Sun L,Zhou Q
Fri Apr 01 00:00:00 EDT 2022
Chrysin Ameliorates Influenza Virus Infection in the Upper Airways by Repressing Virus-Induced Cell Cycle Arrest and Mitochondria-Dependent Apoptosis.
Frontiers in immunology
Liu Y,Song X,Li C,Hu H,Li W,Wang L,Hu J,Liao C,Liang H,He Z,Ye L
Tue Apr 19 00:00:00 EDT 2022
Epigenetic silencing of JAM3 promotes esophageal cancer development by activating Wnt signaling.
Clinical epigenetics
Yang W,Guo C,Herman JG,Zhu C,Lv H,Su X,Zhang L,Zhang M,Guo M
Fri Dec 02 00:00:00 EST 2022
Identification and validation of a potential key gene SGOL1 for poor prognosis in hepatocellular carcinoma based on a bioinformatics approach.
Frontiers in oncology
Fei X,Liu S,Liu P,Wang X,Zhu C,Hou J,Cai J,Pan Y
Tue Nov 29 00:00:00 EST 2022
Porcine Circovirus 2 Activates the PERK-Reactive Oxygen Species Axis To Induce p53 Phosphorylation with Subsequent Cell Cycle Arrest at S Phase in Favor of Its Replication.
Journal of virology
Deng Z,Sun R,Han X,Zhang Y,Zhou Y,Shan Y,Xu J,Li X,He F,Fang W
Wed Nov 23 00:00:00 EST 2022
tRNA-m1A modification promotes T cell expansion via efficient MYC protein synthesis.
Nature immunology
Liu Y,Zhou J,Li X,Zhang X,Shi J,Wang X,Li H,Miao S,Chen H,He X,Dong L,Lee GR,Zheng J,Liu RJ,Su B,Ye Y,Flavell RA,Yi C,Wu Y,Li HB
Sat Oct 01 00:00:00 EDT 2022
Graphene quantum dots rescue angiogenic retinopathy via blocking STAT3/Periostin/ERK signaling.
Journal of nanobiotechnology
Zhao N,Gui X,Fang Q,Zhang R,Zhu W,Zhang H,Li Q,Zhou Y,Zhao J,Cui X,Gao G,Tang H,Shen N,Chen T,Song H,Shen W
Sat Apr 02 00:00:00 EDT 2022
RAB3D, upregulated by aryl hydrocarbon receptor (AhR), promotes the progression of prostate cancer by activating the PI3K/AKT signaling pathway.
Cell biology international
Yu J,Qi H,Wang Z,Zhang Z,Song E,Song W,An R
Thu Dec 01 00:00:00 EST 2022
RNF6 promotes the migration and invasion of breast cancer by promoting the ubiquitination and degradation of MST1.
Experimental and therapeutic medicine
Huang Y,Zou Y,Jie Z
Tue Feb 01 00:00:00 EST 2022
FBXW7 Reduces the Cancer Stem Cell-Like Properties of Hepatocellular Carcinoma by Regulating the Ubiquitination and Degradation of ACTL6A.
Stem cells international
Wang X,Li Y,Li Y,Liu P,Liu S,Pan Y
Sat Feb 04 00:00:00 EST 2023
High expression of HOXA4 in patients with glioma indicates unfavorable clinical outcomes.
Cell cycle (Georgetown, Tex.)
Yu Z,Liu Z,Lian X,Cheng X,Liu B,Zhang B,Wang H,Wang J,Li A,Ren Z,Pang B,Qian R,Gao Y
Tue Nov 01 00:00:00 EDT 2022
High expression of serine and arginine-rich splicing factor 9 (SRSF9) is associated with hepatocellular carcinoma progression and a poor prognosis.
BMC medical genomics
Zhang G,Liu B,Shang H,Wu G,Wu D,Wang L,Li S,Wang Z,Wang S,Yuan J
Mon Aug 15 00:00:00 EDT 2022
NRF2-directed PRPS1 upregulation to promote the progression and metastasis of melanoma.
Frontiers in immunology
Xiong G,Feng Y,Yi X,Zhang X,Li X,Yang L,Yi Z,Sai B,Yang Z,Zhang Q,Kuang Y,Zhu Y
Mon Oct 10 00:00:00 EDT 2022
Suppression of the Inhibitory Effect of circ_0036176-Translated Myo9a-208 on Cardiac Fibroblast Proliferation by miR-218-5p.
Journal of cardiovascular translational research
Guo J,Chen LW,Huang ZQ,Guo JS,Li H,Shan Y,Chen ZR,Yan YM,Zhu JN,Guo HM,Fang XH,Shan ZX
Wed Jun 01 00:00:00 EDT 2022
ALG3 Promotes Peritoneal Metastasis of Ovarian Cancer through Increasing Interaction of α1,3-mannosylated uPAR and ADAM8.
Cells
Cui X,Pei X,Wang H,Feng P,Qin H,Liu S,Yan Q,Liu J
Thu Oct 06 00:00:00 EDT 2022
INPP5F translocates into cytoplasm and interacts with ASPH to promote tumor growth in hepatocellular carcinoma.
Journal of experimental & clinical cancer research : CR
Zhou Q,Lin J,Yan Y,Meng S,Liao H,Chen R,He G,Zhu Y,He C,Mao K,Wang J,Zhang J,Zhou Z,Xiao Z
Fri Jan 07 00:00:00 EST 2022
Extracorporeal shockwave relieves endothelial injury and dysfunction in steroid-induced osteonecrosis of the femoral head via miR-135b targeting FOXO1: in vitro and in vivo studies.
Aging
Wu X,Wang Y,Fan X,Xu X,Sun W
Fri Jan 07 00:00:00 EST 2022
The mechanism of the imbalance between proliferation and ferroptosis in pulmonary artery smooth muscle cells based on the activation of SLC7A11.
European journal of pharmacology
Hu P,Xu Y,Jiang Y,Huang J,Liu Y,Wang D,Tao T,Sun Z,Liu Y
Fri Aug 05 00:00:00 EDT 2022
Sinapic acid inhibits pancreatic cancer proliferation, migration, and invasion via downregulation of the AKT/Gsk-3β signal pathway.
Drug development research
Huang Z,Chen H,Tan P,Huang M,Shi H,Sun B,Cheng Y,Li T,Mou Z,Li Q,Fu W
Sun May 01 00:00:00 EDT 2022
NT5DC2 promotes leiomyosarcoma tumour cell growth via stabilizing unpalmitoylated TEAD4 and generating a positive feedback loop.
Journal of cellular and molecular medicine
Hu B,Zhou S,Hu X,Zhang H,Lan X,Li M,Wang Y,Hu Q
Sun May 16 00:00:00 EDT 2021
Overexpression of SHMT2 Predicts a Poor Prognosis and Promotes Tumor Cell Growth in Bladder Cancer.
Frontiers in genetics
Zhang P,Yang Q
Tue Jun 22 00:00:00 EDT 2021
Tacrolimus inhibits oral carcinogenesis through cell cycle control.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li Y,Wang Y,Li J,Ling Z,Chen W,Zhang L,Hu Q,Wu T,Cheng B,Wang Y,Xia J
Thu Jul 01 00:00:00 EDT 2021
Bioinformatics analysis combined with experiments predicts CENPK as a potential prognostic factor for lung adenocarcinoma.
Cancer cell international
Ma J,Chen X,Lin M,Wang Z,Wu Y,Li J
Thu Jan 21 00:00:00 EST 2021
Exogenous Biological Renal Support Improves Kidney Function in Mice With Rhabdomyolysis-Induced Acute Kidney Injury.
Frontiers in medicine
Liu C,Chi K,Geng X,Hong Q,Mao Z,Huang Q,Liu D,Wang Y,Zhang Y,Zhou F,Cai G,Chen X,Sun X
Tue Jun 15 00:00:00 EDT 2021
METTL16 promotes cell proliferation by up-regulating cyclin D1 expression in gastric cancer.
Journal of cellular and molecular medicine
Wang XK,Zhang YW,Wang CM,Li B,Zhang TZ,Zhou WJ,Cheng LJ,Huo MY,Zhang CH,He YL
Thu Jul 01 00:00:00 EDT 2021
Ropivacaine Induces Cell Cycle Arrest in the G0/G1 Phase and Apoptosis of PC12 Cells via Inhibiting Mitochondrial STAT3 Translocation.
Inflammation
Zeng L,Li A,Zhang Z,Zhang F,Chen H,Wang Y,Ding X,Luo H
Wed Dec 01 00:00:00 EST 2021
Role of Glycated High Mobility Group Box-1 in Gastric Cancer.
International journal of molecular sciences
Kishi S,Nishiguchi Y,Honoki K,Mori S,Fujiwara-Tani R,Sasaki T,Fujii K,Kawahara I,Goto K,Nakashima C,Kido A,Tanaka Y,Luo Y,Kuniyasu H
Thu May 13 00:00:00 EDT 2021
HSD17B6 downregulation predicts poor prognosis and drives tumor progression via activating Akt signaling pathway in lung adenocarcinoma.
Cell death discovery
Tian T,Hong F,Wang Z,Hu J,Chen N,Lv L,Yi Q
Mon Nov 08 00:00:00 EST 2021
Pinocembrin Inhibits the Proliferation and Metastasis of Breast Cancer via Suppression of the PI3K/AKT Signaling Pathway.
Frontiers in oncology
Zhu X,Li R,Wang C,Zhou S,Fan Y,Ma S,Gao D,Gai N,Yang J
Tue Aug 03 00:00:00 EDT 2021
Sp1-Mediated circRNA circHipk2 Regulates Myogenesis by Targeting Ribosomal Protein Rpl7.
Genes
Yan J,Yang Y,Fan X,Tang Y,Tang Z
Sat May 08 00:00:00 EDT 2021
Stimulation of ROS Generation by Extract of Warburgia ugandensis Leading to G0/G1 Cell Cycle Arrest and Antiproliferation in A549 Cells.
Antioxidants (Basel, Switzerland)
Zhang YL,Chen GL,Liu Y,Zhuang XC,Guo MQ
Thu Sep 30 00:00:00 EDT 2021
MicroRNA-497 Reduction and Increase of Its Family Member MicroRNA-424 Lead to Dysregulation of Multiple Inflammation Related Genes in Synovial Fibroblasts With Rheumatoid Arthritis.
Frontiers in immunology
Wang S,Xu J,Guo Y,Cai Y,Ren X,Zhu W,Geng M,Meng L,Jiang C,Lu S
Mon Jun 28 00:00:00 EDT 2021
PKMYT1, exacerbating the progression of clear cell renal cell carcinoma, is implied as a biomarker for the diagnosis and prognosis.
Aging
Chen J,Hua X,Chen H,Qiu X,Xiao H,Ge S,Liang C,Zhou Q
Mon Dec 27 00:00:00 EST 2021
NR1D1 suppressed the growth of ovarian cancer by abrogating the JAK/STAT3 signaling pathway.
BMC cancer
Wang H,Fu Y
Fri Jul 30 00:00:00 EDT 2021
Discovery of a novel HDACi structure that inhibits the proliferation of ovarian cancer cells in vivo and in vitro.
International journal of biological sciences
Bai M,Cui M,Li M,Yao X,Wu Y,Zheng L,Sun L,Song Q,Wang S,Liu L,Yu C,Huang Y
Tue Feb 22 00:00:00 EST 2022
SETD4 in the Proliferation, Migration, Angiogenesis, Myogenic Differentiation and Genomic Methylation of Bone Marrow Mesenchymal Stem Cells.
Stem cell reviews and reports
Liao X,Wu C,Shao Z,Zhang S,Zou Y,Wang K,Ha Y,Xing J,Zheng A,Shen Z,Zheng S,Guo J,Jie W
Sun Aug 01 00:00:00 EDT 2021
Ixazomib inhibits myeloma cell proliferation by targeting UBE2K.
Biochemical and biophysical research communications
Wang Q,Dong Z,Su J,Huang J,Xiao P,Tian L,Chen Y,Ma L,Chen X
Fri Apr 16 00:00:00 EDT 2021
CUX1 Facilitates the Development of Oncogenic Properties Via Activating Wnt/β-Catenin Signaling Pathway in Glioma.
Frontiers in molecular biosciences
Feng F,Zhao Z,Zhou Y,Cheng Y,Wu X,Heng X
Tue Aug 24 00:00:00 EDT 2021
Endogenous hydrogen sulfide improves vascular remodeling through PPARδ/SOCS3 signaling.
Journal of advanced research
Tian D,Teng X,Jin S,Chen Y,Xue H,Xiao L,Wu Y
Fri Jan 01 00:00:00 EST 2021
Identification of abnormally high expression of POGZ as a new biomarker associated with a poor prognosis in osteosarcoma.
European journal of histochemistry : EJH
Zheng S,Liu Y,Sun H,Jia J,Wu T,Ding R,Cheng X
Fri Sep 03 00:00:00 EDT 2021
NDUFA4L2 in smooth muscle promotes vascular remodeling in hypoxic pulmonary arterial hypertension.
Journal of cellular and molecular medicine
Liu Y,Nie X,Zhu J,Wang T,Li Y,Wang Q,Sun Z
Fri Jan 01 00:00:00 EST 2021
NUDT21 Promotes Tumor Growth and Metastasis Through Modulating SGPP2 in Human Gastric Cancer.
Frontiers in oncology
Zhu Y,Zhang R,Zhang Y,Cheng X,Li L,Wu Z,Ding K
Wed Sep 13 00:00:00 EDT 2023
Silent information regulator 2 promotes clear cell renal cell carcinoma progression through deacetylation and small ubiquitin-related modifier 1 modification of glucose 6-phosphate dehydrogenase.
Cancer science
Ni Y,Yang Z,Agbana YL,Bai H,Wang L,Yang L,Yi Z,Cheng J,Zhang Q,Kuang Y,Zhu Y
Fri Oct 01 00:00:00 EDT 2021
Ezrin Promotes the Proliferation, Migration, and Invasion of Ovarian Cancer Cells.
Biomedical and environmental sciences : BES
Li MJ,Xiong D,Huang H,Wen ZY
Sat Feb 20 00:00:00 EST 2021
Knockdown of enhancer of rudimentary homolog expression attenuates proliferation, cell cycle and apoptosis of melanoma cells.
Melanoma research
Xiao M,Tang N,Yan Y,Li Z,Shi S,He S,Chen Z,Cao K,Chen J,Zhou J,Chen X
Sun Aug 01 00:00:00 EDT 2021
HHLA2 deficiency inhibits non-small cell lung cancer progression and THP-1 macrophage M2 polarization.
Cancer medicine
Sun W,Li S,Tang G,Sun S,Luo Y,Bai R,Han L,Jiang X,Gao Y,Huang Z,Zhang J,Gong Y,Xie C
Sun Aug 01 00:00:00 EDT 2021
MCC Regulator of WNT Signaling Pathway (MCC) Is a Podocyte Essential Gene.
Frontiers in medicine
Song H,Zhuang L,Xu X,Shi J,Hu W,Liu Z,Shi S
Tue Dec 21 00:00:00 EST 2021
Regulatory effect of microRNA-223-3p on breast cancer cell processes via the Hippo/Yap signaling pathway.
Oncology letters
Du T,Wang D,Wan X,Xu J,Xiao Q,Liu B
Thu Jul 01 00:00:00 EDT 2021
LncRNA CD27-AS1 promotes acute myeloid leukemia progression through the miR-224-5p/PBX3 signaling circuit.
Cell death & disease
Tao Y,Zhang J,Chen L,Liu X,Yao M,Zhang H
Tue May 18 00:00:00 EDT 2021
Epidermal stem cells maintain stemness via a biomimetic micro/nanofiber scaffold that promotes wound healing by activating the Notch signaling pathway.
Stem cell research & therapy
Lin Z,Zhao C,Lei Z,Zhang Y,Huang R,Lin B,Dong Y,Zhang H,Li J,Li X
Thu Jun 10 00:00:00 EDT 2021
Lidocaine inhibits the metastatic potential of ovarian cancer by blocking NaV 1.5-mediated EMT and FAK/Paxillin signaling pathway.
Cancer medicine
Liu C,Yu M,Li Y,Wang H,Xu C,Zhang X,Li M,Guo H,Ma D,Guo X
Fri Jan 01 00:00:00 EST 2021
UHRF1 Knockdown Attenuates Cell Growth, Migration, and Invasion in Cutaneous Squamous Cell Carcinoma.
Cancer investigation
Li Q,Chu Z,Geng S
Fri Jan 01 00:00:00 EST 2021
Cannabisin D from Sinomenium Acutum Inhibits Proliferation and Migration of Glioblastoma Cells through MAPKs Signaling.
Nutrition and cancer
Zhang D,Li C,Zhang L,Li B,Wang Y,Wang R,Chen Z,Xu L,Liu T
Thu Feb 24 00:00:00 EST 2022
ActivinA activates Notch1-Shh signaling to regulate proliferation in C2C12 skeletal muscle cells.
Molecular and cellular endocrinology
Ma L,Li C,Lian S,Xu B,Yuan J,Lu J,Yang H,Guo J,Ji H
Fri Jan 01 00:00:00 EST 2021
S100A11 regulates nasal epithelial cell remodeling and inflammation in CRSwNPs via the RAGE-mediated AMPK-STAT3 pathway.
Molecular immunology
Liu C,Du H,Wang Y,Gong N,Qi W,Zhou X,Shi L
Wed Dec 01 00:00:00 EST 2021
CDCA7 Facilitates Tumor Progression by Directly Regulating CCNA2 Expression in Esophageal Squamous Cell Carcinoma.
Frontiers in oncology
Li H,Weng Y,Wang S,Wang F,Wang Y,Kong P,Zhang L,Cheng C,Cui H,Xu E,Wei S,Guo D,Chen F,Bi Y,Meng Y,Cheng X,Cui Y
Thu Apr 28 00:00:00 EDT 2022
MicroRNA miR-188-5p as a mediator of long non-coding RNA MALAT1 regulates cell proliferation and apoptosis in multiple myeloma.
Bioengineered
Liu H,Chi Z,Jin H,Yang W
Wed Dec 01 00:00:00 EST 2021
Triptolidenol, isolated from Tripterygium wilfordii, disrupted NF-κB/COX-2 pathway by targeting ATP-binding sites of IKKβ in clear cell renal cell carcinoma.
Fitoterapia
Jin J,Zhou M,Wang X,Liu M,Huang H,Yan F,Yu Z,Shu X,Huo X,Feng L,Zhang B,Huang S,Deng S,Wang C,Ma X
Fri Jan 01 00:00:00 EST 2021
Circ_0089823 reinforces malignant behaviors of non-small cell lung cancer by acting as a sponge for microRNAs targeting SOX4.
Neoplasia (New York, N.Y.)
Li J,Zhu Z,Li S,Han Z,Meng F,Wei L
Wed Sep 01 00:00:00 EDT 2021
(20S) Ginsenoside Rh2 Exerts Its Anti-Tumor Effect by Disrupting the HSP90A-Cdc37 System in Human Liver Cancer Cells.
International journal of molecular sciences
Chen C,Wang YS,Zhang ET,Li GA,Liu WY,Li Y,Jin YH
Mon Dec 06 00:00:00 EST 2021
CKAP2L, as an Independent Risk Factor, Closely Related to the Prognosis of Glioma.
BioMed research international
Zhu L,Zheng Y,Hu R,Hu C
Tue Jan 11 00:00:00 EST 2022
Epiberberine ameliorated diabetic nephropathy by inactivating the angiotensinogen (Agt) to repress TGFβ/Smad2 pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Xiao Y,Deng J,Li C,Gong X,Gui Z,Huang J,Zhang Y,Liu Y,Ye X,Li X
Mon Mar 01 00:00:00 EST 2021
STAT3-mediated activation of mitochondrial pathway contributes to antitumor effect of dihydrotanshinone I in esophageal squamous cell carcinoma cells.
World journal of gastrointestinal oncology
Qi MM,He PZ,Zhang L,Dong WG
Sun Aug 15 00:00:00 EDT 2021
Xenobiotic Receptor CAR Is Highly Induced in Psoriasis and Promotes Keratinocyte Proliferation.
The Journal of investigative dermatology
Lai B,Xie X,Li F,Cui Q,Dang E,Luo W,Wang N,Zheng Y,Wang G,Xiao L,Wang N
Wed Dec 01 00:00:00 EST 2021
Astragalin Inhibits the Proliferation and Migration of Human Colon Cancer HCT116 Cells by Regulating the NF-κB Signaling Pathway.
Frontiers in pharmacology
Yang M,Li WY,Xie J,Wang ZL,Wen YL,Zhao CC,Tao L,Li LF,Tian Y,Sheng J
Sat May 08 00:00:00 EDT 2021
Effects of Cynaroside on Cell Proliferation, Apoptosis, Migration and Invasion though the MET/AKT/mTOR Axis in Gastric Cancer.
International journal of molecular sciences
Ji J,Wang Z,Sun W,Li Z,Cai H,Zhao E,Cui H
Tue Nov 09 00:00:00 EST 2021
Long Non-Coding RNA CCAT2 Activates RAB14 and Acts as an Oncogene in Colorectal Cancer.
Frontiers in oncology
Wang D,Li Z,Yin H
Tue Dec 07 00:00:00 EST 2021
Long Non-Coding RNA CCAT2 Activates RAB14 and Acts as an Oncogene in Colorectal Cancer.
Frontiers in oncology
Wang D,Li Z,Yin H
Tue Dec 07 00:00:00 EST 2021
Long Non-Coding RNA CCAT2 Activates RAB14 and Acts as an Oncogene in Colorectal Cancer.
Frontiers in oncology
Wang D,Li Z,Yin H
Tue Dec 07 00:00:00 EST 2021
Long Non-Coding RNA CCAT2 Activates RAB14 and Acts as an Oncogene in Colorectal Cancer.
Frontiers in oncology
Wang D,Li Z,Yin H
Tue Dec 07 00:00:00 EST 2021
Resveratrol inhibits the progression of cervical cancer by suppressing the transcription and expression of HPV E6 and E7 genes.
International journal of molecular medicine
Sun X,Fu P,Xie L,Chai S,Xu Q,Zeng L,Wang X,Jiang N,Sang M
Fri Jan 01 00:00:00 EST 2021
Tissue Inhibitor of Metalloproteinases-2 Mediates Kidney Injury during Sepsis.
Nephron
Wen X,Zhang J,Wan X,Frank AA,Qu X,Kellum JA
Tue Oct 26 00:00:00 EDT 2021
Mechanism of inhibiting proliferation of hepatocellular carcinoma Hepa1-6 cells by embryonic stem cell-conditioned medium.
Experimental and therapeutic medicine
Li L,Zheng Y,Zheng Q,Jiang J
Wed Apr 01 00:00:00 EDT 2020
Elevated expression of minichromosome maintenance 3 indicates poor outcomes and promotes G1/S cell cycle progression, proliferation, migration and invasion in colorectal cancer.
Bioscience reports
Zhou H,Xiong Y,Zhang G,Liu Z,Li L,Hou S,Zhou T
Fri Jul 31 00:00:00 EDT 2020
LncRNA TROJAN promotes proliferation and resistance to CDK4/6 inhibitor via CDK2 transcriptional activation in ER+ breast cancer.
Molecular cancer
Jin X,Ge LP,Li DQ,Shao ZM,Di GH,Xu XE,Jiang YZ
Mon May 11 00:00:00 EDT 2020
Mitochondrial UQCC3 Modulates Hypoxia Adaptation by Orchestrating OXPHOS and Glycolysis in Hepatocellular Carcinoma.
Cell reports
Yang Y,Zhang G,Guo F,Li Q,Luo H,Shu Y,Shen Y,Gan J,Xu L,Yang H
Tue Nov 03 00:00:00 EST 2020
ASPM promotes glioblastoma growth by regulating G1 restriction point progression and Wnt-β-catenin signaling.
Aging
Chen X,Huang L,Yang Y,Chen S,Sun J,Ma C,Xie J,Song Y,Yang J
Mon Jan 06 00:00:00 EST 2020
miR-203a suppresses cell proliferation by targeting RING-finger protein 6 in colorectal cancer.
Anti-cancer drugs
Miao J,Hou N,Yang W,Jiang Q,Xue W,Wang X,Zhang H,Xiong X,Wang L,Zhao L,Huang C
Wed Jul 01 00:00:00 EDT 2020
Aldehyde dehydrogenase 2 protects against sympathetic excitation-induced cardiac fibrosis.
Biochemical and biophysical research communications
Yuan Q,Yang F,Dai S,Wang Z,Xu Y,Xu BC,Sun Y,Zheng B,Zhao Y,Wang W,Liu B,Wang J,Cui S,Cao S,Zhang R,Xue L,Wei S,Xue M,Xu T,Xu F,Chen Y
Thu Dec 17 00:00:00 EST 2020
Silencing of ELK3 Induces S-M Phase Arrest and Apoptosis and Upregulates SERPINE1 Expression Reducing Migration in Prostate Cancer Cells.
BioMed research international
Mao Y,Li W,Hua B,Gu X,Pan W,Chen Q,Xu B,Wang Z,Lu C
Thu Nov 19 00:00:00 EST 2020
UBAC2 promotes bladder cancer proliferation through BCRC-3/miRNA-182-5p/p27 axis.
Cell death & disease
Gu C,Zhao K,Zhou N,Liu F,Xie F,Yu S,Feng Y,Chen L,Yang J,Tian F,Jiang G
Thu Sep 10 00:00:00 EDT 2020
Cancer Cell enters reversible quiescence through Intracellular Acidification to resist Paclitaxel Cytotoxicity.
International journal of medical sciences
Jia M,Zheng D,Wang X,Zhang Y,Chen S,Cai X,Mo L,Hu Z,Li H,Zhou Z,Li J
Mon Mar 08 00:00:00 EST 2021
Knockdown of interferon-induced transmembrane protein 1 inhibited proliferation, induced cell cycle arrest and apoptosis, and suppressed MAPK signaling pathway in pancreatic cancer cells.
Bioscience, biotechnology, and biochemistry
Zhang L,Wang Z,Kong D,Zhao X,Chen X,Chai W
Sat Aug 01 00:00:00 EDT 2020
Down-regulation of NTSR3 inhibits cell growth and metastasis, as well as the PI3K-AKT and MAPK signaling pathways in colorectal cancer.
Biochemistry and cell biology = Biochimie et biologie cellulaire
Liu A,Zuo Z,Liu L,Liu L
Thu Oct 01 00:00:00 EDT 2020
BRCA1 protects cardiac microvascular endothelial cells against irradiation by regulating p21-mediated cell cycle arrest.
Life sciences
Zeng ZM,Du HY,Xiong L,Zeng XL,Zhang P,Cai J,Huang L,Liu AW
Sun Mar 01 00:00:00 EST 2020
Inhibiting effect of MicroRNA-3619-5p/PSMD10 axis on liver cancer cell growth in vivo and in vitro.
Life sciences
Hao P,Yue F,Xian X,Ren Q,Cui H,Wang Y
Sat Aug 01 00:00:00 EDT 2020
Loss of hnRNPA2B1 inhibits malignant capability and promotes apoptosis via down-regulating Lin28B expression in ovarian cancer.
Cancer letters
Yang Y,Wei Q,Tang Y,Luo Q,Zhao H,He M,Wang H,Zeng Q,Lu W,Xu J,Liu T,Yi P
Fri Apr 10 00:00:00 EDT 2020
FAM83A Promotes Lung Cancer Progression by Regulating the Wnt and Hippo Signaling Pathways and Indicates Poor Prognosis.
Frontiers in oncology
Zheng YW,Li ZH,Lei L,Liu CC,Wang Z,Fei LR,Yang MQ,Huang WJ,Xu HT
Mon Sep 28 00:00:00 EDT 2020
Ablation of Gsa impairs renal tubule proliferation after injury via CDK2/cyclin E.
American journal of physiology. Renal physiology
Liu L,Deng Y,Cai Y,Lu P,Guo Y,Zhang C,Li Q,Zhang T,Han M,Xu G
Sun Mar 01 00:00:00 EST 2020
TMED3 Promotes Proliferation and Migration in Breast Cancer Cells by Activating Wnt/β-Catenin Signaling.
OncoTargets and therapy
Zhang X,Luo Y,Li Q
Fri Apr 15 00:00:00 EDT 2022
HOTAIR-EZH2 inhibitor AC1Q3QWB upregulates CWF19L1 and enhances cell cycle inhibition of CDK4/6 inhibitor palbociclib in glioma.
Clinical and translational medicine
Shi J,Lv S,Wu M,Wang X,Deng Y,Li Y,Li K,Zhao H,Zhu X,Ye M
Wed Jan 01 00:00:00 EST 2020
MiR-23a-3p promoted G1/S cell cycle transition by targeting protocadherin17 in hepatocellular carcinoma.
Journal of physiology and biochemistry
Xiang Y,Yang Y,Lin C,Wu J,Zhang X
Sat Feb 01 00:00:00 EST 2020
Novel copper complex CTB regulates methionine cycle induced TERT hypomethylation to promote HCC cells senescence via mitochondrial SLC25A26.
Cell death & disease
Jin C,Li Y,Su Y,Guo Z,Wang X,Wang S,Zhang F,Zhang Z,Shao J,Zheng S
Sun Oct 11 00:00:00 EDT 2020
Cell cycle exit during bortezomib-induced osteogenic differentiation of mesenchymal stem cells was mediated by Xbp1s-upregulated p21Cip1 and p27Kip1.
Journal of cellular and molecular medicine
Zhang D,Fan R,Lei L,Lei L,Wang Y,Lv N,Chen P,Williamson RA,Wang B,Hu J
Sat Aug 01 00:00:00 EDT 2020
Compositional characterization of Pyrus ussuriensis Maxim and their antioxidant activities and induction of apoptosis in Bel-7402 cell.
Journal of food biochemistry
Peng F,Li G,Xie Y,Yin HY,Li XJ,Yang YD
Mon Jun 01 00:00:00 EDT 2020
SLC25A10 performs an oncogenic role in human osteosarcoma.
Oncology letters
Wang G,Xia J,Chen C,Qiu J,Sun P,Peng Z,Chen X,Xu B
Thu Oct 01 00:00:00 EDT 2020
Cell adhesion molecule 4 suppresses cell growth and metastasis by inhibiting the Akt signaling pathway in non-small cell lung cancer.
The international journal of biochemistry & cell biology
Luo F,Zhao Y,Liu J
Mon Jun 01 00:00:00 EDT 2020
LINC00963 targeting miR-128-3p promotes acute kidney injury process by activating JAK2/STAT1 pathway.
Journal of cellular and molecular medicine
Xie LB,Chen B,Liao X,Chen YF,Yang R,He SR,Pei LJ,Jiang R
Fri May 01 00:00:00 EDT 2020
Latcripin-7A, derivative of Lentinula edodes C91-3, reduces migration and induces apoptosis, autophagy, and cell cycle arrest at G1 phase in breast cancer cells.
Applied microbiology and biotechnology
Din SRU,Zhong M,Nisar MA,Saleem MZ,Hussain A,Khinsar KH,Alam S,Ayub G,Kanwal S,Li X,Zhang W,Wang X,Ning A,Cao J,Huang M
Tue Dec 01 00:00:00 EST 2020
Minichromosome maintenance protein 5 is an important pathogenic factor of oral squamous cell carcinoma.
Oncology letters
Hao M,Wang H,Zhang C,Li C,Wang X
Thu Oct 01 00:00:00 EDT 2020
circFOXM1 promotes proliferation of non-small cell lung carcinoma cells by acting as a ceRNA to upregulate FAM83D.
Journal of experimental & clinical cancer research : CR
Yu C,Cheng Z,Cui S,Mao X,Li B,Fu Y,Wang H,Jin H,Ye Q,Zhao X,Jiang L,Qin W
Mon Mar 30 00:00:00 EDT 2020
Epigenetic silencing of IGFBPL1 promotes esophageal cancer growth by activating PI3K-AKT signaling.
Clinical epigenetics
Liu Y,Zhang M,He T,Yang W,Wang L,Zhang L,Guo M
Mon Feb 10 00:00:00 EST 2020
PKMYT1 is associated with prostate cancer malignancy and may serve as a therapeutic target.
Gene
Wang J,Wang L,Chen S,Peng H,Xiao L,Liu Y,Lin D,Wang Y,Xu Y,Yang K
Sat Jun 20 00:00:00 EDT 2020
MicroRNA‑145‑5p inhibits osteosarcoma cell proliferation by targeting E2F transcription factor 3.
International journal of molecular medicine
Li H,Pan R,Lu Q,Ren C,Sun J,Wu H,Wen J,Chen H
Fri May 01 00:00:00 EDT 2020
LncRNA IDH1-AS1 suppresses cell proliferation and tumor growth in glioma.
Biochemistry and cell biology = Biochimie et biologie cellulaire
Wang J,Quan Y,Lv J,Dong Q,Gong S
Thu Oct 01 00:00:00 EDT 2020
MiR-377-3p inhibits atherosclerosis-associated vascular smooth muscle cell proliferation and migration via targeting neuropilin2.
Bioscience reports
Wang H,Wei Z,Li H,Guan Y,Han Z,Wang H,Liu B
Fri Jun 26 00:00:00 EDT 2020
Nicotinamide phosphoribosyl transferase regulates cell growth via the Sirt1/P53 signaling pathway and is a prognosis marker in colorectal cancer.
Journal of cellular physiology
Pan JH,Zhou H,Zhu SB,Huang JL,Zhao XX,Ding H,Qin L,Pan YL
Mon Apr 01 00:00:00 EDT 2019
Nicotinamide phosphoribosyl transferase regulates cell growth via the Sirt1/P53 signaling pathway and is a prognosis marker in colorectal cancer.
Journal of cellular physiology
Pan JH,Zhou H,Zhu SB,Huang JL,Zhao XX,Ding H,Qin L,Pan YL
Mon Apr 01 00:00:00 EDT 2019
Baicalin inhibits proliferation and promotes apoptosis of vascular smooth muscle cells by regulating the MEG3/p53 pathway following treatment with ox‑LDL.
International journal of molecular medicine
Liu Y,Jia L,Min D,Xu Y,Zhu J,Sun Z
Fri Feb 01 00:00:00 EST 2019
Baicalin inhibits proliferation and promotes apoptosis of vascular smooth muscle cells by regulating the MEG3/p53 pathway following treatment with ox‑LDL.
International journal of molecular medicine
Liu Y,Jia L,Min D,Xu Y,Zhu J,Sun Z
Fri Feb 01 00:00:00 EST 2019
miR-193a/b-3p relieves hepatic fibrosis and restrains proliferation and activation of hepatic stellate cells.
Journal of cellular and molecular medicine
Ju B,Nie Y,Yang X,Wang X,Li F,Wang M,Wang C,Zhang H
Sat Jun 01 00:00:00 EDT 2019
DEPDC1 drives hepatocellular carcinoma cell proliferation, invasion and angiogenesis by regulating the CCL20/CCR6 signaling pathway.
Oncology reports
Guo W,Li H,Liu H,Ma X,Yang S,Wang Z
Sun Sep 01 00:00:00 EDT 2019
Silencing TTK expression inhibits the proliferation and progression of prostate cancer.
Experimental cell research
Chen S,Wang J,Wang L,Peng H,Xiao L,Li C,Lin D,Yang K
Sun Dec 01 00:00:00 EST 2019
Pterostilbene, An Active Constituent of Blueberries, Suppresses Proliferation Potential of Human Cholangiocarcinoma via Enhancing the Autophagic Flux.
Frontiers in pharmacology
Wang D,Guo H,Yang H,Wang D,Gao P,Wei W
Tue Sep 29 00:00:00 EDT 2020
MicroRNA-191 promotes hepatocellular carcinoma cell proliferation by has_circ_0000204/miR-191/KLF6 axis.
Cell proliferation
Tian F,Yu C,Wu M,Wu X,Wan L,Zhu X
Sun Sep 01 00:00:00 EDT 2019
Upregulation of miR‑95-3p inhibits growth of osteosarcoma by targeting HDGF.
Pathology, research and practice
Liu X,Ma W,Ma J,Xiao L,Hao D
Thu Aug 01 00:00:00 EDT 2019
Rafoxanide promotes apoptosis and autophagy of gastric cancer cells by suppressing PI3K /Akt/mTOR pathway.
Experimental cell research
Liu JZ,Hu YL,Feng Y,Guo YB,Liu YF,Yang JL,Mao QS,Xue WJ
Sun Dec 15 00:00:00 EST 2019
Bruceine D inhibits tumor growth and stem cell-like traits of osteosarcoma through inhibition of STAT3 signaling pathway.
Cancer medicine
Wang S,Hu H,Zhong B,Shi D,Qing X,Cheng C,Deng X,Zhang Z,Shao Z
Sun Dec 01 00:00:00 EST 2019
Fibroblast growth factor-5 promotes spermatogonial stem cell proliferation via ERK and AKT activation.
Stem cell research & therapy
Tian R,Yao C,Yang C,Zhu Z,Li C,Zhi E,Wang J,Li P,Chen H,Yuan Q,He Z,Li Z
Tue Jan 22 00:00:00 EST 2019
Semaphorin 4C Promotes Macrophage Recruitment and Angiogenesis in Breast Cancer.
Molecular cancer research : MCR
Yang J,Zeng Z,Qiao L,Jiang X,Ma J,Wang J,Ye S,Ma Q,Wei J,Wu M,Huang X,Ma D,Gao Q
Tue Oct 01 00:00:00 EDT 2019
Jingfukang induces anti-cancer activity through oxidative stress-mediated DNA damage in circulating human lung cancer cells.
BMC complementary and alternative medicine
Que Z,Zhou Z,Luo B,Dong C,Jiang Y,Li H,Tian J
Wed Aug 07 00:00:00 EDT 2019
Shuttling SLC2A4RG is regulated by 14-3-3θ to modulate cell survival via caspase-3 and caspase-6 in human glioma.
EBioMedicine
Yun D,Wang H,Wang Y,Chen Y,Zhao Z,Ma J,Ji Y,Huang Q,Chen J,Chen H,Lu D
Fri Feb 01 00:00:00 EST 2019
GTSE1 is involved in breast cancer progression in p53 mutation-dependent manner.
Journal of experimental & clinical cancer research : CR
Lin F,Xie YJ,Zhang XK,Huang TJ,Xu HF,Mei Y,Liang H,Hu H,Lin ST,Luo FF,Lang YH,Peng LX,Qian CN,Huang BJ
Mon Apr 08 00:00:00 EDT 2019
Fibronectin regulates the self-renewal of rabbit limbal epithelial stem cells by stimulating the Wnt11/Fzd7/ROCK non-canonical Wnt pathway.
Experimental eye research
Zheng M,Tian C,Fan T,Xu B
Thu Aug 01 00:00:00 EDT 2019
Identification of predictors of drug sensitivity using patient-derived models of esophageal squamous cell carcinoma.
Nature communications
Su D,Zhang D,Jin J,Ying L,Han M,Chen K,Li B,Wu J,Xie Z,Zhang F,Lin Y,Cheng G,Li JY,Huang M,Wang J,Wang K,Zhang J,Li F,Xiong L,Futreal A,Mao W
Thu Nov 07 00:00:00 EST 2019
Cyclin-dependent kinase 1 disruption inhibits angiogenesis by inducing cell cycle arrest and apoptosis.
Experimental and therapeutic medicine
Gao X,Zhang Y,Zhang R,Zhao Z,Zhang H,Wu J,Shen W,Zhong M
Tue Oct 01 00:00:00 EDT 2019
Exogenous biological renal support ameliorates renal pathology after ischemia reperfusion injury in elderly mice.
Aging
Liu D,Yin Z,Huang Q,Ren Y,Li D,Wang L,Cui S,Zhang Y,Ning Y,Lun L,Cai G,Bai X,Sun X,Chen X
Fri Apr 12 00:00:00 EDT 2019
Phenotypic transition of tumor cells between epithelial- and mesenchymal-like state during adaptation to acidosis.
Cell cycle (Georgetown, Tex.)
Zhang Y,Xu L,Wang P,Jian H,Shi X,Jia M,Mo L,Hu Z,Li H,Li J
Thu Aug 01 00:00:00 EDT 2019
CircNR3C1 inhibits proliferation of bladder cancer cells by sponging miR-27a-3p and downregulating cyclin D1 expression.
Cancer letters
Zheng F,Wang M,Li Y,Huang C,Tao D,Xie F,Zhang H,Sun J,Zhang C,Gu C,Wang Z,Jiang G
Sat Sep 28 00:00:00 EDT 2019
MicroRNA-107 induces cell cycle arrests by directly targeting cyclin E1 in ovarian cancer.
Biochemical and biophysical research communications
Tang Z,Fang Y,Du R
Tue Apr 30 00:00:00 EDT 2019
MicroRNA-107 induces cell cycle arrests by directly targeting cyclin E1 in ovarian cancer.
Biochemical and biophysical research communications
Tang Z,Fang Y,Du R
Tue Apr 30 00:00:00 EDT 2019
Enhanced anti-tumor effect of liposomal Fasudil on hepatocellular carcinoma in vitro and in vivo.
PloS one
Zhao Y,Zhang Y,Vazirinejad Mehdiabad M,Zhou K,Chen Y,Li L,Guo J,Xu C
Wed Mar 11 00:00:00 EDT 2020
Motor Neuron and Pancreas Homeobox 1 (MNX1) Is Involved in Promoting Squamous Cervical Cancer Proliferation via Regulating Cyclin E.
Medical science monitor : international medical journal of experimental and clinical research
Xiao L,Hong L,Zheng W
Thu Aug 22 00:00:00 EDT 2019
Small Nucleolar RNA 71A Promotes Lung Cancer Cell Proliferation, Migration and Invasion via MAPK/ERK Pathway.
Journal of Cancer
Tang G,Zeng Z,Sun W,Li S,You C,Tang F,Peng S,Ma S,Luo Y,Xu J,Tian X,Zhang N,Gong Y,Xie C
Thu Oct 01 00:00:00 EDT 2020
Design, synthesis and biological evaluation of novel uracil derivatives bearing 1, 2, 3-triazole moiety as thymidylate synthase (TS) inhibitors and as potential antitumor drugs.
European journal of medicinal chemistry
Lu GQ,Li XY,Mohamed O K,Wang D,Meng FH
Sat Jun 01 00:00:00 EDT 2019
The Capsid Protein VP1 of Coxsackievirus B Induces Cell Cycle Arrest by Up-Regulating Heat Shock Protein 70.
Frontiers in microbiology
Wang Y,Zhao S,Chen Y,Wang T,Dong C,Wo X,Zhang J,Dong Y,Xu W,Feng X,Qu C,Wang Y,Zhong Z,Zhao W
Thu Oct 01 00:00:00 EDT 2020
The effect of MTHFD2 on the proliferation and migration of colorectal cancer cell lines.
OncoTargets and therapy
Wei Y,Liu P,Li Q,Du J,Chen Y,Wang Y,Shi H,Wang Y,Zhang H,Xue W,Gao Y,Li D,Feng Y,Yan J,Han J,Zhang J
Sun Apr 10 00:00:00 EDT 2022
RNA sequencing analysis of the CAL-27 cell response to over-expressed ZNF750 gene revealed an extensive regulation on cell cycle.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu X,Yang Y,Xu C,Yang H,Chen S,Chen H
Tue Oct 01 00:00:00 EDT 2019
Activation of CXCL5-CXCR2 axis promotes proliferation and accelerates G1 to S phase transition of papillary thyroid carcinoma cells and activates JNK and p38 pathways.
Cancer biology & therapy
Cui D,Zhao Y,Xu J
Mon Jul 06 00:00:00 EDT 2020
Short-term stimulation with histone deacetylase inhibitor trichostatin a induces epithelial-mesenchymal transition in nasopharyngeal carcinoma cells without increasing cell invasion ability.
BMC cancer
Shen Z,Liao X,Shao Z,Feng M,Yuan J,Wang S,Gan S,Ha Y,He Z,Jie W
Fri Mar 22 00:00:00 EDT 2019
SND1 promotes the proliferation of osteosarcoma cells by upregulating COX‑2/PGE2 expression via activation of NF‑κB.
Oncology reports
Zhou M,Wang A,Yin B,Wu D,Han S,Zhang W,Liu J,Sun K
Tue Jan 01 00:00:00 EST 2019
Iron-dependent histone 3 lysine 9 demethylation controls B cell proliferation and humoral immune responses.
Nature communications
Jiang Y,Li C,Wu Q,An P,Huang L,Wang J,Chen C,Chen X,Zhang F,Ma L,Liu S,He H,Xie S,Sun Y,Liu H,Zhan Y,Tao Y,Liu Z,Sun X,Hu Y,Wang Q,Ye D,Zhang J,Zou S,Wang Y,Wei G,Liu Y,Shi Y,Eugene Chin Y,Hao Y,Wang F,Zhang X
Wed Jul 03 00:00:00 EDT 2019
Long non‑coding RNA MALAT1 sponges miR‑124‑3p.1/KLF5 to promote pulmonary vascular remodeling and cell cycle progression of pulmonary artery hypertension.
International journal of molecular medicine
Wang D,Xu H,Wu B,Jiang S,Pan H,Wang R,Chen J
Sun Sep 01 00:00:00 EDT 2019
Biocompatible PEGylated Gold nanorods function As cytokinesis inhibitors to suppress angiogenesis.
Biomaterials
Song H,Guo T,Zhao Z,Wei Y,Luo H,Weng W,Zhang R,Zhong M,Chen C,Su J,Shen W
Sat Sep 01 00:00:00 EDT 2018
Biocompatible PEGylated Gold nanorods function As cytokinesis inhibitors to suppress angiogenesis.
Biomaterials
Song H,Guo T,Zhao Z,Wei Y,Luo H,Weng W,Zhang R,Zhong M,Chen C,Su J,Shen W
Sat Sep 01 00:00:00 EDT 2018
Mitochondrial cytochrome P450 (CYP) 1B1 is responsible for melatonin-induced apoptosis in neural cancer cells.
Journal of pineal research
Yu Z,Tian X,Peng Y,Sun Z,Wang C,Tang N,Li B,Jian Y,Wang W,Huo X,Ma X
Wed Aug 01 00:00:00 EDT 2018
Mitochondrial cytochrome P450 (CYP) 1B1 is responsible for melatonin-induced apoptosis in neural cancer cells.
Journal of pineal research
Yu Z,Tian X,Peng Y,Sun Z,Wang C,Tang N,Li B,Jian Y,Wang W,Huo X,Ma X
Wed Aug 01 00:00:00 EDT 2018
Knockdown of PAICS inhibits malignant proliferation of human breast cancer cell lines.
Biological research
Meng M,Chen Y,Jia J,Li L,Yang S
Fri Aug 10 00:00:00 EDT 2018
Knockdown of PAICS inhibits malignant proliferation of human breast cancer cell lines.
Biological research
Meng M,Chen Y,Jia J,Li L,Yang S
Fri Aug 10 00:00:00 EDT 2018
MCUR1-Mediated Mitochondrial Calcium Signaling Facilitates Cell Survival of Hepatocellular Carcinoma via Reactive Oxygen Species-Dependent P53 Degradation.
Antioxidants & redox signaling
Ren T,Wang J,Zhang H,Yuan P,Zhu J,Wu Y,Huang Q,Guo X,Zhang J,Ji L,Li J,Zhang H,Yang H,Xing J
Fri Apr 20 00:00:00 EDT 2018
MCUR1-Mediated Mitochondrial Calcium Signaling Facilitates Cell Survival of Hepatocellular Carcinoma via Reactive Oxygen Species-Dependent P53 Degradation.
Antioxidants & redox signaling
Ren T,Wang J,Zhang H,Yuan P,Zhu J,Wu Y,Huang Q,Guo X,Zhang J,Ji L,Li J,Zhang H,Yang H,Xing J
Fri Apr 20 00:00:00 EDT 2018
ANP32E induces tumorigenesis of triple-negative breast cancer cells by upregulating E2F1.
Molecular oncology
Xiong Z,Ye L,Zhenyu H,Li F,Xiong Y,Lin C,Wu X,Deng G,Shi W,Song L,Yuan Z,Wang X
Fri Jun 01 00:00:00 EDT 2018
ANP32E induces tumorigenesis of triple-negative breast cancer cells by upregulating E2F1.
Molecular oncology
Xiong Z,Ye L,Zhenyu H,Li F,Xiong Y,Lin C,Wu X,Deng G,Shi W,Song L,Yuan Z,Wang X
Fri Jun 01 00:00:00 EDT 2018
Nesfatin-1 promotes VSMC migration and neointimal hyperplasia by upregulating matrix metalloproteinases and downregulating PPARγ.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang JR,Lu QB,Feng WB,Wang HP,Tang ZH,Cheng H,Du Q,Wang YB,Li KX,Sun HJ
Fri Jun 01 00:00:00 EDT 2018
Nesfatin-1 promotes VSMC migration and neointimal hyperplasia by upregulating matrix metalloproteinases and downregulating PPARγ.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang JR,Lu QB,Feng WB,Wang HP,Tang ZH,Cheng H,Du Q,Wang YB,Li KX,Sun HJ
Fri Jun 01 00:00:00 EDT 2018
8-Cetylberberine inhibits growth of lung cancer in vitro and in vivo.
Life sciences
Xiao Y,Tian C,Huang T,Han B,Wang M,Ma H,Li Z,Ye X,Li X
Mon Jan 01 00:00:00 EST 2018
8-Cetylberberine inhibits growth of lung cancer in vitro and in vivo.
Life sciences
Xiao Y,Tian C,Huang T,Han B,Wang M,Ma H,Li Z,Ye X,Li X
Mon Jan 01 00:00:00 EST 2018
NF-κB and JNK mediated apoptosis and G0/G1 arrest of HeLa cells induced by rubiarbonol G, an arborinane-type triterpenoid from Rubia yunnanensis.
Journal of ethnopharmacology
Zeng GZ,Wang Z,Zhao LM,Fan JT,Tan NH
Thu Jun 28 00:00:00 EDT 2018
NF-κB and JNK mediated apoptosis and G0/G1 arrest of HeLa cells induced by rubiarbonol G, an arborinane-type triterpenoid from Rubia yunnanensis.
Journal of ethnopharmacology
Zeng GZ,Wang Z,Zhao LM,Fan JT,Tan NH
Thu Jun 28 00:00:00 EDT 2018
VEGFC/VEGFR3 Signaling Regulates Mouse Spermatogonial Cell Proliferation via the Activation of AKT/MAPK and Cyclin D1 Pathway and Mediates the Apoptosis by affecting Caspase 3/9 and Bcl-2.
Cell cycle (Georgetown, Tex.)
Zhao L,Zhu Z,Yao C,Huang Y,Zhi E,Chen H,Tian R,Li P,Yuan Q,Xue Y,Wan Z,Yang C,Gong Y,He Z,Li Z
Thu Aug 29 00:00:00 EDT 2019
VEGFC/VEGFR3 Signaling Regulates Mouse Spermatogonial Cell Proliferation via the Activation of AKT/MAPK and Cyclin D1 Pathway and Mediates the Apoptosis by affecting Caspase 3/9 and Bcl-2.
Cell cycle (Georgetown, Tex.)
Zhao L,Zhu Z,Yao C,Huang Y,Zhi E,Chen H,Tian R,Li P,Yuan Q,Xue Y,Wan Z,Yang C,Gong Y,He Z,Li Z
Thu Aug 29 00:00:00 EDT 2019
Antifibrotic cardioprotection of berberine via downregulating myocardial IGF-1 receptor-regulated MMP-2/MMP-9 expression in diabetic rats.
American journal of physiology. Heart and circulatory physiology
Li G,Xing W,Zhang M,Geng F,Yang H,Zhang H,Zhang X,Li J,Dong L,Gao F
Mon Oct 01 00:00:00 EDT 2018
Antifibrotic cardioprotection of berberine via downregulating myocardial IGF-1 receptor-regulated MMP-2/MMP-9 expression in diabetic rats.
American journal of physiology. Heart and circulatory physiology
Li G,Xing W,Zhang M,Geng F,Yang H,Zhang H,Zhang X,Li J,Dong L,Gao F
Mon Oct 01 00:00:00 EDT 2018
miR-155 accelerates proliferation of mouse hepatocytes during liver regeneration by directly targeting SOCS1.
American journal of physiology. Gastrointestinal and liver physiology
Lin X,Chen L,Li H,Liu Y,Guan Y,Li X,Jia Z,Lin X,Jia J,Sun Y,Xiao D
Mon Oct 01 00:00:00 EDT 2018
miR-155 accelerates proliferation of mouse hepatocytes during liver regeneration by directly targeting SOCS1.
American journal of physiology. Gastrointestinal and liver physiology
Lin X,Chen L,Li H,Liu Y,Guan Y,Li X,Jia Z,Lin X,Jia J,Sun Y,Xiao D
Mon Oct 01 00:00:00 EDT 2018
Identification of Grade-associated MicroRNAs in Brainstem Gliomas Based on Microarray Data.
Journal of Cancer
Chen X,Dong D,Pan C,Xu C,Sun Y,Geng Y,Kong L,Xiao X,Zhao Z,Zhou W,Huang L,Song Y,Zhang L
Thu Oct 01 00:00:00 EDT 2020
Identification of Grade-associated MicroRNAs in Brainstem Gliomas Based on Microarray Data.
Journal of Cancer
Chen X,Dong D,Pan C,Xu C,Sun Y,Geng Y,Kong L,Xiao X,Zhao Z,Zhou W,Huang L,Song Y,Zhang L
Thu Oct 01 00:00:00 EDT 2020
Actin-capping protein CapG is associated with prognosis, proliferation and metastasis in human glioma.
Oncology reports
Yun DP,Wang YQ,Meng DL,Ji YY,Chen JX,Chen HY,Lu DR
Thu Mar 01 00:00:00 EST 2018
Actin-capping protein CapG is associated with prognosis, proliferation and metastasis in human glioma.
Oncology reports
Yun DP,Wang YQ,Meng DL,Ji YY,Chen JX,Chen HY,Lu DR
Thu Mar 01 00:00:00 EST 2018
HOXA5 is a tumor suppressor gene that is decreased in gastric cancer.
Oncology reports
Peng X,Zha L,Chen A,Wang Z
Sat Sep 01 00:00:00 EDT 2018
HOXA5 is a tumor suppressor gene that is decreased in gastric cancer.
Oncology reports
Peng X,Zha L,Chen A,Wang Z
Sat Sep 01 00:00:00 EDT 2018
The sialyltransferase ST3Gal3 facilitates the receptivity of the uterine endometrium in vitro and in vivo.
FEBS letters
Yu M,Wang H,Liu J,Qin H,Liu S,Yan Q
Thu Nov 01 00:00:00 EDT 2018
The sialyltransferase ST3Gal3 facilitates the receptivity of the uterine endometrium in vitro and in vivo.
FEBS letters
Yu M,Wang H,Liu J,Qin H,Liu S,Yan Q
Thu Nov 01 00:00:00 EDT 2018
Overexpression of SMARCA2 or CAMK2D is associated with cisplatin resistance in human epithelial ovarian cancer.
Oncology letters
Xu X,Zheng Z,Jia L,Suo S,Liu B,Shao T,Tu Q,Hua Y,Xu H
Sat Sep 01 00:00:00 EDT 2018
Overexpression of SMARCA2 or CAMK2D is associated with cisplatin resistance in human epithelial ovarian cancer.
Oncology letters
Xu X,Zheng Z,Jia L,Suo S,Liu B,Shao T,Tu Q,Hua Y,Xu H
Sat Sep 01 00:00:00 EDT 2018
Coxsackievirus A6 Induces Cell Cycle Arrest in G0/G1 Phase for Viral Production.
Frontiers in cellular and infection microbiology
Wang Z,Wang Y,Wang S,Meng X,Song F,Huo W,Zhang S,Chang J,Li J,Zheng B,Liu Y,Zhang Y,Zhang W,Yu J
Fri Aug 16 00:00:00 EDT 2019
Coxsackievirus A6 Induces Cell Cycle Arrest in G0/G1 Phase for Viral Production.
Frontiers in cellular and infection microbiology
Wang Z,Wang Y,Wang S,Meng X,Song F,Huo W,Zhang S,Chang J,Li J,Zheng B,Liu Y,Zhang Y,Zhang W,Yu J
Fri Aug 16 00:00:00 EDT 2019
Arachidonic acid induces macrophage cell cycle arrest through the JNK signaling pathway.
Lipids in health and disease
Shen Z,Ma Y,Ji Z,Hao Y,Yan X,Zhong Y,Tang X,Ren W
Fri Feb 09 00:00:00 EST 2018
Arachidonic acid induces macrophage cell cycle arrest through the JNK signaling pathway.
Lipids in health and disease
Shen Z,Ma Y,Ji Z,Hao Y,Yan X,Zhong Y,Tang X,Ren W
Fri Feb 09 00:00:00 EST 2018
HMGA1 exacerbates tumor growth through regulating the cell cycle and accelerates migration/invasion via targeting miR-221/222 in cervical cancer.
Cell death & disease
Fu F,Wang T,Wu Z,Feng Y,Wang W,Zhou S,Ma X,Wang S
Tue May 22 00:00:00 EDT 2018
HMGA1 exacerbates tumor growth through regulating the cell cycle and accelerates migration/invasion via targeting miR-221/222 in cervical cancer.
Cell death & disease
Fu F,Wang T,Wu Z,Feng Y,Wang W,Zhou S,Ma X,Wang S
Tue May 22 00:00:00 EDT 2018
HMGA1 exacerbates tumor growth through regulating the cell cycle and accelerates migration/invasion via targeting miR-221/222 in cervical cancer.
Cell death & disease
Fu F,Wang T,Wu Z,Feng Y,Wang W,Zhou S,Ma X,Wang S
Tue May 22 00:00:00 EDT 2018
HMGA1 exacerbates tumor growth through regulating the cell cycle and accelerates migration/invasion via targeting miR-221/222 in cervical cancer.
Cell death & disease
Fu F,Wang T,Wu Z,Feng Y,Wang W,Zhou S,Ma X,Wang S
Tue May 22 00:00:00 EDT 2018
Overexpression of Annexin A2 Receptor Inhibits Neovascularization via the Promotion of Krüppel-Like Transcription Factor 2.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Guo T,Song H,Zhao Z,Qi Z,Zhao S
Thu Jul 05 00:00:00 EDT 2018
Overexpression of Annexin A2 Receptor Inhibits Neovascularization via the Promotion of Krüppel-Like Transcription Factor 2.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Guo T,Song H,Zhao Z,Qi Z,Zhao S
Thu Jul 05 00:00:00 EDT 2018
Deficiency of VCP-Interacting Membrane Selenoprotein (VIMP) Leads to G1 Cell Cycle Arrest and Cell Death in MIN6 Insulinoma Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Men L,Sun J,Ren D
Fri Jan 25 00:00:00 EST 2019
Deficiency of VCP-Interacting Membrane Selenoprotein (VIMP) Leads to G1 Cell Cycle Arrest and Cell Death in MIN6 Insulinoma Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Men L,Sun J,Ren D
Fri Jan 25 00:00:00 EST 2019
Motor neuron and pancreas homeobox 1/HLXB9 promotes sustained proliferation in bladder cancer by upregulating CCNE1/2.
Journal of experimental & clinical cancer research : CR
Chen M,Wu R,Li G,Liu C,Tan L,Xiao K,Ye Y,Qin Z
Mon Jul 16 00:00:00 EDT 2018
Motor neuron and pancreas homeobox 1/HLXB9 promotes sustained proliferation in bladder cancer by upregulating CCNE1/2.
Journal of experimental & clinical cancer research : CR
Chen M,Wu R,Li G,Liu C,Tan L,Xiao K,Ye Y,Qin Z
Mon Jul 16 00:00:00 EDT 2018
Circular RNA BCRC-3 suppresses bladder cancer proliferation through miR-182-5p/p27 axis.
Molecular cancer
Xie F,Li Y,Wang M,Huang C,Tao D,Zheng F,Zhang H,Zeng F,Xiao X,Jiang G
Wed Oct 03 00:00:00 EDT 2018
Circular RNA BCRC-3 suppresses bladder cancer proliferation through miR-182-5p/p27 axis.
Molecular cancer
Xie F,Li Y,Wang M,Huang C,Tao D,Zheng F,Zhang H,Zeng F,Xiao X,Jiang G
Wed Oct 03 00:00:00 EDT 2018
IL-2Rα up-regulation is mediated by latent membrane protein 1 and promotes lymphomagenesis and chemotherapy resistance in natural killer/T-cell lymphoma.
Cancer communications (London, England)
Wang L,Bi XW,Zhu YJ,He YZ,Lai QY,Xia ZJ,Cai QQ
Fri Oct 19 00:00:00 EDT 2018
IL-2Rα up-regulation is mediated by latent membrane protein 1 and promotes lymphomagenesis and chemotherapy resistance in natural killer/T-cell lymphoma.
Cancer communications (London, England)
Wang L,Bi XW,Zhu YJ,He YZ,Lai QY,Xia ZJ,Cai QQ
Fri Oct 19 00:00:00 EDT 2018
(-)-Curine induces cell cycle arrest and cell death in hepatocellular carcinoma cells in a p53-independent way.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Gong S,Xu D,Zou F,Peng R
Mon May 01 00:00:00 EDT 2017
(-)-Curine induces cell cycle arrest and cell death in hepatocellular carcinoma cells in a p53-independent way.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Gong S,Xu D,Zou F,Peng R
Mon May 01 00:00:00 EDT 2017
Methylation of ZNF331 is an independent prognostic marker of colorectal cancer and promotes colorectal cancer growth.
Clinical epigenetics
Wang Y,He T,Herman JG,Linghu E,Yang Y,Fuks F,Zhou F,Song L,Guo M
Fri Jul 13 00:00:00 EDT 2018
Methylation of ZNF331 is an independent prognostic marker of colorectal cancer and promotes colorectal cancer growth.
Clinical epigenetics
Wang Y,He T,Herman JG,Linghu E,Yang Y,Fuks F,Zhou F,Song L,Guo M
Fri Jul 13 00:00:00 EDT 2018
The role of lncRNA MALAT1 in the regulation of hepatocyte proliferation during liver regeneration.
International journal of molecular medicine
Li C,Chang L,Chen Z,Liu Z,Wang Y,Ye Q
Wed Feb 01 00:00:00 EST 2017
Pseudolaric acid B inhibits neuroglioma cell proliferation through DNA damage response.
Oncology reports
Song F,Yu X,Zhang H,Wang Z,Wang Y,Meng X,Yu J
Sun Oct 01 00:00:00 EDT 2017
Pseudolaric acid B inhibits neuroglioma cell proliferation through DNA damage response.
Oncology reports
Song F,Yu X,Zhang H,Wang Z,Wang Y,Meng X,Yu J
Sun Oct 01 00:00:00 EDT 2017
Identification, expression pattern and functional characterization of As-kip2 in diapause embryo restarting process of Artemia sinica.
Gene
Zhang M,Yao F,Qin T,Hou L,Zou X
Sat Apr 15 00:00:00 EDT 2017
Identification, expression pattern and functional characterization of As-kip2 in diapause embryo restarting process of Artemia sinica.
Gene
Zhang M,Yao F,Qin T,Hou L,Zou X
Sat Apr 15 00:00:00 EDT 2017
miR-873 suppresses H9C2 cardiomyocyte proliferation by targeting GLI1.
Gene
Zhang JS,Zhao Y,Lv Y,Liu PY,Ruan JX,Sun YL,Gong TX,Wan N,Qiu GR
Wed Aug 30 00:00:00 EDT 2017
miR-873 suppresses H9C2 cardiomyocyte proliferation by targeting GLI1.
Gene
Zhang JS,Zhao Y,Lv Y,Liu PY,Ruan JX,Sun YL,Gong TX,Wan N,Qiu GR
Wed Aug 30 00:00:00 EDT 2017
Cloning, expression pattern, and potential role of apoptosis inhibitor 5 in the termination of embryonic diapause and early embryo development of Artemia sinica.
Gene
Zhang S,Yao F,Jing T,Zhang M,Zhao W,Zou X,Sui L,Hou L
Sun Sep 10 00:00:00 EDT 2017
EDIL3 knockdown inhibits retinal angiogenesis through the induction of cell cycle arrest in vitro.
Molecular medicine reports
Shen W,Zhu S,Qin H,Zhong M,Wu J,Zhang R,Song H
Sun Oct 01 00:00:00 EDT 2017
Rhodium (II) complex with 2-benzoylpyridine, a novel potential chemotherapeutic drug, induces cell cycle arrest and apoptosis in HepG2 cells.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine
Wang X,Li Y,Lin M,Jin J,Huang Z
Fri Dec 01 00:00:00 EST 2017
Rhodium (II) complex with 2-benzoylpyridine, a novel potential chemotherapeutic drug, induces cell cycle arrest and apoptosis in HepG2 cells.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine
Wang X,Li Y,Lin M,Jin J,Huang Z
Fri Dec 01 00:00:00 EST 2017
Lnc-ATB contributes to gastric cancer growth through a MiR-141-3p/TGFβ2 feedback loop.
Biochemical and biophysical research communications
Lei K,Liang X,Gao Y,Xu B,Xu Y,Li Y,Tao Y,Shi W,Liu J
Sat Mar 11 00:00:00 EST 2017
Lnc-ATB contributes to gastric cancer growth through a MiR-141-3p/TGFβ2 feedback loop.
Biochemical and biophysical research communications
Lei K,Liang X,Gao Y,Xu B,Xu Y,Li Y,Tao Y,Shi W,Liu J
Sat Mar 11 00:00:00 EST 2017
Coptisine from Rhizoma Coptidis Suppresses HCT-116 Cells-related Tumor Growth in vitro and in vivo.
Scientific reports
Huang T,Xiao Y,Yi L,Li L,Wang M,Tian C,Ma H,He K,Wang Y,Han B,Ye X,Li X
Mon Feb 06 00:00:00 EST 2017
Coptisine from Rhizoma Coptidis Suppresses HCT-116 Cells-related Tumor Growth in vitro and in vivo.
Scientific reports
Huang T,Xiao Y,Yi L,Li L,Wang M,Tian C,Ma H,He K,Wang Y,Han B,Ye X,Li X
Mon Feb 06 00:00:00 EST 2017
Salusin-β mediates high glucose-induced endothelial injury via disruption of AMPK signaling pathway.
Biochemical and biophysical research communications
Zhu X,Zhou Y,Cai W,Sun H,Qiu L
Sat Sep 16 00:00:00 EDT 2017
Human Enterovirus 68 Interferes with the Host Cell Cycle to Facilitate Viral Production.
Frontiers in cellular and infection microbiology
Wang ZY,Zhong T,Wang Y,Song FM,Yu XF,Xing LP,Zhang WY,Yu JH,Hua SC,Yu XF
Thu Jun 29 00:00:00 EDT 2017
Human Enterovirus 68 Interferes with the Host Cell Cycle to Facilitate Viral Production.
Frontiers in cellular and infection microbiology
Wang ZY,Zhong T,Wang Y,Song FM,Yu XF,Xing LP,Zhang WY,Yu JH,Hua SC,Yu XF
Thu Jun 29 00:00:00 EDT 2017
MiR-16-5p mediates a positive feedback loop in EV71-induced apoptosis and suppresses virus replication.
Scientific reports
Zheng C,Zheng Z,Sun J,Zhang Y,Wei C,Ke X,Liu Y,Deng L,Wang H
Mon Nov 27 00:00:00 EST 2017
MiR-16-5p mediates a positive feedback loop in EV71-induced apoptosis and suppresses virus replication.
Scientific reports
Zheng C,Zheng Z,Sun J,Zhang Y,Wei C,Ke X,Liu Y,Deng L,Wang H
Mon Nov 27 00:00:00 EST 2017
MiR-16-5p mediates a positive feedback loop in EV71-induced apoptosis and suppresses virus replication.
Scientific reports
Zheng C,Zheng Z,Sun J,Zhang Y,Wei C,Ke X,Liu Y,Deng L,Wang H
Mon Nov 27 00:00:00 EST 2017
CAPE-pNO2 ameliorated diabetic nephropathy through regulating the Akt/NF-κB/ iNOS pathway in STZ-induced diabetic mice.
Oncotarget
Wang X,Li D,Fan L,Xiao Q,Zuo H,Li Z
Fri Dec 29 00:00:00 EST 2017
CAPE-pNO2 ameliorated diabetic nephropathy through regulating the Akt/NF-κB/ iNOS pathway in STZ-induced diabetic mice.
Oncotarget
Wang X,Li D,Fan L,Xiao Q,Zuo H,Li Z
Fri Dec 29 00:00:00 EST 2017
Methylation of DACT2 promotes breast cancer development by activating Wnt signaling.
Scientific reports
Li J,Zhang M,He T,Li H,Cao T,Zheng L,Guo M
Mon Jun 12 00:00:00 EDT 2017
Methylation of DACT2 promotes breast cancer development by activating Wnt signaling.
Scientific reports
Li J,Zhang M,He T,Li H,Cao T,Zheng L,Guo M
Mon Jun 12 00:00:00 EDT 2017
Methylation of SLFN11 is a marker of poor prognosis and cisplatin resistance in colorectal cancer.
Epigenomics
He T,Zhang M,Zheng R,Zheng S,Linghu E,Herman JG,Guo M
Thu Jun 01 00:00:00 EDT 2017
Methylation of SLFN11 is a marker of poor prognosis and cisplatin resistance in colorectal cancer.
Epigenomics
He T,Zhang M,Zheng R,Zheng S,Linghu E,Herman JG,Guo M
Thu Jun 01 00:00:00 EDT 2017
LSD1 is Required for Hair Cell Regeneration in Zebrafish.
Molecular neurobiology
He Y,Tang D,Cai C,Chai R,Li H
Sun May 01 00:00:00 EDT 2016
LSD1 is Required for Hair Cell Regeneration in Zebrafish.
Molecular neurobiology
He Y,Tang D,Cai C,Chai R,Li H
Sun May 01 00:00:00 EDT 2016
The AhR is involved in the regulation of LoVo cell proliferation through cell cycle-associated proteins.
Cell biology international
Yin J,Sheng B,Han B,Pu A,Yang K,Li P,Wang Q,Xiao W,Yang H
Sun May 01 00:00:00 EDT 2016
The AhR is involved in the regulation of LoVo cell proliferation through cell cycle-associated proteins.
Cell biology international
Yin J,Sheng B,Han B,Pu A,Yang K,Li P,Wang Q,Xiao W,Yang H
Sun May 01 00:00:00 EDT 2016
Shikonin induces apoptosis of HaCaT cells via the mitochondrial, Erk and Akt pathways.
Molecular medicine reports
Jing H,Sun W,Fan J,Zhang Y,Yang J,Jia J,Li J,Guo J,Luo S,Zheng Y
Fri Apr 01 00:00:00 EDT 2016
Shikonin induces apoptosis of HaCaT cells via the mitochondrial, Erk and Akt pathways.
Molecular medicine reports
Jing H,Sun W,Fan J,Zhang Y,Yang J,Jia J,Li J,Guo J,Luo S,Zheng Y
Fri Apr 01 00:00:00 EDT 2016
Carfilzomib induces G2/M cell cycle arrest in human endometrial cancer cells via upregulation of p21Waf1/Cip1 and p27Kip1.
Taiwanese journal of obstetrics & gynecology
Zhou Y,Wang K,Zhen S,Wang R,Luo W
Thu Dec 01 00:00:00 EST 2016
Carfilzomib induces G2/M cell cycle arrest in human endometrial cancer cells via upregulation of p21Waf1/Cip1 and p27Kip1.
Taiwanese journal of obstetrics & gynecology
Zhou Y,Wang K,Zhen S,Wang R,Luo W
Thu Dec 01 00:00:00 EST 2016
PDIA6 promotes the proliferation of HeLa cells through activating the Wnt/β-catenin signaling pathway.
Oncotarget
Gao H,Sun B,Fu H,Chi X,Wang F,Qi X,Hu J,Shao S
Tue Aug 16 00:00:00 EDT 2016
PDIA6 promotes the proliferation of HeLa cells through activating the Wnt/β-catenin signaling pathway.
Oncotarget
Gao H,Sun B,Fu H,Chi X,Wang F,Qi X,Hu J,Shao S
Tue Aug 16 00:00:00 EDT 2016
Upregulation of E2F8 promotes cell proliferation and tumorigenicity in breast cancer by modulating G1/S phase transition.
Oncotarget
Ye L,Guo L,He Z,Wang X,Lin C,Zhang X,Wu S,Bao Y,Yang Q,Song L,Lin H
Tue Apr 26 00:00:00 EDT 2016
Upregulation of E2F8 promotes cell proliferation and tumorigenicity in breast cancer by modulating G1/S phase transition.
Oncotarget
Ye L,Guo L,He Z,Wang X,Lin C,Zhang X,Wu S,Bao Y,Yang Q,Song L,Lin H
Tue Apr 26 00:00:00 EDT 2016
Inhibition of tumor suppressor p53 preserves glycation-serum induced pancreatic beta-cell demise.
Endocrine
Li Y,Zhang T,Huang Q,Sun Y,Chang X,Zhang H,Zhu Y,Han X
Tue Nov 01 00:00:00 EDT 2016
Down-regulation of dihydrofolate reductase inhibits the growth of endothelial EA.hy926 cell through induction of G1 cell cycle arrest via up-regulating p53 and p21(waf1/cip1) expression.
Journal of clinical biochemistry and nutrition
Fei Z,Gao Y,Qiu M,Qi X,Dai Y,Wang S,Quan Z,Liu Y,Ou J
Tue Mar 01 00:00:00 EST 2016
Paper-based microreactor array for rapid screening of cell signaling cascades.
Lab on a chip
Huang CH,Lei KF,Tsang NM
Sun Aug 07 00:00:00 EDT 2016
Paper-based microreactor array for rapid screening of cell signaling cascades.
Lab on a chip
Huang CH,Lei KF,Tsang NM
Sun Aug 07 00:00:00 EDT 2016
PRMT5 competitively binds to CDK4 to promote G1-S transition upon glucose induction in hepatocellular carcinoma.
Oncotarget
Yang H,Zhao X,Zhao L,Liu L,Li J,Jia W,Liu J,Huang G
Tue Nov 01 00:00:00 EDT 2016
PRMT5 competitively binds to CDK4 to promote G1-S transition upon glucose induction in hepatocellular carcinoma.
Oncotarget
Yang H,Zhao X,Zhao L,Liu L,Li J,Jia W,Liu J,Huang G
Tue Nov 01 00:00:00 EDT 2016
Drp1-mediated mitochondrial fission promotes cell proliferation through crosstalk of p53 and NF-κB pathways in hepatocellular carcinoma.
Oncotarget
Zhan L,Cao H,Wang G,Lyu Y,Sun X,An J,Wu Z,Huang Q,Liu B,Xing J
Tue Oct 04 00:00:00 EDT 2016
Drp1-mediated mitochondrial fission promotes cell proliferation through crosstalk of p53 and NF-κB pathways in hepatocellular carcinoma.
Oncotarget
Zhan L,Cao H,Wang G,Lyu Y,Sun X,An J,Wu Z,Huang Q,Liu B,Xing J
Tue Oct 04 00:00:00 EDT 2016
Inhibition of REST Suppresses Proliferation and Migration in Glioblastoma Cells.
International journal of molecular sciences
Zhang D,Li Y,Wang R,Li Y,Shi P,Kan Z,Pang X
Tue May 03 00:00:00 EDT 2016
Inhibition of REST Suppresses Proliferation and Migration in Glioblastoma Cells.
International journal of molecular sciences
Zhang D,Li Y,Wang R,Li Y,Shi P,Kan Z,Pang X
Tue May 03 00:00:00 EDT 2016
Novel taspine derivative 12k inhibits cell growth and induces apoptosis in lung cell carcinoma.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Dai B,Wang W,Liu R,Wang H,Zhang Y
Sun Mar 01 00:00:00 EST 2015
Novel taspine derivative 12k inhibits cell growth and induces apoptosis in lung cell carcinoma.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Dai B,Wang W,Liu R,Wang H,Zhang Y
Sun Mar 01 00:00:00 EST 2015
Tetrandrine induces G1/S cell cycle arrest through the ROS/Akt pathway in EOMA cells and inhibits angiogenesis in vivo.
International journal of oncology
Xiao W,Jiang Y,Men Q,Yuan L,Huang Z,Liu T,Li W,Liu X
Thu Jan 01 00:00:00 EST 2015
Tetrandrine induces G1/S cell cycle arrest through the ROS/Akt pathway in EOMA cells and inhibits angiogenesis in vivo.
International journal of oncology
Xiao W,Jiang Y,Men Q,Yuan L,Huang Z,Liu T,Li W,Liu X
Thu Jan 01 00:00:00 EST 2015
Down-regulation of cancer/testis antigen OY-TES-1 attenuates malignant behaviors of hepatocellular carcinoma cells in vitro.
International journal of clinical and experimental pathology
Fu J,Luo B,Guo WW,Zhang QM,Shi L,Hu QP,Chen F,Xiao SW,Xie XX
Wed Dec 14 00:00:00 EST 2016
Down-regulation of cancer/testis antigen OY-TES-1 attenuates malignant behaviors of hepatocellular carcinoma cells in vitro.
International journal of clinical and experimental pathology
Fu J,Luo B,Guo WW,Zhang QM,Shi L,Hu QP,Chen F,Xiao SW,Xie XX
Wed Dec 14 00:00:00 EST 2016
Eupolyphaga sinensis Walker inhibits human chronic myeloid leukemia cell K562 growth by inducing G2-M phase cell cycle arrest and targeting EGFR signaling pathway and in S180 tumor-bearing mice.
Environmental toxicology and pharmacology
Dai B,Zhan Y,Qi J,Zhang Y
Thu May 01 00:00:00 EDT 2014
Eupolyphaga sinensis Walker inhibits human chronic myeloid leukemia cell K562 growth by inducing G2-M phase cell cycle arrest and targeting EGFR signaling pathway and in S180 tumor-bearing mice.
Environmental toxicology and pharmacology
Dai B,Zhan Y,Qi J,Zhang Y
Thu May 01 00:00:00 EDT 2014
Notch1 is a potential therapeutic target for the treatment of human hepatitis B virus X protein-associated hepatocellular carcinoma.
Oncology reports
Sun Q,Wang R,Wang Y,Luo J,Wang P,Cheng B
Sat Feb 01 00:00:00 EST 2014
Notch1 is a potential therapeutic target for the treatment of human hepatitis B virus X protein-associated hepatocellular carcinoma.
Oncology reports
Sun Q,Wang R,Wang Y,Luo J,Wang P,Cheng B
Sat Feb 01 00:00:00 EST 2014
Eupolyphaga sinensis walker displays inhibition on hepatocellular carcinoma through regulating cell growth and metastasis signaling.
Scientific reports
Zhang Y,Zhan Y,Zhang D,Dai B,Ma W,Qi J,Liu R,He L
Tue Jul 01 00:00:00 EDT 2014
Eupolyphaga sinensis walker displays inhibition on hepatocellular carcinoma through regulating cell growth and metastasis signaling.
Scientific reports
Zhang Y,Zhan Y,Zhang D,Dai B,Ma W,Qi J,Liu R,He L
Tue Jul 01 00:00:00 EDT 2014
Ginsenoside Rg3 inhibits melanoma cell proliferation through down-regulation of histone deacetylase 3 (HDAC3) and increase of p53 acetylation.
PloS one
Shan X,Fu YS,Aziz F,Wang XQ,Yan Q,Liu JW
Thu Dec 17 00:00:00 EST 2015
Ginsenoside Rg3 inhibits melanoma cell proliferation through down-regulation of histone deacetylase 3 (HDAC3) and increase of p53 acetylation.
PloS one
Shan X,Fu YS,Aziz F,Wang XQ,Yan Q,Liu JW
Thu Dec 17 00:00:00 EST 2015
Sj7170, a unique dual-function peptide with a specific α-chymotrypsin inhibitory activity and a potent tumor-activating effect from scorpion venom.
The Journal of biological chemistry
Song Y,Gong K,Yan H,Hong W,Wang L,Wu Y,Li W,Li W,Cao Z
Fri Apr 25 00:00:00 EDT 2014
Sj7170, a unique dual-function peptide with a specific α-chymotrypsin inhibitory activity and a potent tumor-activating effect from scorpion venom.
The Journal of biological chemistry
Song Y,Gong K,Yan H,Hong W,Wang L,Wu Y,Li W,Li W,Cao Z
Fri Apr 25 00:00:00 EDT 2014
C1orf61 acts as a tumor activator in human hepatocellular carcinoma and is associated with tumorigenesis and metastasis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Hu HM,Chen Y,Liu L,Zhang CG,Wang W,Gong K,Huang Z,Guo MX,Li WX,Li W
Tue Jan 01 00:00:00 EST 2013
C1orf61 acts as a tumor activator in human hepatocellular carcinoma and is associated with tumorigenesis and metastasis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Hu HM,Chen Y,Liu L,Zhang CG,Wang W,Gong K,Huang Z,Guo MX,Li WX,Li W
Tue Jan 01 00:00:00 EST 2013
Brucine, an effective natural compound derived from nux-vomica, induces G1 phase arrest and apoptosis in LoVo cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Zheng L,Wang X,Luo W,Zhan Y,Zhang Y
Thu Aug 01 00:00:00 EDT 2013
Brucine, an effective natural compound derived from nux-vomica, induces G1 phase arrest and apoptosis in LoVo cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Zheng L,Wang X,Luo W,Zhan Y,Zhang Y
Thu Aug 01 00:00:00 EDT 2013
The tumor-suppressive miR-497-195 cluster targets multiple cell-cycle regulators in hepatocellular carcinoma.
PloS one
Furuta M,Kozaki K,Tanimoto K,Tanaka S,Arii S,Shimamura T,Niida A,Miyano S,Inazawa J
Tue Sep 17 00:00:00 EDT 2013
The tumor-suppressive miR-497-195 cluster targets multiple cell-cycle regulators in hepatocellular carcinoma.
PloS one
Furuta M,Kozaki K,Tanimoto K,Tanaka S,Arii S,Shimamura T,Niida A,Miyano S,Inazawa J
Tue Sep 17 00:00:00 EDT 2013
CS055 (Chidamide/HBI-8000), a novel histone deacetylase inhibitor, induces G1 arrest, ROS-dependent apoptosis and differentiation in human leukaemia cells.
The Biochemical journal
Gong K,Xie J,Yi H,Li W
Tue May 01 00:00:00 EDT 2012
CS055 (Chidamide/HBI-8000), a novel histone deacetylase inhibitor, induces G1 arrest, ROS-dependent apoptosis and differentiation in human leukaemia cells.
The Biochemical journal
Gong K,Xie J,Yi H,Li W
Tue May 01 00:00:00 EDT 2012