Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MIPGGLSEA KPATPEIQEI VDKVKPQLEE KTNETYGKLE AVQYKTQVVA GTNYYIKVRA GDNKYMHLKV FKSLPGQNED LVLTGYQVDK NKDDELTGF (1-98 aa encoded by BC010379)
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Stefins have been proposed as prognostic and diagnostic tools for cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cystatin A; cystatin A (stefin A); cystatin AS; Cystatin-A; Cystatin-A1; Cystatin-AS; Stefin A; Stefin-A
Gene Aliases: AREI; CSTA; Csta1; PSS4; STF1; STFA
UniProt ID: (Human) P01040, (Mouse) P56567
Entrez Gene ID: (Human) 1475, (Mouse) 209294
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support