Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
In sandwich ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Cystatin D is observed and less than 2% cross-reactivity with rhCystatin S/A, rhCystatin A, recombinant mouse (rm) Cystatin B, rhCystatin S/N, and rmCystatin E/M is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AD8; cystatin 3; Cystatin-3; Cystatin-C; Cystatin3; CystatinC; MGC117328; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; RP23-250M11.9
Gene Aliases: Cst3; CysC
UniProt ID: (Mouse) P21460
Entrez Gene ID: (Mouse) 13010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support