Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Required for correct development of the embryonic midline glial cells which are necessary for the formation of distinct segmental commissures.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG10594; CG10594-PA; CG10594-PC; Cyp307a1; CYPCCCVIIA1; Cytochrome P450 307a1; Protein spook; spo; spo-PA; spo-PC
Gene Aliases: 307a1; CG10594; CYP307A1; CYP307A1-Dm; Dmel\CG10594; Dmel_CG10594; Dmspo; Spo
UniProt ID: (Fruit fly) Q9VRM7
Entrez Gene ID: (Fruit fly) 38631
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support