Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEQSDEAV KYYTLEEIQK HNHSKSTWLI LHHKVYDLTK FLEEHPGGEE VLREQAGGDA TENFEDVGHS TDAREMSKTF IIGELHPDDR PKLNKPPETL ITTIDSSSS (1-108 aa encoded by BC015182 )
Cytochrome b5 is a membrane-bound member of the cytochrome b family. A heme protein that functions as an electron carrier for many membrane-bound oxygenases, cytochrome b5 possesses two heme groups, which are not covalently attached to the protein. Two isoforms of cytochrome b5, a microsomal membrane-bound form and a cytoplasmic form, are produced by alternative splicing. Mutations in cytochrome b5 are associated with Leber's hereditary optic neuropathy and with myopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytochrome b-5; Cytochrome b5; cytochrome b5 type A (microsomal); MCB5; Microsomal cytochrome b5 type A; type 1 cyt-b5
Gene Aliases: 0610009N12Rik; CYB5; CYB5A; MCB5
UniProt ID: (Human) P00167, (Rat) P00173, (Mouse) P56395
Entrez Gene ID: (Human) 1528, (Rat) 64001, (Mouse) 109672
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support