Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Positive test controls include: HeLa, A-431.
Immunogen sequence: GGHGGGHGGS SGGGYGGGSS GGGSSGGGYG GGSSSGGHGG SSSGGYGGGS SGGGGGGYGG GSSGGGSSSG GGYGGGSSSG GHKSSSSGSV GESSSKGPRY
Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (i.e., skin) and non-keratinizing cells (i.e. mesothelial cells). Although not a traditional marker for endothelial cells, cytokeratins have also been found in some microvascular endothelial cells. At least 20 different cytokeratins (CK) in the molecular range of 40-70 kDa and isoelectric points of 5-8. 5 can be identified using two dimensional gel electrophoresis. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells. Monoclonal antibodies to cytokeratin proteins can be useful markers for tumor identification and classification.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 56 kDa cytokeratin; CK-10; cytokeratin 10; Cytokeratin-10; cytokeratin-17; epidermal keratin 10; intermediate filament protein; K10; keratin 10, type I; keratin complex 1, acidic, gene 10; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 59 kDa; Keratin-10; keratin-17; krt10 {ECO:0000250|UniProtKB:P13645}; suprabasal cytokeratin 10; type I keratin KA10
Gene Aliases: BCIE; BIE; CK10; D130054E02Rik; EHK; K10; K1C1; Ka10; KPP; Krt-1.10; Krt1-10; KRT10
UniProt ID: (Human) P13645, (Mouse) P02535
Entrez Gene ID: (Human) 3858, (Mouse) 16661, (Rat) 450225
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support