Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQNLNDRLA SYLDKVRALE EANADLEVKI RDWYQRQRPS EIKDYSPYFK TIEDLRNKII AATIENAQPI LQIDNARLAA DDFRTKYEHE LALRQTVEAD VNGLRRVLDE LTLARTDLEM QIEGLKEELA YLRKNHEEEM LALRGQTGGD VNVEMDAAPG VDLSRILNEM RDQYEQMAEK NRRDAETWFL SKTEELNKEV ASNSELVQSS RSEVTELRRV LQGLEIELQS QLSMKASLEN SLEETKGRYC MQLSQIQGLI GSVEEQLAQL RCEMEQQSQE YQILLDVKTR LEQEIATYRR LLEGEDAHLS SQQASGQSYS SREVFTSSSS SSSRQTRPIL KEQSSSSFSQ GQSS (121-473 aa encoded by B C039169)
KRT16 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region of chromosome 17q12-q21. This keratin has been coexpressed with keratin 14 in a number of epithelial tissues, including esophagus, tongue, and hair follicles. Mutations in this gene are associated with type 1 pachyonychia congenita, non-epidermolytic palmoplantar keratoderma and unilateral palmoplantar verrucous nevus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-16; cytokeratin 16; Cytokeratin-16; focal non-epidermolytic palmoplantar keratoderma; K16; keratin 16, type I; keratin complex 1, acidic, gene 16; keratin intermediate filament 16a; keratin intermediate filament 16b; Keratin, type I cytoskeletal 16; Keratin-16
Gene Aliases: AI324768; CK16; FNEPPK; K16; K1CP; Krt1-16; KRT16; KRT16A; NEPPK; PC1
UniProt ID: (Human) P08779, (Mouse) Q9Z2K1
Entrez Gene ID: (Human) 3868, (Mouse) 16666
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support