Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
This antibody, generated against full length KRT19 protein, also recognizes KRT17, another type I keratin homologous to KRT19.
Immunogen sequence: MTSYSYRQS SATSSFGGLG GGSVRFGPGV AFRAPSIHGG SGGRGVSVSS ARFVSSSSSG GYGGGYGGVL TASDGLLAGN EKLTMQNLND RLASYLDKVR ALEAANGELE VKIRDWYQKQ GPGPSRDYSH YYTTIQDLRD KILGATIENS RIVLQIDNAR LAADDFRTKF ETEQALRMSV EADINGLRRV LDELTLARTD LEMQIEGLKE ELAYLKKNHE EEISTLRGQV GGQVSVEVDS APGTDLAKIL SDMRSQYEVM AEQNRKDAEA WFTSRTEELN REVAGHTEQL QMSRSEVTDL RRTLQGLEIE LQSQLSMKAA LEDTLAETEA RFGAQLAHIQ ALISGIEAQL GDVRADSERQ NQEYQRLMDI KSRLEQEIAT YRSLLEGQED HYNNLSASKV L (1-400 aa encoded by BC002539)
Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40-kDa keratin intermediate filament; C K1CS; CK 19; CK-19; cytokeratin 19; Cytokeratin-19; K19; keratin 19, type I; keratin complex 1, acidic, gene 19; Keratin, type I cytoskeletal 19; keratin, type I, 40-kd; Keratin-19; MGC15366; Type I keratin Ka19
Gene Aliases: AI663979; CK19; EndoC; K19; K1CS; Ka19; Krt-1.19; Krt1-19; KRT19
UniProt ID: (Human) P08727, (Mouse) P19001, (Rat) Q63279
Entrez Gene ID: (Human) 3880, (Mouse) 16669, (Rat) 360626
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support