Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: IDIYCNLLDG EERKSKSTCY KSKGYRPVNS GNQAKDSTEE TIVKTVVEEL
Sequence homology: Human: 100%; Rabbit: 77%
The protein encoded by this gene is a member of the keratin superfamily. This keratin protein is a type I keratin that is specific for the inner root sheath of the hair follicle. This gene exists in a cluster with other keratin genes on chromosome 17q21.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-26; Cytokeratin-26; K25B; K26; keratin 25B; keratin 26, type I; Keratin, type I cytoskeletal 26; Keratin-25B; Keratin-26; KRT26; type I inner root sheath specific keratin 25 irs2; Type I inner root sheath-specific keratin-K25irs2
Gene Aliases: CK26; K25; K25IRS2; K26; KRT25B; KRT26
UniProt ID: (Human) Q7Z3Y9
Entrez Gene ID: (Human) 353288
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support